Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11796185-11796680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV6
Entrez Gene ID
2120
Gene Name
ETS variant 6
Gene Aliases
TEL, TEL/ABL, THC5
UniGene
Hs.504765
Gene Chromosome Location
Chr.12: 11649601-11895402 on Build GRCh38
Transcripts
NM_001987.4 XM_011520607.2 XM_011520608.2 XM_011520611.2 XM_017018990.1 XM_017018991.1

SNP Detail

SNP ID Chromosome Location
rs1056972463 Chr.12: 11796278-11796278 on Build GRCh38
rs536031919 Chr.12: 11796272-11796272 on Build GRCh38
rs866869676 Chr.12: 11796610-11796610 on Build GRCh38
rs780497130 Chr.12: 11796277-11796277 on Build GRCh38
rs953120181 Chr.12: 11796386-11796386 on Build GRCh38
rs115940849 Chr.12: 11796637-11796637 on Build GRCh38
rs1019238494 Chr.12: 11796484-11796484 on Build GRCh38
rs150262417 Chr.12: 11796509-11796509 on Build GRCh38
rs989084105 Chr.12: 11796462-11796462 on Build GRCh38
rs1015224194 Chr.12: 11796342-11796342 on Build GRCh38
rs749887038 Chr.12: 11796217-11796217 on Build GRCh38
rs1027146210 Chr.12: 11796376-11796376 on Build GRCh38
rs953769586 Chr.12: 11796460-11796460 on Build GRCh38
rs138950528 Chr.12: 11796614-11796614 on Build GRCh38
rs965000060 Chr.12: 11796658-11796658 on Build GRCh38
rs2723822 Chr.12: 11796568-11796568 on Build GRCh38
rs965618258 Chr.12: 11796503-11796503 on Build GRCh38
rs1036323442 Chr.12: 11796627-11796627 on Build GRCh38
rs985535190 Chr.12: 11796471-11796471 on Build GRCh38
rs1002769188 Chr.12: 11796316-11796316 on Build GRCh38
rs895662665 Chr.12: 11796612-11796612 on Build GRCh38
rs79350133 Chr.12: 11796635-11796635 on Build GRCh38
rs11831325 Chr.12: 11796651-11796651 on Build GRCh38
rs755383343 Chr.12: 11796371-11796371 on Build GRCh38
rs758830386 Chr.12: 11796256-11796256 on Build GRCh38
rs895757306 Chr.12: 11796313-11796313 on Build GRCh38
rs948126528 Chr.12: 11796556-11796556 on Build GRCh38
rs1004818538 Chr.12: 11796327-11796327 on Build GRCh38
rs924087671 Chr.12: 11796231-11796231 on Build GRCh38
rs937389496 Chr.12: 11796602-11796602 on Build GRCh38
rs965490578 Chr.12: 11796461-11796461 on Build GRCh38
rs938611192 Chr.12: 11796605-11796605 on Build GRCh38
rs558549998 Chr.12: 11796381-11796381 on Build GRCh38
rs936839924 Chr.12: 11796539-11796539 on Build GRCh38
rs916659632 Chr.12: 11796551-11796551 on Build GRCh38
rs750254684 Chr.12: 11796601-11796601 on Build GRCh38
rs940481795 Chr.12: 11796622-11796622 on Build GRCh38
rs1046358363 Chr.12: 11796286-11796286 on Build GRCh38
rs527533440 Chr.12: 11796650-11796650 on Build GRCh38
rs924136779 Chr.12: 11796537-11796537 on Build GRCh38
rs565372216 Chr.12: 11796628-11796628 on Build GRCh38
rs116687990 Chr.12: 11796273-11796273 on Build GRCh38
rs896979797 Chr.12: 11796357-11796357 on Build GRCh38
rs995329732 Chr.12: 11796632-11796632 on Build GRCh38
rs947341757 Chr.12: 11796218-11796218 on Build GRCh38
rs1019758680 Chr.12: 11796453-11796453 on Build GRCh38
rs115122458 Chr.12: 11796603-11796603 on Build GRCh38
rs1028598364 Chr.12: 11796633-11796633 on Build GRCh38
rs796197179 Chr.12: 11796659-11796659 on Build GRCh38
rs907119797 Chr.12: 11796308-11796308 on Build GRCh38
rs770043823 Chr.12: 11796640-11796640 on Build GRCh38
rs188044975 Chr.12: 11796353-11796353 on Build GRCh38
rs142559925 Chr.12: 11796351-11796351 on Build GRCh38
rs985617431 Chr.12: 11796427-11796427 on Build GRCh38
rs936769721 Chr.12: 11796246-11796246 on Build GRCh38
rs576901557 Chr.12: 11796559-11796559 on Build GRCh38
rs978284191 Chr.12: 11796519-11796519 on Build GRCh38
rs989614774 Chr.12: 11796545-11796545 on Build GRCh38
rs994909451 Chr.12: 11796354-11796354 on Build GRCh38

View Full Product Details