Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48771848-48772348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY6
Entrez Gene ID
112
Gene Name
adenylate cyclase 6
Gene Aliases
AC6, LCCS8
UniGene
Hs.525401
Gene Chromosome Location
Chr.12: 48766191-48789096 on Build GRCh38
Transcripts
NM_015270.4 XM_006719210.3 XM_017018743.1 NR_039850.2

Gene Symbol
MIR4701
Entrez Gene ID
100616262
Gene Name
microRNA 4701
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.12: 48771975-48772037 on Build GRCh38
Transcripts
NM_015270.4 XM_006719210.3 XM_017018743.1 NR_039850.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4659929 c.2896C>T p.R966C Chr.12: 48771865-48771865 on Build GRCh38
4659930 c.2759C>T p.A920V Chr.12: 48772323-48772323 on Build GRCh38
5495439 c.2820A>T p.L940L Chr.12: 48771941-48771941 on Build GRCh38
7088013 c.2900A>G p.N967S Chr.12: 48771861-48771861 on Build GRCh38
1676924 c.2869G>A p.A957T Chr.12: 48771892-48771892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752304189 Chr.12: 48772029-48772029 on Build GRCh38
rs754462882 Chr.12: 48771907-48771907 on Build GRCh38
rs906075876 Chr.12: 48771912-48771912 on Build GRCh38
rs377559275 Chr.12: 48772021-48772021 on Build GRCh38
rs767887910 Chr.12: 48771997-48771997 on Build GRCh38
rs746847957 Chr.12: 48771962-48771962 on Build GRCh38
rs755219383 Chr.12: 48771967-48771968 on Build GRCh38
rs778563944 Chr.12: 48771927-48771927 on Build GRCh38
rs187203937 Chr.12: 48772109-48772109 on Build GRCh38
rs750808509 Chr.12: 48771999-48771999 on Build GRCh38
rs756192807 Chr.12: 48772063-48772064 on Build GRCh38
rs374254371 Chr.12: 48772087-48772087 on Build GRCh38
rs953933030 Chr.12: 48772239-48772239 on Build GRCh38
rs549003118 Chr.12: 48772092-48772092 on Build GRCh38
rs757059005 Chr.12: 48772051-48772051 on Build GRCh38
rs777523139 Chr.12: 48771944-48771944 on Build GRCh38
rs748323230 Chr.12: 48772080-48772080 on Build GRCh38
rs774582629 Chr.12: 48771979-48771979 on Build GRCh38
rs750863326 Chr.12: 48771884-48771884 on Build GRCh38
rs200061969 Chr.12: 48772320-48772320 on Build GRCh38
rs200640184 Chr.12: 48771968-48771968 on Build GRCh38
rs986329942 Chr.12: 48772221-48772221 on Build GRCh38
rs747790458 Chr.12: 48771932-48771932 on Build GRCh38
rs756995528 Chr.12: 48772328-48772328 on Build GRCh38
rs1030326903 Chr.12: 48772175-48772175 on Build GRCh38
rs778215409 Chr.12: 48772070-48772073 on Build GRCh38
rs753383547 Chr.12: 48771896-48771896 on Build GRCh38
rs766883993 Chr.12: 48772267-48772267 on Build GRCh38
rs745814578 Chr.12: 48772061-48772061 on Build GRCh38
rs770852283 Chr.12: 48771963-48771963 on Build GRCh38
rs759941976 Chr.12: 48772275-48772275 on Build GRCh38
rs777151176 Chr.12: 48772268-48772268 on Build GRCh38
rs751732732 Chr.12: 48771873-48771873 on Build GRCh38
rs754474561 Chr.12: 48772008-48772008 on Build GRCh38
rs771856644 Chr.12: 48771935-48771935 on Build GRCh38
rs1013320636 Chr.12: 48772023-48772023 on Build GRCh38
rs2291727 Chr.12: 48772216-48772216 on Build GRCh38
rs763680788 Chr.12: 48772289-48772289 on Build GRCh38
rs12427399 Chr.12: 48772144-48772144 on Build GRCh38
rs897307392 Chr.12: 48771880-48771880 on Build GRCh38
rs1010022062 Chr.12: 48772160-48772160 on Build GRCh38
rs892469615 Chr.12: 48772153-48772153 on Build GRCh38
rs563129186 Chr.12: 48771876-48771876 on Build GRCh38
rs761056051 Chr.12: 48772252-48772252 on Build GRCh38
rs761918018 Chr.12: 48771994-48771994 on Build GRCh38
rs918705602 Chr.12: 48772303-48772303 on Build GRCh38
rs371352626 Chr.12: 48771890-48771890 on Build GRCh38
rs781331177 Chr.12: 48772001-48772004 on Build GRCh38
rs1000968505 Chr.12: 48772116-48772116 on Build GRCh38
rs761176427 Chr.12: 48772002-48772002 on Build GRCh38
rs3729970 Chr.12: 48772276-48772276 on Build GRCh38
rs756654985 Chr.12: 48771889-48771889 on Build GRCh38
rs754307704 Chr.12: 48772005-48772005 on Build GRCh38
rs774259312 Chr.12: 48772072-48772072 on Build GRCh38
rs777470134 Chr.12: 48772048-48772048 on Build GRCh38
rs781065644 Chr.12: 48772058-48772058 on Build GRCh38
rs752768128 Chr.12: 48772011-48772012 on Build GRCh38
rs112212130 Chr.12: 48772251-48772251 on Build GRCh38
rs776609767 Chr.12: 48771964-48771964 on Build GRCh38
rs1032807037 Chr.12: 48772125-48772125 on Build GRCh38
rs201896260 Chr.12: 48772280-48772280 on Build GRCh38
rs535357432 Chr.12: 48772184-48772184 on Build GRCh38
rs113434461 Chr.12: 48772286-48772286 on Build GRCh38
rs769818987 Chr.12: 48772068-48772068 on Build GRCh38
rs758015419 Chr.12: 48772033-48772033 on Build GRCh38
rs879124998 Chr.12: 48772057-48772057 on Build GRCh38
rs963483143 Chr.12: 48772250-48772250 on Build GRCh38
rs368945637 Chr.12: 48771874-48771874 on Build GRCh38
rs772406411 Chr.12: 48772248-48772248 on Build GRCh38
rs147278695 Chr.12: 48771965-48771965 on Build GRCh38
rs369144920 Chr.12: 48772004-48772004 on Build GRCh38
rs115351639 Chr.12: 48772321-48772321 on Build GRCh38
rs538084003 Chr.12: 48771983-48771983 on Build GRCh38
rs751347638 Chr.12: 48771912-48771913 on Build GRCh38

View Full Product Details