Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64900437-64900939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ41278
Entrez Gene ID
400046
Gene Name
uncharacterized LOC400046
Gene Aliases
-
UniGene
Hs.591032
Gene Chromosome Location
Chr.12: 64883774-64977522 on Build GRCh38
Transcripts
NR_033988.1

SNP Detail

SNP ID Chromosome Location
rs919208296 Chr.12: 64900689-64900689 on Build GRCh38
rs12231352 Chr.12: 64900747-64900747 on Build GRCh38
rs981001839 Chr.12: 64900632-64900632 on Build GRCh38
rs28524936 Chr.12: 64900871-64900871 on Build GRCh38
rs904754814 Chr.12: 64900570-64900570 on Build GRCh38
rs534007933 Chr.12: 64900492-64900492 on Build GRCh38
rs755841738 Chr.12: 64900839-64900839 on Build GRCh38
rs374683075 Chr.12: 64900481-64900481 on Build GRCh38
rs572031659 Chr.12: 64900668-64900668 on Build GRCh38
rs73327250 Chr.12: 64900488-64900488 on Build GRCh38
rs368986809 Chr.12: 64900615-64900615 on Build GRCh38
rs941904263 Chr.12: 64900639-64900639 on Build GRCh38
rs187981769 Chr.12: 64900521-64900521 on Build GRCh38
rs941958083 Chr.12: 64900516-64900516 on Build GRCh38
rs999080557 Chr.12: 64900575-64900575 on Build GRCh38
rs763844638 Chr.12: 64900908-64900908 on Build GRCh38
rs536451905 Chr.12: 64900522-64900522 on Build GRCh38
rs1144416 Chr.12: 64900519-64900519 on Build GRCh38
rs556491850 Chr.12: 64900585-64900585 on Build GRCh38
rs1051793149 Chr.12: 64900741-64900741 on Build GRCh38
rs904701709 Chr.12: 64900841-64900841 on Build GRCh38
rs999028270 Chr.12: 64900864-64900864 on Build GRCh38
rs937731656 Chr.12: 64900826-64900826 on Build GRCh38
rs1031877898 Chr.12: 64900867-64900867 on Build GRCh38
rs1008120102 Chr.12: 64900544-64900544 on Build GRCh38
rs372307662 Chr.12: 64900467-64900467 on Build GRCh38
rs139842161 Chr.12: 64900658-64900658 on Build GRCh38
rs551395341 Chr.12: 64900485-64900485 on Build GRCh38
rs41434944 Chr.12: 64900633-64900633 on Build GRCh38
rs761995141 Chr.12: 64900558-64900558 on Build GRCh38
rs767486076 Chr.12: 64900663-64900663 on Build GRCh38
rs917762301 Chr.12: 64900860-64900860 on Build GRCh38
rs933304742 Chr.12: 64900710-64900710 on Build GRCh38
rs943479220 Chr.12: 64900784-64900784 on Build GRCh38
rs896434271 Chr.12: 64900540-64900540 on Build GRCh38
rs956382577 Chr.12: 64900603-64900603 on Build GRCh38
rs1021843993 Chr.12: 64900782-64900782 on Build GRCh38
rs1052272728 Chr.12: 64900524-64900524 on Build GRCh38
rs992017737 Chr.12: 64900849-64900849 on Build GRCh38
rs896112189 Chr.12: 64900870-64900870 on Build GRCh38
rs79756910 Chr.12: 64900785-64900785 on Build GRCh38
rs1036336897 Chr.12: 64900517-64900517 on Build GRCh38
rs1030584348 Chr.12: 64900576-64900576 on Build GRCh38
rs192493594 Chr.12: 64900894-64900894 on Build GRCh38
rs549508752 Chr.12: 64900736-64900736 on Build GRCh38
rs1025142527 Chr.12: 64900625-64900625 on Build GRCh38
rs889243086 Chr.12: 64900527-64900527 on Build GRCh38
rs756995298 Chr.12: 64900627-64900627 on Build GRCh38
rs765264757 Chr.12: 64900602-64900602 on Build GRCh38

View Full Product Details