Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64975587-64976085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ41278
Entrez Gene ID
400046
Gene Name
uncharacterized LOC400046
Gene Aliases
-
UniGene
Hs.591032
Gene Chromosome Location
Chr.12: 64883774-64977522 on Build GRCh38
Transcripts
NR_033988.1

SNP Detail

SNP ID Chromosome Location
rs897533392 Chr.12: 64975671-64975671 on Build GRCh38
rs545823474 Chr.12: 64976051-64976051 on Build GRCh38
rs941315112 Chr.12: 64976006-64976006 on Build GRCh38
rs998733798 Chr.12: 64975876-64975876 on Build GRCh38
rs983052102 Chr.12: 64975959-64975959 on Build GRCh38
rs933074897 Chr.12: 64975673-64975673 on Build GRCh38
rs540190931 Chr.12: 64976052-64976052 on Build GRCh38
rs910052972 Chr.12: 64975992-64975992 on Build GRCh38
rs980403419 Chr.12: 64975880-64975880 on Build GRCh38
rs189367665 Chr.12: 64975914-64975914 on Build GRCh38
rs990411317 Chr.12: 64975773-64975773 on Build GRCh38
rs973014562 Chr.12: 64976050-64976050 on Build GRCh38
rs183792735 Chr.12: 64975856-64975856 on Build GRCh38
rs777529362 Chr.12: 64975812-64975812 on Build GRCh38
rs1038810351 Chr.12: 64975646-64975646 on Build GRCh38
rs1031825376 Chr.12: 64975731-64975731 on Build GRCh38
rs7960921 Chr.12: 64975965-64975965 on Build GRCh38
rs1007779865 Chr.12: 64975803-64975803 on Build GRCh38
rs79809040 Chr.12: 64976043-64976043 on Build GRCh38
rs957891626 Chr.12: 64975770-64975770 on Build GRCh38
rs572220219 Chr.12: 64975709-64975709 on Build GRCh38
rs574657959 Chr.12: 64975890-64975890 on Build GRCh38
rs992913881 Chr.12: 64975934-64975934 on Build GRCh38
rs941277808 Chr.12: 64975616-64975616 on Build GRCh38
rs145063198 Chr.12: 64976060-64976060 on Build GRCh38
rs1038294298 Chr.12: 64976024-64976024 on Build GRCh38
rs888935287 Chr.12: 64975730-64975730 on Build GRCh38
rs1023597078 Chr.12: 64975794-64975794 on Build GRCh38
rs970187567 Chr.12: 64975984-64975984 on Build GRCh38
rs899829747 Chr.12: 64975661-64975661 on Build GRCh38
rs534811811 Chr.12: 64975713-64975713 on Build GRCh38
rs984660599 Chr.12: 64975985-64975985 on Build GRCh38

View Full Product Details