Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109742007-109742509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222A
Entrez Gene ID
84915
Gene Name
family with sequence similarity 222 member A
Gene Aliases
C12orf34
UniGene
Hs.661785
Gene Chromosome Location
Chr.12: 109714382-109770509 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

Gene Symbol
FAM222A-AS1
Entrez Gene ID
84983
Gene Name
FAM222A antisense RNA 1
Gene Aliases
-
UniGene
Hs.675961
Gene Chromosome Location
Chr.12: 109734209-109773487 on Build GRCh38
Transcripts
NM_032829.2 XM_006719654.2 XM_017020055.1 NR_026661.2 NR_026662.2

SNP Detail

SNP ID Chromosome Location
rs938337957 Chr.12: 109742135-109742135 on Build GRCh38
rs1011682493 Chr.12: 109742153-109742153 on Build GRCh38
rs938617844 Chr.12: 109742344-109742344 on Build GRCh38
rs1002811816 Chr.12: 109742212-109742212 on Build GRCh38
rs1008147496 Chr.12: 109742459-109742459 on Build GRCh38
rs991566426 Chr.12: 109742309-109742309 on Build GRCh38
rs966231891 Chr.12: 109742239-109742239 on Build GRCh38
rs1041754488 Chr.12: 109742469-109742469 on Build GRCh38
rs958537857 Chr.12: 109742305-109742305 on Build GRCh38
rs1026308178 Chr.12: 109742271-109742271 on Build GRCh38
rs537279925 Chr.12: 109742098-109742098 on Build GRCh38
rs970311230 Chr.12: 109742315-109742315 on Build GRCh38
rs565062456 Chr.12: 109742466-109742466 on Build GRCh38
rs371932060 Chr.12: 109742213-109742213 on Build GRCh38
rs942418838 Chr.12: 109742109-109742109 on Build GRCh38
rs749948783 Chr.12: 109742332-109742332 on Build GRCh38
rs943580112 Chr.12: 109742415-109742415 on Build GRCh38
rs533627709 Chr.12: 109742164-109742164 on Build GRCh38
rs558703155 Chr.12: 109742125-109742125 on Build GRCh38
rs909276057 Chr.12: 109742310-109742310 on Build GRCh38
rs903071011 Chr.12: 109742188-109742188 on Build GRCh38
rs1051855692 Chr.12: 109742062-109742062 on Build GRCh38
rs553105134 Chr.12: 109742269-109742269 on Build GRCh38
rs765349095 Chr.12: 109742385-109742385 on Build GRCh38
rs528879247 Chr.12: 109742085-109742085 on Build GRCh38
rs985304505 Chr.12: 109742296-109742296 on Build GRCh38
rs375693314 Chr.12: 109742268-109742268 on Build GRCh38
rs4766627 Chr.12: 109742346-109742346 on Build GRCh38
rs902013380 Chr.12: 109742475-109742475 on Build GRCh38
rs574732734 Chr.12: 109742316-109742316 on Build GRCh38
rs764000323 Chr.12: 109742146-109742146 on Build GRCh38
rs1052733494 Chr.12: 109742140-109742140 on Build GRCh38
rs891463111 Chr.12: 109742145-109742145 on Build GRCh38
rs947015074 Chr.12: 109742359-109742359 on Build GRCh38
rs557393159 Chr.12: 109742372-109742372 on Build GRCh38
rs762954921 Chr.12: 109742142-109742142 on Build GRCh38
rs892906474 Chr.12: 109742430-109742430 on Build GRCh38
rs17616406 Chr.12: 109742414-109742414 on Build GRCh38
rs1035485384 Chr.12: 109742241-109742241 on Build GRCh38
rs1039411275 Chr.12: 109742428-109742428 on Build GRCh38
rs912928197 Chr.12: 109742357-109742357 on Build GRCh38
rs780643316 Chr.12: 109742205-109742205 on Build GRCh38
rs751300756 Chr.12: 109742154-109742154 on Build GRCh38
rs752646144 Chr.12: 109742100-109742100 on Build GRCh38
rs570854412 Chr.12: 109742210-109742210 on Build GRCh38
rs933497847 Chr.12: 109742078-109742078 on Build GRCh38
rs950707586 Chr.12: 109742283-109742283 on Build GRCh38
rs1018286216 Chr.12: 109742220-109742220 on Build GRCh38
rs543672173 Chr.12: 109742465-109742465 on Build GRCh38
rs928547338 Chr.12: 109742333-109742333 on Build GRCh38

View Full Product Details