Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49982437-49982946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU2
Entrez Gene ID
8847
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
Gene Aliases
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS
UniGene
Hs.547964
Gene Chromosome Location
Chr.13: 49982552-50125541 on Build GRCh38
Transcripts
NR_002612.1

SNP Detail

SNP ID Chromosome Location
rs546353784 Chr.13: 49982694-49982694 on Build GRCh38
rs554920667 Chr.13: 49982707-49982707 on Build GRCh38
rs905383573 Chr.13: 49982557-49982557 on Build GRCh38
rs944663019 Chr.13: 49982547-49982547 on Build GRCh38
rs575208641 Chr.13: 49982923-49982923 on Build GRCh38
rs561844650 Chr.13: 49982766-49982766 on Build GRCh38
rs986803729 Chr.13: 49982907-49982907 on Build GRCh38
rs1036468294 Chr.13: 49982575-49982575 on Build GRCh38
rs150476347 Chr.13: 49982736-49982736 on Build GRCh38
rs558162066 Chr.13: 49982696-49982696 on Build GRCh38
rs1050695192 Chr.13: 49982468-49982468 on Build GRCh38
rs921168651 Chr.13: 49982727-49982727 on Build GRCh38
rs890492244 Chr.13: 49982523-49982523 on Build GRCh38
rs989906175 Chr.13: 49982768-49982768 on Build GRCh38
rs987037177 Chr.13: 49982749-49982749 on Build GRCh38
rs764824666 Chr.13: 49982834-49982834 on Build GRCh38
rs375079190 Chr.13: 49982820-49982820 on Build GRCh38
rs1052502243 Chr.13: 49982859-49982859 on Build GRCh38
rs544075390 Chr.13: 49982798-49982798 on Build GRCh38
rs1011175932 Chr.13: 49982510-49982510 on Build GRCh38
rs760295247 Chr.13: 49982490-49982490 on Build GRCh38
rs866315533 Chr.13: 49982822-49982822 on Build GRCh38
rs189314369 Chr.13: 49982738-49982738 on Build GRCh38
rs181219087 Chr.13: 49982915-49982915 on Build GRCh38
rs867132681 Chr.13: 49982656-49982656 on Build GRCh38
rs184542033 Chr.13: 49982635-49982635 on Build GRCh38
rs1034808821 Chr.13: 49982598-49982598 on Build GRCh38
rs971051002 Chr.13: 49982718-49982718 on Build GRCh38
rs897874944 Chr.13: 49982628-49982628 on Build GRCh38
rs1027936553 Chr.13: 49982691-49982691 on Build GRCh38
rs776108589 Chr.13: 49982593-49982593 on Build GRCh38
rs1024211407 Chr.13: 49982713-49982713 on Build GRCh38
rs1031385406 Chr.13: 49982737-49982737 on Build GRCh38
rs768378848 Chr.13: 49982568-49982568 on Build GRCh38
rs1045317871 Chr.13: 49982531-49982531 on Build GRCh38
rs953551355 Chr.13: 49982874-49982874 on Build GRCh38
rs537537676 Chr.13: 49982548-49982548 on Build GRCh38
rs571915643 Chr.13: 49982461-49982461 on Build GRCh38
rs535337591 Chr.13: 49982693-49982693 on Build GRCh38
rs557578447 Chr.13: 49982646-49982646 on Build GRCh38
rs78118837 Chr.13: 49982727-49982727 on Build GRCh38
rs529038839 Chr.13: 49982790-49982790 on Build GRCh38
rs893777693 Chr.13: 49982489-49982489 on Build GRCh38
rs781634335 Chr.13: 49982475-49982475 on Build GRCh38

View Full Product Details