Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95496898-95497401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_182848.3 XM_017020843.1 XM_017020844.1 NR_046533.1

Gene Symbol
CLDN10-AS1
Entrez Gene ID
100874194
Gene Name
CLDN10 antisense RNA 1
Gene Aliases
-
UniGene
Hs.569229
Gene Chromosome Location
Chr.13: 95479444-95533910 on Build GRCh38
Transcripts
NM_001160100.1 NM_182848.3 XM_017020843.1 XM_017020844.1 NR_046533.1

SNP Detail

SNP ID Chromosome Location
rs931363859 Chr.13: 95497021-95497021 on Build GRCh38
rs912501801 Chr.13: 95497176-95497176 on Build GRCh38
rs968539198 Chr.13: 95497037-95497037 on Build GRCh38
rs1025254123 Chr.13: 95497079-95497079 on Build GRCh38
rs959688793 Chr.13: 95497072-95497072 on Build GRCh38
rs775785158 Chr.13: 95497334-95497334 on Build GRCh38
rs985670909 Chr.13: 95497154-95497154 on Build GRCh38
rs932475615 Chr.13: 95496935-95496935 on Build GRCh38
rs571009471 Chr.13: 95497208-95497208 on Build GRCh38
rs890430518 Chr.13: 95496954-95496954 on Build GRCh38
rs549884433 Chr.13: 95497236-95497236 on Build GRCh38
rs1031301189 Chr.13: 95497105-95497105 on Build GRCh38
rs867239416 Chr.13: 95497001-95497001 on Build GRCh38
rs985420120 Chr.13: 95496965-95496965 on Build GRCh38
rs915643177 Chr.13: 95497081-95497081 on Build GRCh38
rs912458876 Chr.13: 95496986-95496986 on Build GRCh38
rs116879212 Chr.13: 95497317-95497317 on Build GRCh38
rs890750559 Chr.13: 95497190-95497190 on Build GRCh38
rs895354669 Chr.13: 95497051-95497051 on Build GRCh38
rs924538791 Chr.13: 95497041-95497041 on Build GRCh38
rs749286228 Chr.13: 95497101-95497101 on Build GRCh38
rs563654638 Chr.13: 95497080-95497080 on Build GRCh38
rs543681044 Chr.13: 95497043-95497043 on Build GRCh38
rs1021368551 Chr.13: 95497119-95497119 on Build GRCh38
rs1046974004 Chr.13: 95497369-95497369 on Build GRCh38
rs138318707 Chr.13: 95497115-95497115 on Build GRCh38
rs972717655 Chr.13: 95497201-95497201 on Build GRCh38
rs377536190 Chr.13: 95497372-95497372 on Build GRCh38
rs968489093 Chr.13: 95497303-95497303 on Build GRCh38
rs577896154 Chr.13: 95497040-95497040 on Build GRCh38
rs948502608 Chr.13: 95497095-95497095 on Build GRCh38
rs754767835 Chr.13: 95497106-95497106 on Build GRCh38
rs1008936738 Chr.13: 95496992-95496992 on Build GRCh38
rs564845005 Chr.13: 95497224-95497224 on Build GRCh38
rs775286457 Chr.13: 95497217-95497219 on Build GRCh38
rs1021847501 Chr.13: 95497029-95497029 on Build GRCh38
rs955780656 Chr.13: 95497107-95497107 on Build GRCh38
rs897249123 Chr.13: 95497020-95497020 on Build GRCh38
rs944068560 Chr.13: 95496988-95496988 on Build GRCh38
rs906750060 Chr.13: 95497096-95497096 on Build GRCh38
rs149625529 Chr.13: 95497198-95497198 on Build GRCh38
rs540979322 Chr.13: 95496944-95496944 on Build GRCh38
rs771101228 Chr.13: 95497169-95497169 on Build GRCh38
rs1022380322 Chr.13: 95497255-95497255 on Build GRCh38
rs948259672 Chr.13: 95497367-95497367 on Build GRCh38
rs1012810344 Chr.13: 95497063-95497063 on Build GRCh38
rs1011383582 Chr.13: 95497250-95497254 on Build GRCh38
rs543252336 Chr.13: 95497120-95497120 on Build GRCh38
rs138179094 Chr.13: 95497127-95497128 on Build GRCh38
rs559174038 Chr.13: 95497180-95497180 on Build GRCh38

View Full Product Details