Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95533710-95534243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_182848.3 XM_017020843.1 XM_017020844.1 NR_046533.1

Gene Symbol
CLDN10-AS1
Entrez Gene ID
100874194
Gene Name
CLDN10 antisense RNA 1
Gene Aliases
-
UniGene
Hs.569229
Gene Chromosome Location
Chr.13: 95479444-95533910 on Build GRCh38
Transcripts
NM_001160100.1 NM_182848.3 XM_017020843.1 XM_017020844.1 NR_046533.1

SNP Detail

SNP ID Chromosome Location
rs954406432 Chr.13: 95533989-95533989 on Build GRCh38
rs982739169 Chr.13: 95534202-95534202 on Build GRCh38
rs995844545 Chr.13: 95534212-95534212 on Build GRCh38
rs143848622 Chr.13: 95534200-95534200 on Build GRCh38
rs536075237 Chr.13: 95534017-95534017 on Build GRCh38
rs529541705 Chr.13: 95533866-95533866 on Build GRCh38
rs550247488 Chr.13: 95533806-95533806 on Build GRCh38
rs927231786 Chr.13: 95534065-95534065 on Build GRCh38
rs796646233 Chr.13: 95533901-95533901 on Build GRCh38
rs774435715 Chr.13: 95533756-95533756 on Build GRCh38
rs572162860 Chr.13: 95534041-95534041 on Build GRCh38
rs73547024 Chr.13: 95533788-95533788 on Build GRCh38
rs541176992 Chr.13: 95534091-95534091 on Build GRCh38
rs936180647 Chr.13: 95534176-95534176 on Build GRCh38
rs941919899 Chr.13: 95533830-95533830 on Build GRCh38
rs41277664 Chr.13: 95533962-95533962 on Build GRCh38
rs981024370 Chr.13: 95533997-95533997 on Build GRCh38
rs956479788 Chr.13: 95533949-95533949 on Build GRCh38
rs564327010 Chr.13: 95533767-95533767 on Build GRCh38
rs549086581 Chr.13: 95533904-95533904 on Build GRCh38
rs901751223 Chr.13: 95533867-95533867 on Build GRCh38
rs900048232 Chr.13: 95534206-95534206 on Build GRCh38
rs182209544 Chr.13: 95534008-95534008 on Build GRCh38
rs1025165422 Chr.13: 95534153-95534153 on Build GRCh38
rs887221592 Chr.13: 95533850-95533850 on Build GRCh38
rs919900135 Chr.13: 95533781-95533781 on Build GRCh38
rs907796555 Chr.13: 95533814-95533814 on Build GRCh38
rs904783863 Chr.13: 95533916-95533916 on Build GRCh38
rs969556386 Chr.13: 95534169-95534169 on Build GRCh38
rs551307361 Chr.13: 95533914-95533914 on Build GRCh38
rs552773004 Chr.13: 95534040-95534040 on Build GRCh38
rs558123084 Chr.13: 95534100-95534100 on Build GRCh38
rs1000881907 Chr.13: 95533935-95533935 on Build GRCh38
rs946103948 Chr.13: 95534108-95534108 on Build GRCh38
rs577997496 Chr.13: 95534110-95534110 on Build GRCh38
rs534440309 Chr.13: 95533911-95533911 on Build GRCh38
rs61734428 Chr.13: 95533908-95533908 on Build GRCh38
rs1050351795 Chr.13: 95534079-95534079 on Build GRCh38
rs953005524 Chr.13: 95534201-95534201 on Build GRCh38
rs920194317 Chr.13: 95534007-95534007 on Build GRCh38
rs904710833 Chr.13: 95534149-95534149 on Build GRCh38
rs995834693 Chr.13: 95533964-95533964 on Build GRCh38
rs866842520 Chr.13: 95534133-95534133 on Build GRCh38
rs967676610 Chr.13: 95533926-95533926 on Build GRCh38
rs1018509461 Chr.13: 95533936-95533936 on Build GRCh38
rs927314784 Chr.13: 95533855-95533855 on Build GRCh38
rs1041844043 Chr.13: 95533915-95533915 on Build GRCh38
rs761096540 Chr.13: 95534095-95534095 on Build GRCh38
rs189212537 Chr.13: 95533817-95533817 on Build GRCh38
rs61736021 Chr.13: 95533733-95533733 on Build GRCh38
rs1030092502 Chr.13: 95533981-95533981 on Build GRCh38
rs556882808 Chr.13: 95533975-95533975 on Build GRCh38
rs1030705652 Chr.13: 95534132-95534132 on Build GRCh38
rs907673181 Chr.13: 95534024-95534024 on Build GRCh38
rs901652743 Chr.13: 95534048-95534048 on Build GRCh38
rs571162425 Chr.13: 95533941-95533941 on Build GRCh38
rs931820040 Chr.13: 95534050-95534050 on Build GRCh38
rs386773607 Chr.13: 95534040-95534041 on Build GRCh38
rs992726912 Chr.13: 95533961-95533961 on Build GRCh38
rs1037721357 Chr.13: 95533833-95533833 on Build GRCh38

View Full Product Details