Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21392041-21392544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD8
Entrez Gene ID
57680
Gene Name
chromodomain helicase DNA binding protein 8
Gene Aliases
AUTS18, HELSNF1
UniGene
Hs.530698
Gene Chromosome Location
Chr.14: 21385194-21437298 on Build GRCh38
Transcripts
NM_001170629.1 NM_020920.3 NR_003029.2

Gene Symbol
SNORD9
Entrez Gene ID
692053
Gene Name
small nucleolar RNA, C/D box 9
Gene Aliases
mgU6-53B
UniGene
Hs.675950
Gene Chromosome Location
Chr.14: 21392151-21392253 on Build GRCh38
Transcripts
NM_001170629.1 NM_020920.3 NR_003029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6583099 c.5934+1G>T p.? Chr.14: 21392506-21392506 on Build GRCh38
6075353 c.6769G>C p.D2257H Chr.14: 21392509-21392509 on Build GRCh38
6001100 c.6742G>T p.E2248* Chr.14: 21392536-21392536 on Build GRCh38
6075352 c.5932G>C p.D1978H Chr.14: 21392509-21392509 on Build GRCh38
6583100 c.6771+1G>T p.? Chr.14: 21392506-21392506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766216751 Chr.14: 21392282-21392282 on Build GRCh38
rs759593522 Chr.14: 21392470-21392470 on Build GRCh38
rs901063076 Chr.14: 21392429-21392429 on Build GRCh38
rs894725097 Chr.14: 21392499-21392499 on Build GRCh38
rs928196951 Chr.14: 21392252-21392252 on Build GRCh38
rs756832960 Chr.14: 21392112-21392112 on Build GRCh38
rs746023449 Chr.14: 21392293-21392294 on Build GRCh38
rs767928606 Chr.14: 21392235-21392235 on Build GRCh38
rs185676637 Chr.14: 21392337-21392337 on Build GRCh38
rs1048120920 Chr.14: 21392317-21392317 on Build GRCh38
rs376064377 Chr.14: 21392114-21392114 on Build GRCh38
rs374528240 Chr.14: 21392265-21392265 on Build GRCh38
rs749597133 Chr.14: 21392167-21392170 on Build GRCh38
rs139558741 Chr.14: 21392108-21392108 on Build GRCh38
rs1021755285 Chr.14: 21392167-21392167 on Build GRCh38
rs938313731 Chr.14: 21392358-21392358 on Build GRCh38
rs751445294 Chr.14: 21392283-21392283 on Build GRCh38
rs894674911 Chr.14: 21392120-21392120 on Build GRCh38
rs986711139 Chr.14: 21392280-21392280 on Build GRCh38
rs954857843 Chr.14: 21392245-21392245 on Build GRCh38
rs72684751 Chr.14: 21392335-21392335 on Build GRCh38
rs965305781 Chr.14: 21392145-21392145 on Build GRCh38
rs550938546 Chr.14: 21392080-21392080 on Build GRCh38
rs776881930 Chr.14: 21392375-21392375 on Build GRCh38
rs777956601 Chr.14: 21392465-21392465 on Build GRCh38
rs771318761 Chr.14: 21392225-21392225 on Build GRCh38
rs776437407 Chr.14: 21392237-21392237 on Build GRCh38
rs746753327 Chr.14: 21392064-21392064 on Build GRCh38
rs201511497 Chr.14: 21392509-21392509 on Build GRCh38
rs764875493 Chr.14: 21392259-21392259 on Build GRCh38
rs771478747 Chr.14: 21392203-21392203 on Build GRCh38
rs996788249 Chr.14: 21392452-21392452 on Build GRCh38
rs982872215 Chr.14: 21392334-21392334 on Build GRCh38
rs989597002 Chr.14: 21392184-21392184 on Build GRCh38
rs754779015 Chr.14: 21392290-21392290 on Build GRCh38
rs192489152 Chr.14: 21392179-21392179 on Build GRCh38
rs749840472 Chr.14: 21392222-21392222 on Build GRCh38
rs373765021 Chr.14: 21392158-21392158 on Build GRCh38
rs973838814 Chr.14: 21392311-21392311 on Build GRCh38
rs755258387 Chr.14: 21392172-21392172 on Build GRCh38
rs914010402 Chr.14: 21392189-21392189 on Build GRCh38
rs370488043 Chr.14: 21392256-21392256 on Build GRCh38
rs377437025 Chr.14: 21392234-21392234 on Build GRCh38
rs543559054 Chr.14: 21392291-21392291 on Build GRCh38
rs919678815 Chr.14: 21392324-21392324 on Build GRCh38
rs543459682 Chr.14: 21392238-21392238 on Build GRCh38
rs1046695507 Chr.14: 21392082-21392082 on Build GRCh38
rs764511403 Chr.14: 21392249-21392250 on Build GRCh38
rs938921833 Chr.14: 21392101-21392101 on Build GRCh38
rs187908001 Chr.14: 21392165-21392165 on Build GRCh38
rs920793611 Chr.14: 21392307-21392307 on Build GRCh38
rs776582187 Chr.14: 21392226-21392226 on Build GRCh38
rs749313449 Chr.14: 21392477-21392477 on Build GRCh38
rs930831698 Chr.14: 21392309-21392309 on Build GRCh38
rs781394216 Chr.14: 21392199-21392199 on Build GRCh38
rs779272813 Chr.14: 21392243-21392243 on Build GRCh38
rs756297076 Chr.14: 21392296-21392296 on Build GRCh38
rs907462443 Chr.14: 21392090-21392090 on Build GRCh38
rs769881262 Chr.14: 21392218-21392218 on Build GRCh38
rs929714154 Chr.14: 21392328-21392328 on Build GRCh38
rs773126066 Chr.14: 21392221-21392221 on Build GRCh38
rs556888476 Chr.14: 21392295-21392295 on Build GRCh38
rs758413800 Chr.14: 21392281-21392281 on Build GRCh38
rs369233599 Chr.14: 21392166-21392166 on Build GRCh38
rs771537778 Chr.14: 21392478-21392478 on Build GRCh38
rs1998332 Chr.14: 21392201-21392201 on Build GRCh38
rs774726221 Chr.14: 21392487-21392487 on Build GRCh38
rs1040689640 Chr.14: 21392406-21392406 on Build GRCh38
rs939687679 Chr.14: 21392378-21392378 on Build GRCh38
rs534350751 Chr.14: 21392169-21392169 on Build GRCh38

View Full Product Details