Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23940163-23940702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS4-AS1
Entrez Gene ID
55449
Gene Name
DHRS4 antisense RNA 1
Gene Aliases
AS1DHRS4, C14orf167, DHRS4AS1, PRO1488
UniGene
Hs.569361
Gene Chromosome Location
Chr.14: 23938731-23955112 on Build GRCh38
Transcripts
NR_023921.2 NR_023922.2 NR_023923.2 NR_023924.2

SNP Detail

SNP ID Chromosome Location
rs531918955 Chr.14: 23940557-23940557 on Build GRCh38
rs987428326 Chr.14: 23940230-23940230 on Build GRCh38
rs956963029 Chr.14: 23940200-23940200 on Build GRCh38
rs1008811933 Chr.14: 23940531-23940531 on Build GRCh38
rs936338493 Chr.14: 23940519-23940519 on Build GRCh38
rs964358517 Chr.14: 23940237-23940237 on Build GRCh38
rs980658596 Chr.14: 23940640-23940640 on Build GRCh38
rs898612277 Chr.14: 23940574-23940574 on Build GRCh38
rs1052131656 Chr.14: 23940413-23940413 on Build GRCh38
rs113551151 Chr.14: 23940590-23940590 on Build GRCh38
rs924910370 Chr.14: 23940513-23940513 on Build GRCh38
rs769723287 Chr.14: 23940278-23940278 on Build GRCh38
rs1038515782 Chr.14: 23940572-23940572 on Build GRCh38
rs940554408 Chr.14: 23940294-23940294 on Build GRCh38
rs560494134 Chr.14: 23940473-23940473 on Build GRCh38
rs947693816 Chr.14: 23940345-23940345 on Build GRCh38
rs1016085833 Chr.14: 23940605-23940605 on Build GRCh38
rs112851730 Chr.14: 23940478-23940478 on Build GRCh38
rs892221190 Chr.14: 23940527-23940527 on Build GRCh38
rs761815423 Chr.14: 23940247-23940247 on Build GRCh38
rs983249605 Chr.14: 23940457-23940457 on Build GRCh38
rs184272760 Chr.14: 23940188-23940188 on Build GRCh38
rs925191524 Chr.14: 23940663-23940663 on Build GRCh38
rs1001675045 Chr.14: 23940329-23940329 on Build GRCh38
rs185970772 Chr.14: 23940411-23940411 on Build GRCh38
rs917824184 Chr.14: 23940482-23940482 on Build GRCh38
rs574129480 Chr.14: 23940351-23940352 on Build GRCh38
rs868017211 Chr.14: 23940365-23940365 on Build GRCh38
rs917784431 Chr.14: 23940627-23940627 on Build GRCh38
rs1052099259 Chr.14: 23940260-23940260 on Build GRCh38
rs906045625 Chr.14: 23940298-23940298 on Build GRCh38
rs975420934 Chr.14: 23940251-23940251 on Build GRCh38
rs79821036 Chr.14: 23940396-23940396 on Build GRCh38
rs1025585741 Chr.14: 23940348-23940348 on Build GRCh38
rs931422708 Chr.14: 23940268-23940268 on Build GRCh38
rs758528448 Chr.14: 23940340-23940341 on Build GRCh38
rs141875463 Chr.14: 23940408-23940408 on Build GRCh38
rs952675909 Chr.14: 23940593-23940593 on Build GRCh38
rs373328780 Chr.14: 23940608-23940608 on Build GRCh38
rs113459990 Chr.14: 23940520-23940520 on Build GRCh38
rs368656986 Chr.14: 23940479-23940479 on Build GRCh38
rs975368558 Chr.14: 23940446-23940446 on Build GRCh38
rs903585535 Chr.14: 23940401-23940401 on Build GRCh38
rs1017467554 Chr.14: 23940340-23940340 on Build GRCh38
rs572395136 Chr.14: 23940440-23940440 on Build GRCh38
rs140666882 Chr.14: 23940509-23940509 on Build GRCh38
rs1026973635 Chr.14: 23940449-23940449 on Build GRCh38
rs964229782 Chr.14: 23940382-23940382 on Build GRCh38
rs562016820 Chr.14: 23940594-23940594 on Build GRCh38
rs540600860 Chr.14: 23940443-23940443 on Build GRCh38
rs1056744956 Chr.14: 23940321-23940321 on Build GRCh38
rs896410763 Chr.14: 23940341-23940341 on Build GRCh38
rs916182810 Chr.14: 23940372-23940372 on Build GRCh38
rs952729779 Chr.14: 23940450-23940450 on Build GRCh38
rs1041242075 Chr.14: 23940536-23940536 on Build GRCh38
rs188611386 Chr.14: 23940254-23940254 on Build GRCh38
rs759804403 Chr.14: 23940634-23940634 on Build GRCh38
rs76392638 Chr.14: 23940393-23940393 on Build GRCh38
rs76175404 Chr.14: 23940397-23940397 on Build GRCh38
rs907673320 Chr.14: 23940292-23940292 on Build GRCh38
rs376165566 Chr.14: 23940409-23940409 on Build GRCh38
rs559498081 Chr.14: 23940530-23940530 on Build GRCh38
rs947974301 Chr.14: 23940508-23940508 on Build GRCh38
rs900082035 Chr.14: 23940545-23940545 on Build GRCh38
rs911101090 Chr.14: 23940377-23940377 on Build GRCh38
rs374228363 Chr.14: 23940370-23940370 on Build GRCh38
rs962763536 Chr.14: 23940344-23940344 on Build GRCh38
rs984576515 Chr.14: 23940270-23940270 on Build GRCh38
rs955346266 Chr.14: 23940349-23940349 on Build GRCh38
rs992382491 Chr.14: 23940615-23940615 on Build GRCh38
rs997026831 Chr.14: 23940575-23940575 on Build GRCh38
rs527995960 Chr.14: 23940596-23940596 on Build GRCh38
rs942524181 Chr.14: 23940228-23940228 on Build GRCh38
rs1018643100 Chr.14: 23940610-23940610 on Build GRCh38
rs908628600 Chr.14: 23940472-23940472 on Build GRCh38
rs892603500 Chr.14: 23940410-23940410 on Build GRCh38
rs1019881887 Chr.14: 23940232-23940232 on Build GRCh38
rs139190244 Chr.14: 23940649-23940649 on Build GRCh38

View Full Product Details