Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28777750-28778260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01551
Entrez Gene ID
387978
Gene Name
long intergenic non-protein coding RNA 1551
Gene Aliases
C14orf23, c14_5148
UniGene
Hs.633254
Gene Chromosome Location
Chr.14: 28772704-28794794 on Build GRCh38
Transcripts
NR_026731.1 NR_026732.1

SNP Detail

SNP ID Chromosome Location
rs943508327 Chr.14: 28778173-28778173 on Build GRCh38
rs1021355574 Chr.14: 28778239-28778239 on Build GRCh38
rs772210784 Chr.14: 28778080-28778081 on Build GRCh38
rs762894630 Chr.14: 28778095-28778095 on Build GRCh38
rs969855775 Chr.14: 28778058-28778058 on Build GRCh38
rs919250353 Chr.14: 28778085-28778085 on Build GRCh38
rs375225478 Chr.14: 28778072-28778072 on Build GRCh38
rs543586662 Chr.14: 28778043-28778043 on Build GRCh38
rs571998521 Chr.14: 28777861-28777861 on Build GRCh38
rs1007613696 Chr.14: 28777975-28777975 on Build GRCh38
rs937766938 Chr.14: 28778148-28778148 on Build GRCh38
rs768360006 Chr.14: 28777929-28777929 on Build GRCh38
rs765606800 Chr.14: 28778217-28778217 on Build GRCh38
rs896101542 Chr.14: 28777804-28777804 on Build GRCh38
rs777953838 Chr.14: 28777842-28777843 on Build GRCh38
rs917604334 Chr.14: 28778183-28778183 on Build GRCh38
rs767293994 Chr.14: 28778017-28778017 on Build GRCh38
rs929419323 Chr.14: 28778127-28778127 on Build GRCh38
rs976372913 Chr.14: 28778029-28778029 on Build GRCh38
rs576153235 Chr.14: 28777980-28777980 on Build GRCh38
rs1011318535 Chr.14: 28778032-28778032 on Build GRCh38
rs565358825 Chr.14: 28778044-28778044 on Build GRCh38
rs957433756 Chr.14: 28777937-28777937 on Build GRCh38
rs919487286 Chr.14: 28778083-28778083 on Build GRCh38
rs1049251298 Chr.14: 28777782-28777782 on Build GRCh38
rs541654172 Chr.14: 28777824-28777824 on Build GRCh38
rs1042669969 Chr.14: 28777847-28777847 on Build GRCh38
rs565333332 Chr.14: 28778079-28778080 on Build GRCh38
rs547804440 Chr.14: 28778082-28778082 on Build GRCh38
rs943762277 Chr.14: 28778073-28778073 on Build GRCh38
rs950954825 Chr.14: 28778097-28778097 on Build GRCh38
rs80214643 Chr.14: 28778092-28778092 on Build GRCh38
rs984918568 Chr.14: 28778122-28778122 on Build GRCh38
rs532449149 Chr.14: 28778065-28778065 on Build GRCh38
rs11365680 Chr.14: 28777842-28777842 on Build GRCh38
rs777888073 Chr.14: 28778169-28778169 on Build GRCh38
rs772487066 Chr.14: 28777872-28777872 on Build GRCh38
rs966414891 Chr.14: 28777988-28777988 on Build GRCh38
rs757679085 Chr.14: 28777841-28777842 on Build GRCh38
rs559744993 Chr.14: 28778141-28778141 on Build GRCh38
rs780434596 Chr.14: 28778080-28778082 on Build GRCh38
rs766237487 Chr.14: 28778098-28778098 on Build GRCh38
rs772852737 Chr.14: 28778088-28778089 on Build GRCh38
rs997554141 Chr.14: 28777908-28777908 on Build GRCh38
rs1039667113 Chr.14: 28777863-28777863 on Build GRCh38
rs566209730 Chr.14: 28778192-28778192 on Build GRCh38
rs78640311 Chr.14: 28778178-28778178 on Build GRCh38
rs776421154 Chr.14: 28777938-28777938 on Build GRCh38
rs1013523380 Chr.14: 28777841-28777841 on Build GRCh38
rs61982298 Chr.14: 28778078-28778078 on Build GRCh38
rs1007664579 Chr.14: 28778231-28778231 on Build GRCh38
rs1018075379 Chr.14: 28778237-28778237 on Build GRCh38
rs72669758 Chr.14: 28778210-28778210 on Build GRCh38
rs75294746 Chr.14: 28778093-28778093 on Build GRCh38
rs560927885 Chr.14: 28777971-28777971 on Build GRCh38
rs148412023 Chr.14: 28777803-28777803 on Build GRCh38
rs1050336555 Chr.14: 28777814-28777814 on Build GRCh38
rs553696162 Chr.14: 28777860-28777860 on Build GRCh38
rs775304025 Chr.14: 28778140-28778140 on Build GRCh38
rs943645573 Chr.14: 28777840-28777843 on Build GRCh38
rs763830400 Chr.14: 28778191-28778191 on Build GRCh38
rs761620038 Chr.14: 28777967-28777967 on Build GRCh38
rs949060255 Chr.14: 28778184-28778184 on Build GRCh38
rs771059013 Chr.14: 28778080-28778080 on Build GRCh38
rs759525641 Chr.14: 28777974-28777974 on Build GRCh38
rs11316936 Chr.14: 28777855-28777855 on Build GRCh38
rs775018004 Chr.14: 28777885-28777885 on Build GRCh38
rs77255418 Chr.14: 28778080-28778080 on Build GRCh38
rs765063006 Chr.14: 28778067-28778067 on Build GRCh38
rs891450199 Chr.14: 28778233-28778233 on Build GRCh38
rs375203506 Chr.14: 28777881-28777881 on Build GRCh38
rs542473273 Chr.14: 28777927-28777927 on Build GRCh38
rs937714800 Chr.14: 28777785-28777785 on Build GRCh38
rs901520187 Chr.14: 28777901-28777901 on Build GRCh38
rs1000734563 Chr.14: 28778201-28778201 on Build GRCh38
rs756180285 Chr.14: 28778138-28778138 on Build GRCh38
rs1035982328 Chr.14: 28778077-28778077 on Build GRCh38
rs869129101 Chr.14: 28777854-28777855 on Build GRCh38
rs760276610 Chr.14: 28777903-28777903 on Build GRCh38
rs909400201 Chr.14: 28778143-28778143 on Build GRCh38
rs562693408 Chr.14: 28778239-28778239 on Build GRCh38

View Full Product Details