Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 28792598-28793134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01551
Entrez Gene ID
387978
Gene Name
long intergenic non-protein coding RNA 1551
Gene Aliases
C14orf23, c14_5148
UniGene
Hs.633254
Gene Chromosome Location
Chr.14: 28772704-28794794 on Build GRCh38
Transcripts
NR_026731.1

SNP Detail

SNP ID Chromosome Location
rs565231561 Chr.14: 28792886-28792886 on Build GRCh38
rs572112609 Chr.14: 28792727-28792727 on Build GRCh38
rs916727593 Chr.14: 28792799-28792799 on Build GRCh38
rs915176105 Chr.14: 28792973-28792973 on Build GRCh38
rs987483467 Chr.14: 28793078-28793078 on Build GRCh38
rs969573897 Chr.14: 28792824-28792824 on Build GRCh38
rs1950192 Chr.14: 28792768-28792768 on Build GRCh38
rs150245637 Chr.14: 28793064-28793064 on Build GRCh38
rs58499139 Chr.14: 28793103-28793103 on Build GRCh38
rs145837889 Chr.14: 28793066-28793066 on Build GRCh38
rs981866096 Chr.14: 28792859-28792859 on Build GRCh38
rs987678183 Chr.14: 28792782-28792782 on Build GRCh38
rs745857023 Chr.14: 28793047-28793047 on Build GRCh38
rs191531211 Chr.14: 28793021-28793021 on Build GRCh38
rs1010703514 Chr.14: 28792689-28792689 on Build GRCh38
rs543845728 Chr.14: 28792869-28792869 on Build GRCh38
rs927659596 Chr.14: 28792866-28792866 on Build GRCh38
rs953571207 Chr.14: 28792779-28792779 on Build GRCh38
rs900596897 Chr.14: 28792707-28792707 on Build GRCh38
rs1051847213 Chr.14: 28793093-28793093 on Build GRCh38
rs996261510 Chr.14: 28792732-28792732 on Build GRCh38
rs1004096709 Chr.14: 28792739-28792739 on Build GRCh38
rs954729184 Chr.14: 28792751-28792751 on Build GRCh38
rs886899067 Chr.14: 28792974-28792974 on Build GRCh38
rs1015190438 Chr.14: 28792767-28792767 on Build GRCh38
rs745842308 Chr.14: 28792770-28792770 on Build GRCh38
rs898395494 Chr.14: 28793003-28793003 on Build GRCh38
rs887034816 Chr.14: 28792688-28792688 on Build GRCh38
rs769885822 Chr.14: 28792787-28792787 on Build GRCh38
rs555079577 Chr.14: 28792755-28792755 on Build GRCh38
rs577488469 Chr.14: 28793041-28793041 on Build GRCh38
rs973588299 Chr.14: 28792791-28792791 on Build GRCh38
rs1024120819 Chr.14: 28792671-28792671 on Build GRCh38
rs946607524 Chr.14: 28792993-28792993 on Build GRCh38
rs1019073639 Chr.14: 28793086-28793086 on Build GRCh38
rs1018038752 Chr.14: 28792690-28792690 on Build GRCh38
rs949810445 Chr.14: 28792956-28792956 on Build GRCh38
rs866056328 Chr.14: 28792761-28792761 on Build GRCh38
rs878977639 Chr.14: 28792758-28792758 on Build GRCh38
rs1055849575 Chr.14: 28792891-28792891 on Build GRCh38
rs946309955 Chr.14: 28792822-28792822 on Build GRCh38
rs1950191 Chr.14: 28792740-28792740 on Build GRCh38
rs775092271 Chr.14: 28792888-28792888 on Build GRCh38
rs963410768 Chr.14: 28792752-28792752 on Build GRCh38
rs1052017157 Chr.14: 28792923-28792923 on Build GRCh38
rs1004044427 Chr.14: 28792984-28792984 on Build GRCh38
rs994874382 Chr.14: 28793062-28793062 on Build GRCh38

View Full Product Details