Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51650706-51651202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD6
Entrez Gene ID
122786
Gene Name
FERM domain containing 6
Gene Aliases
C14orf31, EX1, Willin, c14_5320
UniGene
Hs.434914
Gene Chromosome Location
Chr.14: 51489100-51730727 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

Gene Symbol
FRMD6-AS1
Entrez Gene ID
145438
Gene Name
FRMD6 antisense RNA 1
Gene Aliases
C14orf82
UniGene
Hs.645410
Gene Chromosome Location
Chr.14: 51649516-51651744 on Build GRCh38
Transcripts
NM_001042481.2 XM_011536423.1 XM_011536424.1 NR_037676.1

SNP Detail

SNP ID Chromosome Location
rs533332366 Chr.14: 51650907-51650907 on Build GRCh38
rs907205473 Chr.14: 51650808-51650808 on Build GRCh38
rs1034285256 Chr.14: 51650850-51650850 on Build GRCh38
rs888436576 Chr.14: 51651095-51651095 on Build GRCh38
rs570847042 Chr.14: 51650876-51650876 on Build GRCh38
rs967134464 Chr.14: 51650872-51650872 on Build GRCh38
rs775550073 Chr.14: 51650982-51650984 on Build GRCh38
rs1001918112 Chr.14: 51650836-51650836 on Build GRCh38
rs1049243385 Chr.14: 51650735-51650735 on Build GRCh38
rs916348548 Chr.14: 51650985-51650985 on Build GRCh38
rs994086586 Chr.14: 51650824-51650824 on Build GRCh38
rs963763808 Chr.14: 51650912-51650912 on Build GRCh38
rs542012889 Chr.14: 51650771-51650771 on Build GRCh38
rs1008451870 Chr.14: 51651155-51651155 on Build GRCh38
rs940801210 Chr.14: 51650971-51650971 on Build GRCh38
rs865811727 Chr.14: 51650818-51650818 on Build GRCh38
rs928683237 Chr.14: 51651010-51651010 on Build GRCh38
rs148132586 Chr.14: 51651131-51651131 on Build GRCh38
rs893041813 Chr.14: 51650855-51650855 on Build GRCh38
rs1047038059 Chr.14: 51651093-51651093 on Build GRCh38
rs575429298 Chr.14: 51651088-51651088 on Build GRCh38
rs977366171 Chr.14: 51650899-51650899 on Build GRCh38
rs977658502 Chr.14: 51650954-51650954 on Build GRCh38
rs899878158 Chr.14: 51651173-51651173 on Build GRCh38
rs45598040 Chr.14: 51651003-51651003 on Build GRCh38
rs868836638 Chr.14: 51650803-51650803 on Build GRCh38
rs984993676 Chr.14: 51650961-51650961 on Build GRCh38
rs746585446 Chr.14: 51650823-51650823 on Build GRCh38
rs943598671 Chr.14: 51651019-51651019 on Build GRCh38
rs757754713 Chr.14: 51650960-51650960 on Build GRCh38
rs1019741124 Chr.14: 51650910-51650910 on Build GRCh38
rs376926938 Chr.14: 51651057-51651057 on Build GRCh38
rs112843208 Chr.14: 51650821-51650821 on Build GRCh38
rs922234899 Chr.14: 51650931-51650931 on Build GRCh38
rs1020071133 Chr.14: 51650866-51650866 on Build GRCh38
rs886564119 Chr.14: 51650744-51650744 on Build GRCh38
rs947908513 Chr.14: 51650777-51650778 on Build GRCh38
rs1041581199 Chr.14: 51651096-51651096 on Build GRCh38
rs536084834 Chr.14: 51651091-51651091 on Build GRCh38
rs975454681 Chr.14: 51650922-51650922 on Build GRCh38
rs1046617005 Chr.14: 51651073-51651073 on Build GRCh38
rs371989509 Chr.14: 51650863-51650863 on Build GRCh38
rs940750300 Chr.14: 51650792-51650792 on Build GRCh38
rs555676559 Chr.14: 51651105-51651105 on Build GRCh38
rs900813661 Chr.14: 51651030-51651030 on Build GRCh38
rs894958596 Chr.14: 51650747-51650747 on Build GRCh38
rs3751464 Chr.14: 51651174-51651174 on Build GRCh38
rs937386925 Chr.14: 51651040-51651040 on Build GRCh38
rs1039788774 Chr.14: 51651171-51651171 on Build GRCh38
rs1029757530 Chr.14: 51651175-51651175 on Build GRCh38
rs908051221 Chr.14: 51650962-51650962 on Build GRCh38
rs200364647 Chr.14: 51651025-51651025 on Build GRCh38

View Full Product Details