Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91126068-91126580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf159
Entrez Gene ID
80017
Gene Name
chromosome 14 open reading frame 159
Gene Aliases
-
UniGene
Hs.309849
Gene Chromosome Location
Chr.14: 91114013-91225396 on Build GRCh38
Transcripts
NM_001102366.2 NM_001102367.2 NM_001102368.2 NM_001102369.2 NM_001286470.1 NM_001286471.1 NM_001286472.1 NM_001286473.1 NM_024952.7 NR_003709.1

Gene Symbol
SNORA11B
Entrez Gene ID
100124539
Gene Name
small nucleolar RNA, H/ACA box 11B
Gene Aliases
-
UniGene
Hs.693444
Gene Chromosome Location
Chr.14: 91126426-91126552 on Build GRCh38
Transcripts
NM_001102366.2 NM_001102367.2 NM_001102368.2 NM_001102369.2 NM_001286470.1 NM_001286471.1 NM_001286472.1 NM_001286473.1 NM_024952.7 NR_003709.1

SNP Detail

SNP ID Chromosome Location
rs894648582 Chr.14: 91126405-91126405 on Build GRCh38
rs1033329240 Chr.14: 91126435-91126435 on Build GRCh38
rs397817202 Chr.14: 91126409-91126410 on Build GRCh38
rs958475699 Chr.14: 91126193-91126193 on Build GRCh38
rs950365053 Chr.14: 91126386-91126386 on Build GRCh38
rs1002712525 Chr.14: 91126365-91126365 on Build GRCh38
rs911299846 Chr.14: 91126271-91126271 on Build GRCh38
rs1005713760 Chr.14: 91126372-91126372 on Build GRCh38
rs769439030 Chr.14: 91126292-91126292 on Build GRCh38
rs1024350710 Chr.14: 91126202-91126202 on Build GRCh38
rs138878456 Chr.14: 91126388-91126388 on Build GRCh38
rs922564959 Chr.14: 91126335-91126335 on Build GRCh38
rs1046474566 Chr.14: 91126539-91126539 on Build GRCh38
rs901423706 Chr.14: 91126133-91126133 on Build GRCh38
rs1054222781 Chr.14: 91126525-91126525 on Build GRCh38
rs534936447 Chr.14: 91126200-91126200 on Build GRCh38
rs977714679 Chr.14: 91126207-91126207 on Build GRCh38
rs974929857 Chr.14: 91126450-91126450 on Build GRCh38
rs68124656 Chr.14: 91126405-91126406 on Build GRCh38
rs150423024 Chr.14: 91126512-91126512 on Build GRCh38
rs971313132 Chr.14: 91126117-91126117 on Build GRCh38
rs1026191381 Chr.14: 91126383-91126383 on Build GRCh38
rs954557325 Chr.14: 91126308-91126317 on Build GRCh38
rs888589227 Chr.14: 91126145-91126145 on Build GRCh38
rs200360927 Chr.14: 91126404-91126404 on Build GRCh38
rs144945750 Chr.14: 91126162-91126162 on Build GRCh38
rs895230726 Chr.14: 91126526-91126526 on Build GRCh38
rs536741449 Chr.14: 91126233-91126233 on Build GRCh38
rs762706250 Chr.14: 91126489-91126489 on Build GRCh38
rs1039632729 Chr.14: 91126212-91126212 on Build GRCh38
rs765298314 Chr.14: 91126304-91126304 on Build GRCh38
rs950395750 Chr.14: 91126137-91126137 on Build GRCh38
rs796762388 Chr.14: 91126100-91126100 on Build GRCh38
rs576004234 Chr.14: 91126363-91126363 on Build GRCh38
rs923515313 Chr.14: 91126224-91126224 on Build GRCh38
rs957416909 Chr.14: 91126234-91126234 on Build GRCh38
rs933593866 Chr.14: 91126228-91126228 on Build GRCh38
rs895251344 Chr.14: 91126310-91126310 on Build GRCh38
rs78016680 Chr.14: 91126410-91126410 on Build GRCh38
rs971282275 Chr.14: 91126351-91126351 on Build GRCh38
rs527588790 Chr.14: 91126219-91126219 on Build GRCh38
rs201143531 Chr.14: 91126402-91126402 on Build GRCh38
rs187073759 Chr.14: 91126245-91126245 on Build GRCh38
rs948172844 Chr.14: 91126530-91126530 on Build GRCh38
rs57456676 Chr.14: 91126416-91126416 on Build GRCh38
rs981073894 Chr.14: 91126119-91126119 on Build GRCh38
rs548316993 Chr.14: 91126204-91126204 on Build GRCh38
rs202204979 Chr.14: 91126403-91126403 on Build GRCh38
rs957318590 Chr.14: 91126457-91126457 on Build GRCh38
rs1012736007 Chr.14: 91126315-91126315 on Build GRCh38
rs1034084688 Chr.14: 91126192-91126192 on Build GRCh38
rs57615276 Chr.14: 91126404-91126405 on Build GRCh38
rs923399364 Chr.14: 91126460-91126460 on Build GRCh38
rs1022794708 Chr.14: 91126328-91126328 on Build GRCh38
rs745427697 Chr.14: 91126267-91126267 on Build GRCh38
rs1022870917 Chr.14: 91126110-91126110 on Build GRCh38
rs1047743788 Chr.14: 91126342-91126342 on Build GRCh38
rs907006652 Chr.14: 91126554-91126554 on Build GRCh38
rs902825997 Chr.14: 91126422-91126422 on Build GRCh38
rs192852795 Chr.14: 91126499-91126499 on Build GRCh38
rs1041060691 Chr.14: 91126102-91126102 on Build GRCh38
rs918959246 Chr.14: 91126136-91126136 on Build GRCh38
rs936670602 Chr.14: 91126519-91126519 on Build GRCh38
rs930010886 Chr.14: 91126336-91126336 on Build GRCh38
rs76554363 Chr.14: 91126428-91126428 on Build GRCh38
rs568170600 Chr.14: 91126230-91126230 on Build GRCh38
rs764046574 Chr.14: 91126488-91126488 on Build GRCh38

View Full Product Details