Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93910466-93910963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM181A-AS1
Entrez Gene ID
283592
Gene Name
FAM181A antisense RNA 1
Gene Aliases
C14orf86
UniGene
Hs.585648
Gene Chromosome Location
Chr.14: 93904730-93926372 on Build GRCh38
Transcripts
NR_027004.1

SNP Detail

SNP ID Chromosome Location
rs910953278 Chr.14: 93910571-93910571 on Build GRCh38
rs570701024 Chr.14: 93910752-93910754 on Build GRCh38
rs931093622 Chr.14: 93910883-93910883 on Build GRCh38
rs999807960 Chr.14: 93910701-93910701 on Build GRCh38
rs1037881025 Chr.14: 93910858-93910858 on Build GRCh38
rs1046586337 Chr.14: 93910664-93910664 on Build GRCh38
rs1049418603 Chr.14: 93910630-93910630 on Build GRCh38
rs552658618 Chr.14: 93910562-93910562 on Build GRCh38
rs927497174 Chr.14: 93910529-93910529 on Build GRCh38
rs1000181443 Chr.14: 93910526-93910526 on Build GRCh38
rs980130184 Chr.14: 93910502-93910502 on Build GRCh38
rs916576583 Chr.14: 93910837-93910837 on Build GRCh38
rs772593112 Chr.14: 93910671-93910671 on Build GRCh38
rs960261608 Chr.14: 93910821-93910821 on Build GRCh38
rs17129336 Chr.14: 93910620-93910620 on Build GRCh38
rs1049364656 Chr.14: 93910899-93910899 on Build GRCh38
rs919640638 Chr.14: 93910863-93910863 on Build GRCh38
rs1046902392 Chr.14: 93910881-93910881 on Build GRCh38
rs957926402 Chr.14: 93910557-93910557 on Build GRCh38
rs898246875 Chr.14: 93910609-93910609 on Build GRCh38
rs762581019 Chr.14: 93910732-93910732 on Build GRCh38
rs942012767 Chr.14: 93910608-93910608 on Build GRCh38
rs889674118 Chr.14: 93910645-93910645 on Build GRCh38
rs938928801 Chr.14: 93910569-93910569 on Build GRCh38
rs967019214 Chr.14: 93910517-93910517 on Build GRCh38
rs370537798 Chr.14: 93910781-93910781 on Build GRCh38
rs764722309 Chr.14: 93910752-93910752 on Build GRCh38
rs17129338 Chr.14: 93910849-93910849 on Build GRCh38
rs767062456 Chr.14: 93910936-93910936 on Build GRCh38
rs142080277 Chr.14: 93910602-93910602 on Build GRCh38
rs528431921 Chr.14: 93910812-93910812 on Build GRCh38
rs573485089 Chr.14: 93910582-93910582 on Build GRCh38
rs1013667668 Chr.14: 93910490-93910490 on Build GRCh38
rs908038200 Chr.14: 93910846-93910846 on Build GRCh38
rs796474402 Chr.14: 93910572-93910572 on Build GRCh38
rs111966617 Chr.14: 93910583-93910583 on Build GRCh38
rs751916608 Chr.14: 93910795-93910795 on Build GRCh38
rs546568883 Chr.14: 93910832-93910832 on Build GRCh38
rs976550009 Chr.14: 93910573-93910573 on Build GRCh38
rs1032027903 Chr.14: 93910543-93910543 on Build GRCh38
rs544733237 Chr.14: 93910711-93910711 on Build GRCh38
rs943948673 Chr.14: 93910937-93910937 on Build GRCh38
rs984073650 Chr.14: 93910785-93910785 on Build GRCh38
rs75807715 Chr.14: 93910760-93910760 on Build GRCh38
rs77519604 Chr.14: 93910733-93910733 on Build GRCh38
rs1013615204 Chr.14: 93910648-93910648 on Build GRCh38
rs1008121233 Chr.14: 93910667-93910667 on Build GRCh38
rs929950151 Chr.14: 93910581-93910581 on Build GRCh38
rs142966607 Chr.14: 93910935-93910935 on Build GRCh38
rs146300825 Chr.14: 93910712-93910712 on Build GRCh38
rs909751233 Chr.14: 93910801-93910801 on Build GRCh38
rs201585420 Chr.14: 93910757-93910757 on Build GRCh38
rs1018046402 Chr.14: 93910749-93910749 on Build GRCh38
rs927297523 Chr.14: 93910762-93910762 on Build GRCh38
rs993109051 Chr.14: 93910822-93910822 on Build GRCh38
rs902690900 Chr.14: 93910921-93910921 on Build GRCh38
rs908065876 Chr.14: 93910579-93910579 on Build GRCh38

View Full Product Details