Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95179557-95180103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DICER1-AS1
Entrez Gene ID
400242
Gene Name
DICER1 antisense RNA 1
Gene Aliases
DICER1-AS
UniGene
Hs.497573
Gene Chromosome Location
Chr.14: 95157688-95179933 on Build GRCh38
Transcripts
NR_015415.1

SNP Detail

SNP ID Chromosome Location
rs1005971896 Chr.14: 95179949-95179949 on Build GRCh38
rs866029278 Chr.14: 95180057-95180057 on Build GRCh38
rs368293266 Chr.14: 95179732-95179733 on Build GRCh38
rs924771272 Chr.14: 95179786-95179786 on Build GRCh38
rs529423357 Chr.14: 95179952-95179952 on Build GRCh38
rs996474899 Chr.14: 95180013-95180013 on Build GRCh38
rs574277415 Chr.14: 95179728-95179729 on Build GRCh38
rs146005251 Chr.14: 95180014-95180014 on Build GRCh38
rs1054716414 Chr.14: 95179795-95179795 on Build GRCh38
rs922726428 Chr.14: 95179721-95179721 on Build GRCh38
rs984663065 Chr.14: 95179687-95179687 on Build GRCh38
rs563632826 Chr.14: 95179824-95179824 on Build GRCh38
rs1032719196 Chr.14: 95179609-95179609 on Build GRCh38
rs146179677 Chr.14: 95179787-95179787 on Build GRCh38
rs139418272 Chr.14: 95179853-95179853 on Build GRCh38
rs934518493 Chr.14: 95179852-95179852 on Build GRCh38
rs955482917 Chr.14: 95180073-95180073 on Build GRCh38
rs750358496 Chr.14: 95179778-95179778 on Build GRCh38
rs868702754 Chr.14: 95179649-95179649 on Build GRCh38
rs780150756 Chr.14: 95179932-95179932 on Build GRCh38
rs541662017 Chr.14: 95180078-95180078 on Build GRCh38
rs894694271 Chr.14: 95179820-95179820 on Build GRCh38
rs948694467 Chr.14: 95179890-95179891 on Build GRCh38
rs971525352 Chr.14: 95179650-95179650 on Build GRCh38
rs767519487 Chr.14: 95179740-95179740 on Build GRCh38
rs139588712 Chr.14: 95180029-95180029 on Build GRCh38
rs572298226 Chr.14: 95179735-95179735 on Build GRCh38
rs751306549 Chr.14: 95179622-95179622 on Build GRCh38
rs1044385553 Chr.14: 95179900-95179900 on Build GRCh38
rs778444857 Chr.14: 95179973-95179973 on Build GRCh38
rs556414140 Chr.14: 95179715-95179715 on Build GRCh38
rs568224993 Chr.14: 95179720-95179720 on Build GRCh38
rs753184068 Chr.14: 95179865-95179866 on Build GRCh38
rs14042 Chr.14: 95179683-95179683 on Build GRCh38
rs1000432281 Chr.14: 95179599-95179599 on Build GRCh38
rs74079230 Chr.14: 95179889-95179889 on Build GRCh38
rs866190519 Chr.14: 95179670-95179670 on Build GRCh38
rs1041932145 Chr.14: 95179753-95179753 on Build GRCh38
rs1026083392 Chr.14: 95179672-95179672 on Build GRCh38
rs1024075696 Chr.14: 95179632-95179632 on Build GRCh38
rs894135610 Chr.14: 95179621-95179621 on Build GRCh38
rs541611247 Chr.14: 95180008-95180008 on Build GRCh38
rs142614971 Chr.14: 95179929-95179929 on Build GRCh38
rs886647914 Chr.14: 95179667-95179667 on Build GRCh38
rs190366646 Chr.14: 95179725-95179725 on Build GRCh38
rs778743175 Chr.14: 95179693-95179693 on Build GRCh38
rs968457423 Chr.14: 95179750-95179750 on Build GRCh38
rs1019163261 Chr.14: 95179981-95179981 on Build GRCh38
rs754592691 Chr.14: 95179909-95179909 on Build GRCh38
rs886589554 Chr.14: 95179945-95179945 on Build GRCh38
rs1047839048 Chr.14: 95179938-95179938 on Build GRCh38
rs149714262 Chr.14: 95180056-95180056 on Build GRCh38
rs561956040 Chr.14: 95179951-95179951 on Build GRCh38
rs1019635463 Chr.14: 95179698-95179699 on Build GRCh38
rs951887584 Chr.14: 95179935-95179935 on Build GRCh38
rs964082217 Chr.14: 95179717-95179717 on Build GRCh38

View Full Product Details