Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95534606-95535103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX5
Entrez Gene ID
51218
Gene Name
glutaredoxin 5
Gene Aliases
C14orf87, FLB4739, GRX5, PR01238, PRO1238, PRSA, SIDBA3, SPAHGC
UniGene
Hs.744943
Gene Chromosome Location
Chr.14: 95534986-95544718 on Build GRCh38
Transcripts
NM_016417.2 NR_001459.2 NR_003138.3

Gene Symbol
SNHG10
Entrez Gene ID
283596
Gene Name
small nucleolar RNA host gene 10
Gene Aliases
C14orf62, LINC00063, NCRNA00063
UniGene
Hs.448753
Gene Chromosome Location
Chr.14: 95532912-95534872 on Build GRCh38
Transcripts
NM_016417.2 NR_001459.2 NR_003138.3

SNP Detail

SNP ID Chromosome Location
rs748678949 Chr.14: 95535045-95535045 on Build GRCh38
rs909960803 Chr.14: 95534680-95534680 on Build GRCh38
rs911681 Chr.14: 95535046-95535046 on Build GRCh38
rs373155250 Chr.14: 95534876-95534876 on Build GRCh38
rs771656027 Chr.14: 95534904-95534904 on Build GRCh38
rs912861603 Chr.14: 95535004-95535004 on Build GRCh38
rs554982689 Chr.14: 95534718-95534718 on Build GRCh38
rs897410235 Chr.14: 95534897-95534897 on Build GRCh38
rs776972957 Chr.14: 95535068-95535068 on Build GRCh38
rs889911240 Chr.14: 95534747-95534747 on Build GRCh38
rs538318447 Chr.14: 95535036-95535036 on Build GRCh38
rs781442697 Chr.14: 95534694-95534694 on Build GRCh38
rs959586131 Chr.14: 95534883-95534884 on Build GRCh38
rs976815628 Chr.14: 95534991-95534991 on Build GRCh38
rs968591 Chr.14: 95534887-95534887 on Build GRCh38
rs746009921 Chr.14: 95535072-95535072 on Build GRCh38
rs1003377239 Chr.14: 95534875-95534875 on Build GRCh38
rs962872356 Chr.14: 95535013-95535013 on Build GRCh38
rs943672903 Chr.14: 95534764-95534764 on Build GRCh38
rs553055759 Chr.14: 95534969-95534969 on Build GRCh38
rs779975494 Chr.14: 95534751-95534751 on Build GRCh38
rs112431749 Chr.14: 95534982-95534982 on Build GRCh38
rs45582537 Chr.14: 95534813-95534813 on Build GRCh38
rs972776103 Chr.14: 95535028-95535028 on Build GRCh38
rs1034898471 Chr.14: 95534878-95534878 on Build GRCh38
rs1035128636 Chr.14: 95534976-95534976 on Build GRCh38
rs555474190 Chr.14: 95535073-95535073 on Build GRCh38
rs554961847 Chr.14: 95535074-95535074 on Build GRCh38
rs1018019243 Chr.14: 95534953-95534953 on Build GRCh38
rs1013848133 Chr.14: 95534689-95534689 on Build GRCh38
rs755851239 Chr.14: 95534716-95534716 on Build GRCh38
rs765691081 Chr.14: 95534962-95534962 on Build GRCh38
rs1027121478 Chr.14: 95534710-95534710 on Build GRCh38
rs1042127085 Chr.14: 95534946-95534946 on Build GRCh38
rs1054610664 Chr.14: 95534932-95534932 on Build GRCh38
rs1012597948 Chr.14: 95534995-95534995 on Build GRCh38
rs571025127 Chr.14: 95535033-95535033 on Build GRCh38
rs982012259 Chr.14: 95534654-95534655 on Build GRCh38
rs952828514 Chr.14: 95534927-95534927 on Build GRCh38
rs778338455 Chr.14: 95534814-95534814 on Build GRCh38
rs961588586 Chr.14: 95534703-95534703 on Build GRCh38
rs895043981 Chr.14: 95534983-95534983 on Build GRCh38
rs569336771 Chr.14: 95535062-95535062 on Build GRCh38
rs946439166 Chr.14: 95534941-95534941 on Build GRCh38
rs927771816 Chr.14: 95534655-95534655 on Build GRCh38
rs1041903041 Chr.14: 95535066-95535066 on Build GRCh38
rs913139450 Chr.14: 95534807-95534807 on Build GRCh38
rs573147293 Chr.14: 95534743-95534743 on Build GRCh38
rs914710737 Chr.14: 95535057-95535057 on Build GRCh38
rs2025146 Chr.14: 95534834-95534834 on Build GRCh38
rs972827190 Chr.14: 95534896-95534896 on Build GRCh38
rs548994358 Chr.14: 95534884-95534884 on Build GRCh38
rs572574940 Chr.14: 95534888-95534891 on Build GRCh38
rs563313865 Chr.14: 95534861-95534861 on Build GRCh38
rs1003155925 Chr.14: 95534968-95534968 on Build GRCh38
rs959239856 Chr.14: 95534673-95534673 on Build GRCh38
rs985236413 Chr.14: 95534938-95534938 on Build GRCh38
rs546146190 Chr.14: 95535021-95535021 on Build GRCh38
rs945956481 Chr.14: 95535019-95535019 on Build GRCh38
rs780737253 Chr.14: 95535081-95535081 on Build GRCh38
rs972712615 Chr.14: 95534685-95534685 on Build GRCh38
rs941470771 Chr.14: 95534681-95534681 on Build GRCh38
rs959340656 Chr.14: 95534678-95534678 on Build GRCh38
rs986836212 Chr.14: 95534912-95534912 on Build GRCh38
rs904325414 Chr.14: 95534828-95534828 on Build GRCh38
rs761743525 Chr.14: 95535084-95535084 on Build GRCh38
rs749293851 Chr.14: 95534783-95534783 on Build GRCh38
rs190788920 Chr.14: 95534771-95534771 on Build GRCh38
rs990819733 Chr.14: 95535056-95535056 on Build GRCh38
rs565383676 Chr.14: 95534794-95534794 on Build GRCh38
rs1025729932 Chr.14: 95535035-95535035 on Build GRCh38
rs778132643 Chr.14: 95535050-95535050 on Build GRCh38
rs369520365 Chr.14: 95534867-95534867 on Build GRCh38
rs779466262 Chr.14: 95535043-95535043 on Build GRCh38
rs942878661 Chr.14: 95534752-95534752 on Build GRCh38
rs143059102 Chr.14: 95534626-95534626 on Build GRCh38
rs964303217 Chr.14: 95535051-95535051 on Build GRCh38
rs895177521 Chr.14: 95534866-95534866 on Build GRCh38
rs1027176586 Chr.14: 95534908-95534908 on Build GRCh38
rs542930738 Chr.14: 95534709-95534709 on Build GRCh38
rs985186243 Chr.14: 95534730-95534730 on Build GRCh38
rs976721529 Chr.14: 95534793-95534793 on Build GRCh38
rs978677615 Chr.14: 95535025-95535025 on Build GRCh38
rs747226909 Chr.14: 95535055-95535055 on Build GRCh38
rs911468418 Chr.14: 95534916-95534916 on Build GRCh38
rs902200018 Chr.14: 95534873-95534873 on Build GRCh38

View Full Product Details