Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 101555582-101556082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIO3OS
Entrez Gene ID
64150
Gene Name
DIO3 opposite strand/antisense RNA (head to head)
Gene Aliases
C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
UniGene
Hs.525597
Gene Chromosome Location
Chr.14: 101552223-101555676 on Build GRCh38
Transcripts
NR_002770.1

SNP Detail

SNP ID Chromosome Location
rs570364718 Chr.14: 101555965-101555965 on Build GRCh38
rs61992247 Chr.14: 101555887-101555887 on Build GRCh38
rs111257844 Chr.14: 101555662-101555662 on Build GRCh38
rs869244035 Chr.14: 101555863-101555864 on Build GRCh38
rs181493090 Chr.14: 101555802-101555802 on Build GRCh38
rs562028513 Chr.14: 101555768-101555768 on Build GRCh38
rs866541157 Chr.14: 101555705-101555705 on Build GRCh38
rs562817469 Chr.14: 101555834-101555834 on Build GRCh38
rs201528513 Chr.14: 101555741-101555741 on Build GRCh38
rs555951860 Chr.14: 101555983-101555983 on Build GRCh38
rs1048079172 Chr.14: 101555837-101555837 on Build GRCh38
rs1010263072 Chr.14: 101555895-101555895 on Build GRCh38
rs549736196 Chr.14: 101555915-101555915 on Build GRCh38
rs947779160 Chr.14: 101555870-101555870 on Build GRCh38
rs541040265 Chr.14: 101555788-101555788 on Build GRCh38
rs535642700 Chr.14: 101555982-101555982 on Build GRCh38
rs556420782 Chr.14: 101555963-101555963 on Build GRCh38
rs970298608 Chr.14: 101555771-101555771 on Build GRCh38
rs772469801 Chr.14: 101555844-101555849 on Build GRCh38
rs546949681 Chr.14: 101555930-101555930 on Build GRCh38
rs888629459 Chr.14: 101555768-101555771 on Build GRCh38
rs148122244 Chr.14: 101555816-101555823 on Build GRCh38
rs570294507 Chr.14: 101556030-101556030 on Build GRCh38
rs527436060 Chr.14: 101555735-101555735 on Build GRCh38
rs202030943 Chr.14: 101555846-101555849 on Build GRCh38
rs774015600 Chr.14: 101555705-101555706 on Build GRCh38
rs199537212 Chr.14: 101555747-101555747 on Build GRCh38
rs767260775 Chr.14: 101555823-101555826 on Build GRCh38
rs918253309 Chr.14: 101555841-101555841 on Build GRCh38
rs1000431253 Chr.14: 101555773-101555773 on Build GRCh38
rs918548703 Chr.14: 101555943-101555943 on Build GRCh38
rs1056180890 Chr.14: 101555755-101555755 on Build GRCh38
rs751520701 Chr.14: 101555741-101555742 on Build GRCh38
rs948103308 Chr.14: 101555639-101555639 on Build GRCh38
rs1046464142 Chr.14: 101555684-101555684 on Build GRCh38
rs553612779 Chr.14: 101555849-101555850 on Build GRCh38
rs955425296 Chr.14: 101555796-101555796 on Build GRCh38
rs755782568 Chr.14: 101555839-101555842 on Build GRCh38
rs994470078 Chr.14: 101555706-101555717 on Build GRCh38
rs1047724269 Chr.14: 101555763-101555763 on Build GRCh38
rs373915220 Chr.14: 101555855-101555856 on Build GRCh38
rs549192983 Chr.14: 101555836-101555839 on Build GRCh38
rs749700872 Chr.14: 101555820-101555826 on Build GRCh38
rs929863141 Chr.14: 101555944-101555944 on Build GRCh38
rs34372384 Chr.14: 101555814-101555815 on Build GRCh38
rs368097699 Chr.14: 101555880-101555887 on Build GRCh38
rs542790605 Chr.14: 101555789-101555790 on Build GRCh38
rs1056529951 Chr.14: 101555851-101555851 on Build GRCh38
rs866420896 Chr.14: 101555751-101555751 on Build GRCh38
rs1038457151 Chr.14: 101555739-101555739 on Build GRCh38
rs988872525 Chr.14: 101555702-101555702 on Build GRCh38
rs896794815 Chr.14: 101555743-101555743 on Build GRCh38
rs879815422 Chr.14: 101555732-101555741 on Build GRCh38
rs576225732 Chr.14: 101555744-101555744 on Build GRCh38
rs61992239 Chr.14: 101555833-101555833 on Build GRCh38
rs1024855433 Chr.14: 101555875-101555875 on Build GRCh38
rs532810154 Chr.14: 101555899-101555899 on Build GRCh38
rs61992238 Chr.14: 101555831-101555831 on Build GRCh38
rs964453998 Chr.14: 101555824-101555825 on Build GRCh38
rs886747067 Chr.14: 101555842-101555842 on Build GRCh38
rs556480970 Chr.14: 101555692-101555692 on Build GRCh38
rs779445057 Chr.14: 101556024-101556024 on Build GRCh38
rs572520358 Chr.14: 101555782-101555782 on Build GRCh38
rs36122549 Chr.14: 101555822-101555823 on Build GRCh38
rs61992246 Chr.14: 101555885-101555885 on Build GRCh38
rs1029792248 Chr.14: 101555791-101555791 on Build GRCh38
rs139698122 Chr.14: 101555887-101555888 on Build GRCh38
rs943510337 Chr.14: 101555757-101555757 on Build GRCh38
rs1029654704 Chr.14: 101555903-101555903 on Build GRCh38
rs978762182 Chr.14: 101555630-101555630 on Build GRCh38
rs930991408 Chr.14: 101555835-101555835 on Build GRCh38
rs899578376 Chr.14: 101555815-101555815 on Build GRCh38
rs577077012 Chr.14: 101555627-101555627 on Build GRCh38
rs532405678 Chr.14: 101555822-101555822 on Build GRCh38
rs1004152302 Chr.14: 101555843-101555843 on Build GRCh38
rs1049588361 Chr.14: 101555729-101555729 on Build GRCh38
rs148911882 Chr.14: 101555678-101555678 on Build GRCh38
rs879654683 Chr.14: 101555841-101555842 on Build GRCh38
rs77864095 Chr.14: 101555761-101555761 on Build GRCh38
rs61992243 Chr.14: 101555855-101555855 on Build GRCh38
rs1044562873 Chr.14: 101555871-101555871 on Build GRCh38
rs71305092 Chr.14: 101555740-101555741 on Build GRCh38
rs954373500 Chr.14: 101555952-101555952 on Build GRCh38
rs774254965 Chr.14: 101555836-101555842 on Build GRCh38
rs61992242 Chr.14: 101555853-101555853 on Build GRCh38
rs985318387 Chr.14: 101555801-101555801 on Build GRCh38
rs66885129 Chr.14: 101555820-101555823 on Build GRCh38
rs539092862 Chr.14: 101555960-101555960 on Build GRCh38
rs112968797 Chr.14: 101555739-101555740 on Build GRCh38
rs531383102 Chr.14: 101555850-101555850 on Build GRCh38
rs777384851 Chr.14: 101555740-101555747 on Build GRCh38
rs61992245 Chr.14: 101555879-101555879 on Build GRCh38
rs71464690 Chr.14: 101555916-101555917 on Build GRCh38
rs370895982 Chr.14: 101555799-101555799 on Build GRCh38
rs1000863834 Chr.14: 101555886-101555886 on Build GRCh38
rs868758352 Chr.14: 101555737-101555737 on Build GRCh38
rs61992244 Chr.14: 101555877-101555877 on Build GRCh38
rs376563298 Chr.14: 101555909-101555909 on Build GRCh38
rs139954724 Chr.14: 101556023-101556023 on Build GRCh38
rs933847528 Chr.14: 101555715-101555715 on Build GRCh38
rs558924460 Chr.14: 101555931-101555932 on Build GRCh38
rs985517955 Chr.14: 101555923-101555923 on Build GRCh38
rs973975124 Chr.14: 101555839-101555839 on Build GRCh38
rs527385569 Chr.14: 101555891-101555891 on Build GRCh38
rs903604716 Chr.14: 101555990-101555990 on Build GRCh38
rs71116843 Chr.14: 101555907-101555908 on Build GRCh38
rs957908552 Chr.14: 101555970-101555970 on Build GRCh38
rs879536216 Chr.14: 101555831-101555832 on Build GRCh38
rs61992241 Chr.14: 101555849-101555849 on Build GRCh38
rs570105076 Chr.14: 101555935-101555935 on Build GRCh38
rs896231345 Chr.14: 101555758-101555758 on Build GRCh38
rs1018123202 Chr.14: 101555924-101555924 on Build GRCh38
rs61992240 Chr.14: 101555847-101555847 on Build GRCh38
rs991853934 Chr.14: 101555962-101555962 on Build GRCh38
rs1033432211 Chr.14: 101555780-101555780 on Build GRCh38

View Full Product Details