Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2659586-2660099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVK13-1
Entrez Gene ID
100507321
Gene Name
endogenous retrovirus group K13 member 1
Gene Aliases
-
UniGene
Hs.406976
Gene Chromosome Location
Chr.16: 2658389-2673439 on Build GRCh38
Transcripts
NR_040023.1

SNP Detail

SNP ID Chromosome Location
rs182510412 Chr.16: 2659807-2659807 on Build GRCh38
rs140853650 Chr.16: 2659686-2659686 on Build GRCh38
rs1043131567 Chr.16: 2659778-2659778 on Build GRCh38
rs947371465 Chr.16: 2659770-2659770 on Build GRCh38
rs372657023 Chr.16: 2659704-2659704 on Build GRCh38
rs988718737 Chr.16: 2659765-2659765 on Build GRCh38
rs1005564182 Chr.16: 2659864-2659869 on Build GRCh38
rs772582613 Chr.16: 2659735-2659735 on Build GRCh38
rs529505683 Chr.16: 2659678-2659678 on Build GRCh38
rs896820033 Chr.16: 2659905-2659905 on Build GRCh38
rs111882857 Chr.16: 2659727-2659728 on Build GRCh38
rs980021251 Chr.16: 2659666-2659666 on Build GRCh38
rs1001274145 Chr.16: 2659955-2659955 on Build GRCh38
rs995215504 Chr.16: 2659909-2659909 on Build GRCh38
rs1030450155 Chr.16: 2659718-2659718 on Build GRCh38
rs761379555 Chr.16: 2660011-2660011 on Build GRCh38
rs900682229 Chr.16: 2659709-2659709 on Build GRCh38
rs961447631 Chr.16: 2659630-2659630 on Build GRCh38
rs71148145 Chr.16: 2660029-2660030 on Build GRCh38
rs989138187 Chr.16: 2659973-2659973 on Build GRCh38
rs891784720 Chr.16: 2659687-2659687 on Build GRCh38
rs563897785 Chr.16: 2659631-2659631 on Build GRCh38
rs370542737 Chr.16: 2659746-2659746 on Build GRCh38
rs192746351 Chr.16: 2659769-2659769 on Build GRCh38
rs192122987 Chr.16: 2659979-2659979 on Build GRCh38
rs1026647618 Chr.16: 2659949-2659949 on Build GRCh38
rs187734930 Chr.16: 2659739-2659739 on Build GRCh38
rs535205111 Chr.16: 2659854-2659854 on Build GRCh38
rs544067539 Chr.16: 2660059-2660059 on Build GRCh38
rs996267616 Chr.16: 2659710-2659710 on Build GRCh38
rs377345146 Chr.16: 2660075-2660075 on Build GRCh38
rs1051774459 Chr.16: 2660036-2660036 on Build GRCh38
rs947473935 Chr.16: 2659696-2659696 on Build GRCh38
rs973819795 Chr.16: 2659633-2659633 on Build GRCh38
rs551776798 Chr.16: 2659734-2659734 on Build GRCh38
rs187546911 Chr.16: 2659950-2659950 on Build GRCh38
rs549735450 Chr.16: 2659682-2659682 on Build GRCh38
rs373070426 Chr.16: 2659900-2659900 on Build GRCh38
rs969786729 Chr.16: 2659738-2659738 on Build GRCh38
rs1043123736 Chr.16: 2659703-2659703 on Build GRCh38
rs116392201 Chr.16: 2659853-2659853 on Build GRCh38
rs922055427 Chr.16: 2659791-2659791 on Build GRCh38
rs7190820 Chr.16: 2659660-2659660 on Build GRCh38
rs149712661 Chr.16: 2660006-2660006 on Build GRCh38
rs978771811 Chr.16: 2660014-2660014 on Build GRCh38
rs1005121440 Chr.16: 2659722-2659722 on Build GRCh38
rs756868528 Chr.16: 2660071-2660071 on Build GRCh38
rs921922854 Chr.16: 2660016-2660016 on Build GRCh38
rs543783645 Chr.16: 2659626-2659626 on Build GRCh38
rs113236777 Chr.16: 2659728-2659728 on Build GRCh38
rs533062000 Chr.16: 2659653-2659653 on Build GRCh38
rs1015688853 Chr.16: 2659724-2659725 on Build GRCh38
rs1036576913 Chr.16: 2659899-2659899 on Build GRCh38
rs112303566 Chr.16: 2660022-2660022 on Build GRCh38
rs1035538173 Chr.16: 2659957-2659957 on Build GRCh38
rs960260000 Chr.16: 2659756-2659756 on Build GRCh38
rs112258297 Chr.16: 2660037-2660037 on Build GRCh38
rs890730218 Chr.16: 2659722-2659724 on Build GRCh38
rs560739150 Chr.16: 2659672-2659672 on Build GRCh38
rs919775922 Chr.16: 2659641-2659641 on Build GRCh38

View Full Product Details