Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21300747-21301239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYM
Entrez Gene ID
1428
Gene Name
crystallin mu
Gene Aliases
DFNA40, THBP
UniGene
Hs.924
Gene Chromosome Location
Chr.16: 21258518-21303136 on Build GRCh38
Transcripts
NM_001888.4 NR_026675.1

Gene Symbol
CRYM-AS1
Entrez Gene ID
400508
Gene Name
CRYM antisense RNA 1
Gene Aliases
NCRNA00169
UniGene
Hs.578949
Gene Chromosome Location
Chr.16: 21300849-21318591 on Build GRCh38
Transcripts
NM_001888.4 NR_026675.1

SNP Detail

SNP ID Chromosome Location
rs967398091 Chr.16: 21301143-21301143 on Build GRCh38
rs925455345 Chr.16: 21301165-21301165 on Build GRCh38
rs945965052 Chr.16: 21300946-21300946 on Build GRCh38
rs192089586 Chr.16: 21300885-21300885 on Build GRCh38
rs925447032 Chr.16: 21300893-21300893 on Build GRCh38
rs771496360 Chr.16: 21300987-21300987 on Build GRCh38
rs541936482 Chr.16: 21300886-21300886 on Build GRCh38
rs1024034505 Chr.16: 21300873-21300873 on Build GRCh38
rs746427183 Chr.16: 21301062-21301062 on Build GRCh38
rs945860545 Chr.16: 21300944-21300944 on Build GRCh38
rs962489449 Chr.16: 21300792-21300792 on Build GRCh38
rs143385344 Chr.16: 21301065-21301065 on Build GRCh38
rs546140830 Chr.16: 21301067-21301067 on Build GRCh38
rs986581795 Chr.16: 21300967-21300967 on Build GRCh38
rs78095551 Chr.16: 21300902-21300902 on Build GRCh38
rs1033401873 Chr.16: 21301090-21301090 on Build GRCh38
rs226059 Chr.16: 21301139-21301139 on Build GRCh38
rs992437286 Chr.16: 21300858-21300858 on Build GRCh38
rs76719139 Chr.16: 21300970-21300970 on Build GRCh38
rs563855254 Chr.16: 21300935-21300935 on Build GRCh38
rs921985795 Chr.16: 21301199-21301199 on Build GRCh38
rs1039052415 Chr.16: 21300927-21300927 on Build GRCh38
rs74012160 Chr.16: 21301124-21301124 on Build GRCh38
rs935706880 Chr.16: 21301214-21301214 on Build GRCh38
rs1039562837 Chr.16: 21301011-21301011 on Build GRCh38
rs74012159 Chr.16: 21300876-21300876 on Build GRCh38
rs155931 Chr.16: 21300837-21300837 on Build GRCh38
rs974809909 Chr.16: 21301187-21301187 on Build GRCh38
rs34308235 Chr.16: 21301015-21301016 on Build GRCh38
rs936805483 Chr.16: 21301167-21301167 on Build GRCh38
rs567890425 Chr.16: 21301154-21301154 on Build GRCh38
rs891067501 Chr.16: 21301006-21301006 on Build GRCh38
rs1015478146 Chr.16: 21301057-21301057 on Build GRCh38
rs886568384 Chr.16: 21300795-21300795 on Build GRCh38
rs12447154 Chr.16: 21300775-21300775 on Build GRCh38
rs1005167966 Chr.16: 21301091-21301091 on Build GRCh38
rs959265439 Chr.16: 21300855-21300855 on Build GRCh38
rs528334417 Chr.16: 21301136-21301136 on Build GRCh38
rs992256684 Chr.16: 21301131-21301131 on Build GRCh38
rs973816742 Chr.16: 21300836-21300836 on Build GRCh38
rs995262893 Chr.16: 21301066-21301066 on Build GRCh38
rs1024462795 Chr.16: 21301099-21301099 on Build GRCh38
rs990049947 Chr.16: 21300913-21300913 on Build GRCh38
rs397827381 Chr.16: 21301018-21301019 on Build GRCh38
rs979874440 Chr.16: 21300878-21300878 on Build GRCh38
rs577565257 Chr.16: 21300782-21300782 on Build GRCh38
rs541768515 Chr.16: 21300820-21300820 on Build GRCh38
rs1015425984 Chr.16: 21300776-21300776 on Build GRCh38
rs529131105 Chr.16: 21301179-21301179 on Build GRCh38
rs957297690 Chr.16: 21300908-21300909 on Build GRCh38
rs930640221 Chr.16: 21301021-21301021 on Build GRCh38
rs907410165 Chr.16: 21301107-21301107 on Build GRCh38
rs1004128096 Chr.16: 21301031-21301031 on Build GRCh38
rs766169644 Chr.16: 21301097-21301097 on Build GRCh38
rs995089696 Chr.16: 21300767-21300767 on Build GRCh38
rs1034524942 Chr.16: 21301126-21301126 on Build GRCh38

View Full Product Details