Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21301077-21301584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYM
Entrez Gene ID
1428
Gene Name
crystallin mu
Gene Aliases
DFNA40, THBP
UniGene
Hs.924
Gene Chromosome Location
Chr.16: 21258518-21303136 on Build GRCh38
Transcripts
NM_001888.4 NR_026675.1

Gene Symbol
CRYM-AS1
Entrez Gene ID
400508
Gene Name
CRYM antisense RNA 1
Gene Aliases
NCRNA00169
UniGene
Hs.578949
Gene Chromosome Location
Chr.16: 21300849-21318591 on Build GRCh38
Transcripts
NM_001888.4 NR_026675.1

SNP Detail

SNP ID Chromosome Location
rs894976619 Chr.16: 21301515-21301515 on Build GRCh38
rs967398091 Chr.16: 21301143-21301143 on Build GRCh38
rs564121174 Chr.16: 21301507-21301507 on Build GRCh38
rs1036916434 Chr.16: 21301262-21301262 on Build GRCh38
rs922028411 Chr.16: 21301238-21301238 on Build GRCh38
rs187766804 Chr.16: 21301421-21301421 on Build GRCh38
rs925455345 Chr.16: 21301165-21301165 on Build GRCh38
rs553731657 Chr.16: 21301395-21301395 on Build GRCh38
rs545994870 Chr.16: 21301474-21301474 on Build GRCh38
rs975664004 Chr.16: 21301420-21301420 on Build GRCh38
rs226058 Chr.16: 21301362-21301362 on Build GRCh38
rs542453290 Chr.16: 21301408-21301408 on Build GRCh38
rs966361248 Chr.16: 21301361-21301361 on Build GRCh38
rs75541665 Chr.16: 21301310-21301310 on Build GRCh38
rs557709338 Chr.16: 21301374-21301374 on Build GRCh38
rs1025100190 Chr.16: 21301343-21301343 on Build GRCh38
rs1056348769 Chr.16: 21301513-21301514 on Build GRCh38
rs907868658 Chr.16: 21301458-21301458 on Build GRCh38
rs933408868 Chr.16: 21301239-21301239 on Build GRCh38
rs374384309 Chr.16: 21301327-21301327 on Build GRCh38
rs867576853 Chr.16: 21301371-21301371 on Build GRCh38
rs1046111317 Chr.16: 21301518-21301518 on Build GRCh38
rs1002114320 Chr.16: 21301287-21301287 on Build GRCh38
rs115162868 Chr.16: 21301550-21301550 on Build GRCh38
rs226059 Chr.16: 21301139-21301139 on Build GRCh38
rs892891678 Chr.16: 21301329-21301329 on Build GRCh38
rs369243202 Chr.16: 21301258-21301258 on Build GRCh38
rs370858127 Chr.16: 21301492-21301492 on Build GRCh38
rs1033493558 Chr.16: 21301286-21301286 on Build GRCh38
rs921985795 Chr.16: 21301199-21301199 on Build GRCh38
rs150513547 Chr.16: 21301405-21301405 on Build GRCh38
rs919865976 Chr.16: 21301488-21301488 on Build GRCh38
rs74012160 Chr.16: 21301124-21301124 on Build GRCh38
rs935706880 Chr.16: 21301214-21301214 on Build GRCh38
rs777027139 Chr.16: 21301309-21301309 on Build GRCh38
rs1045978953 Chr.16: 21301477-21301477 on Build GRCh38
rs967049326 Chr.16: 21301379-21301379 on Build GRCh38
rs141614390 Chr.16: 21301393-21301393 on Build GRCh38
rs974809909 Chr.16: 21301187-21301187 on Build GRCh38
rs182640587 Chr.16: 21301252-21301252 on Build GRCh38
rs886197483 Chr.16: 21301387-21301408 on Build GRCh38
rs936805483 Chr.16: 21301167-21301167 on Build GRCh38
rs567890425 Chr.16: 21301154-21301154 on Build GRCh38
rs1013754871 Chr.16: 21301516-21301516 on Build GRCh38
rs952211191 Chr.16: 21301449-21301449 on Build GRCh38
rs1036970388 Chr.16: 21301487-21301487 on Build GRCh38
rs995316698 Chr.16: 21301277-21301277 on Build GRCh38
rs1031948302 Chr.16: 21301552-21301552 on Build GRCh38
rs528334417 Chr.16: 21301136-21301136 on Build GRCh38
rs992256684 Chr.16: 21301131-21301131 on Build GRCh38
rs182625697 Chr.16: 21301456-21301456 on Build GRCh38
rs155932 Chr.16: 21301486-21301486 on Build GRCh38
rs1024462795 Chr.16: 21301099-21301099 on Build GRCh38
rs1039451615 Chr.16: 21301225-21301225 on Build GRCh38
rs1055401329 Chr.16: 21301514-21301514 on Build GRCh38
rs942544717 Chr.16: 21301223-21301224 on Build GRCh38
rs919797970 Chr.16: 21301423-21301423 on Build GRCh38
rs529131105 Chr.16: 21301179-21301179 on Build GRCh38
rs907410165 Chr.16: 21301107-21301107 on Build GRCh38
rs928891868 Chr.16: 21301503-21301503 on Build GRCh38
rs955001806 Chr.16: 21301396-21301396 on Build GRCh38
rs372282239 Chr.16: 21301519-21301519 on Build GRCh38
rs886440904 Chr.16: 21301256-21301256 on Build GRCh38
rs557284140 Chr.16: 21301415-21301415 on Build GRCh38
rs931205343 Chr.16: 21301511-21301511 on Build GRCh38
rs375967280 Chr.16: 21301420-21301421 on Build GRCh38
rs974815101 Chr.16: 21301224-21301224 on Build GRCh38
rs148383611 Chr.16: 21301383-21301383 on Build GRCh38
rs535321372 Chr.16: 21301385-21301385 on Build GRCh38
rs766169644 Chr.16: 21301097-21301097 on Build GRCh38
rs958215664 Chr.16: 21301376-21301376 on Build GRCh38
rs74012161 Chr.16: 21301271-21301271 on Build GRCh38
rs1034524942 Chr.16: 21301126-21301126 on Build GRCh38
rs191876673 Chr.16: 21301430-21301430 on Build GRCh38

View Full Product Details