Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 54918757-54919258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRNDE
Entrez Gene ID
643911
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
Gene Aliases
CRNDEP, LINC00180, NCRNA00180, PNAS-108, lincIRX5
UniGene
Hs.237396
Gene Chromosome Location
Chr.16: 54918863-54929189 on Build GRCh38
Transcripts
NM_001308963.1 NR_034105.3 NR_034106.2 NR_110453.1 NR_110454.1 XM_017023562.1 XM_017023563.1

SNP Detail

SNP ID Chromosome Location
rs184419940 Chr.16: 54919027-54919027 on Build GRCh38
rs549490600 Chr.16: 54918879-54918879 on Build GRCh38
rs985477413 Chr.16: 54918841-54918841 on Build GRCh38
rs1010806237 Chr.16: 54919068-54919068 on Build GRCh38
rs930178486 Chr.16: 54919183-54919183 on Build GRCh38
rs906273412 Chr.16: 54918965-54918965 on Build GRCh38
rs1029677742 Chr.16: 54918809-54918809 on Build GRCh38
rs546065826 Chr.16: 54918791-54918791 on Build GRCh38
rs1018245525 Chr.16: 54919151-54919151 on Build GRCh38
rs1047435051 Chr.16: 54918962-54918962 on Build GRCh38
rs550827978 Chr.16: 54919217-54919217 on Build GRCh38
rs188796926 Chr.16: 54919201-54919201 on Build GRCh38
rs79203900 Chr.16: 54919225-54919226 on Build GRCh38
rs974255649 Chr.16: 54919168-54919168 on Build GRCh38
rs974224401 Chr.16: 54918900-54918900 on Build GRCh38
rs200015173 Chr.16: 54918784-54918787 on Build GRCh38
rs567771465 Chr.16: 54919148-54919148 on Build GRCh38
rs148646307 Chr.16: 54919204-54919204 on Build GRCh38
rs115515594 Chr.16: 54919202-54919202 on Build GRCh38
rs918762328 Chr.16: 54919178-54919178 on Build GRCh38
rs1029793737 Chr.16: 54919135-54919135 on Build GRCh38
rs955595412 Chr.16: 54918829-54918829 on Build GRCh38
rs770089003 Chr.16: 54918950-54918950 on Build GRCh38
rs1022142130 Chr.16: 54919107-54919107 on Build GRCh38
rs188692108 Chr.16: 54918896-54918896 on Build GRCh38
rs191597929 Chr.16: 54918957-54918957 on Build GRCh38
rs540492107 Chr.16: 54918917-54918917 on Build GRCh38
rs999354259 Chr.16: 54919132-54919132 on Build GRCh38
rs373443202 Chr.16: 54919226-54919226 on Build GRCh38
rs902314632 Chr.16: 54919120-54919120 on Build GRCh38
rs966803963 Chr.16: 54918783-54918783 on Build GRCh38
rs541993151 Chr.16: 54919055-54919055 on Build GRCh38
rs552469560 Chr.16: 54919150-54919150 on Build GRCh38
rs546558199 Chr.16: 54919193-54919193 on Build GRCh38
rs939039346 Chr.16: 54919045-54919045 on Build GRCh38
rs748334957 Chr.16: 54918945-54918945 on Build GRCh38
rs563253266 Chr.16: 54919056-54919056 on Build GRCh38
rs535323882 Chr.16: 54918924-54918924 on Build GRCh38
rs748196780 Chr.16: 54918863-54918863 on Build GRCh38
rs568211832 Chr.16: 54918874-54918875 on Build GRCh38
rs143939661 Chr.16: 54919159-54919159 on Build GRCh38

View Full Product Details