Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 54924963-54925460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRNDE
Entrez Gene ID
643911
Gene Name
colorectal neoplasia differentially expressed (non-protein coding)
Gene Aliases
CRNDEP, LINC00180, NCRNA00180, PNAS-108, lincIRX5
UniGene
Hs.237396
Gene Chromosome Location
Chr.16: 54918863-54929189 on Build GRCh38
Transcripts
NM_001308963.1 NR_034105.3 NR_034106.2 NR_110453.1 NR_110454.1 XM_017023562.1 XM_017023563.1

SNP Detail

SNP ID Chromosome Location
rs962891305 Chr.16: 54925345-54925345 on Build GRCh38
rs767481017 Chr.16: 54925401-54925401 on Build GRCh38
rs112523309 Chr.16: 54925083-54925083 on Build GRCh38
rs574523365 Chr.16: 54925233-54925233 on Build GRCh38
rs1022793816 Chr.16: 54925023-54925023 on Build GRCh38
rs999914454 Chr.16: 54925040-54925040 on Build GRCh38
rs1010960167 Chr.16: 54925019-54925019 on Build GRCh38
rs557244484 Chr.16: 54925254-54925254 on Build GRCh38
rs999543683 Chr.16: 54925279-54925279 on Build GRCh38
rs966774265 Chr.16: 54925027-54925027 on Build GRCh38
rs548532172 Chr.16: 54925054-54925054 on Build GRCh38
rs765242105 Chr.16: 54925012-54925012 on Build GRCh38
rs151201998 Chr.16: 54925246-54925246 on Build GRCh38
rs973214232 Chr.16: 54925136-54925136 on Build GRCh38
rs766529953 Chr.16: 54925252-54925252 on Build GRCh38
rs1018360458 Chr.16: 54925109-54925109 on Build GRCh38
rs183873262 Chr.16: 54925305-54925305 on Build GRCh38
rs530012752 Chr.16: 54925052-54925052 on Build GRCh38
rs1029593636 Chr.16: 54925300-54925300 on Build GRCh38
rs201723508 Chr.16: 54925021-54925021 on Build GRCh38
rs142148268 Chr.16: 54925165-54925165 on Build GRCh38
rs955592083 Chr.16: 54925076-54925076 on Build GRCh38
rs139730954 Chr.16: 54925402-54925402 on Build GRCh38
rs983508978 Chr.16: 54925181-54925181 on Build GRCh38
rs754765462 Chr.16: 54925364-54925364 on Build GRCh38
rs964440268 Chr.16: 54925114-54925114 on Build GRCh38
rs974629481 Chr.16: 54925350-54925350 on Build GRCh38
rs950491461 Chr.16: 54925393-54925393 on Build GRCh38
rs1018475329 Chr.16: 54925325-54925325 on Build GRCh38
rs565718464 Chr.16: 54924986-54924986 on Build GRCh38
rs939228259 Chr.16: 54925213-54925213 on Build GRCh38
rs773282745 Chr.16: 54925095-54925095 on Build GRCh38
rs116512596 Chr.16: 54925137-54925137 on Build GRCh38
rs573135152 Chr.16: 54925153-54925153 on Build GRCh38
rs946539956 Chr.16: 54925221-54925221 on Build GRCh38
rs990775043 Chr.16: 54925436-54925436 on Build GRCh38
rs777127667 Chr.16: 54925103-54925104 on Build GRCh38
rs552949284 Chr.16: 54925220-54925220 on Build GRCh38
rs535162567 Chr.16: 54925184-54925184 on Build GRCh38
rs1054972888 Chr.16: 54925219-54925219 on Build GRCh38
rs756127900 Chr.16: 54925418-54925418 on Build GRCh38
rs1007064407 Chr.16: 54925315-54925315 on Build GRCh38
rs938989791 Chr.16: 54925412-54925412 on Build GRCh38
rs534558323 Chr.16: 54925416-54925416 on Build GRCh38

View Full Product Details