Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86478230-86478752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FENDRR
Entrez Gene ID
400550
Gene Name
FOXF1 adjacent non-coding developmental regulatory RNA
Gene Aliases
FOXF1-AS1, TCONS_00024240, lincFOXF1, onco-lncRNA-21
UniGene
Hs.448825
Gene Chromosome Location
Chr.16: 86474525-86508860 on Build GRCh38
Transcripts
NR_033925.1

SNP Detail

SNP ID Chromosome Location
rs974739278 Chr.16: 86478489-86478489 on Build GRCh38
rs983959069 Chr.16: 86478696-86478696 on Build GRCh38
rs919752951 Chr.16: 86478693-86478693 on Build GRCh38
rs1025192152 Chr.16: 86478586-86478586 on Build GRCh38
rs954484050 Chr.16: 86478307-86478307 on Build GRCh38
rs1015310672 Chr.16: 86478414-86478414 on Build GRCh38
rs569759726 Chr.16: 86478505-86478505 on Build GRCh38
rs1042465489 Chr.16: 86478327-86478327 on Build GRCh38
rs7187191 Chr.16: 86478252-86478252 on Build GRCh38
rs541357836 Chr.16: 86478619-86478619 on Build GRCh38
rs970262285 Chr.16: 86478681-86478681 on Build GRCh38
rs556272434 Chr.16: 86478331-86478331 on Build GRCh38
rs13335567 Chr.16: 86478716-86478716 on Build GRCh38
rs139883239 Chr.16: 86478422-86478422 on Build GRCh38
rs1032455310 Chr.16: 86478619-86478619 on Build GRCh38
rs756002079 Chr.16: 86478585-86478585 on Build GRCh38
rs527599594 Chr.16: 86478368-86478368 on Build GRCh38
rs74035784 Chr.16: 86478461-86478461 on Build GRCh38
rs753188446 Chr.16: 86478587-86478587 on Build GRCh38
rs775723117 Chr.16: 86478358-86478358 on Build GRCh38
rs899731606 Chr.16: 86478370-86478370 on Build GRCh38
rs959865522 Chr.16: 86478350-86478350 on Build GRCh38
rs559626078 Chr.16: 86478340-86478340 on Build GRCh38
rs546094705 Chr.16: 86478702-86478702 on Build GRCh38
rs998473717 Chr.16: 86478633-86478633 on Build GRCh38
rs1004324036 Chr.16: 86478258-86478258 on Build GRCh38
rs922705240 Chr.16: 86478583-86478583 on Build GRCh38
rs568224637 Chr.16: 86478576-86478576 on Build GRCh38
rs764062555 Chr.16: 86478434-86478434 on Build GRCh38
rs890033215 Chr.16: 86478463-86478463 on Build GRCh38
rs998770937 Chr.16: 86478284-86478284 on Build GRCh38
rs1036887148 Chr.16: 86478686-86478686 on Build GRCh38
rs771371323 Chr.16: 86478714-86478714 on Build GRCh38
rs551710994 Chr.16: 86478705-86478705 on Build GRCh38
rs565147250 Chr.16: 86478351-86478351 on Build GRCh38
rs8054409 Chr.16: 86478618-86478618 on Build GRCh38
rs147284842 Chr.16: 86478508-86478508 on Build GRCh38
rs143825099 Chr.16: 86478490-86478490 on Build GRCh38
rs750191037 Chr.16: 86478567-86478567 on Build GRCh38
rs890960291 Chr.16: 86478399-86478401 on Build GRCh38
rs185175162 Chr.16: 86478347-86478347 on Build GRCh38
rs909862073 Chr.16: 86478400-86478400 on Build GRCh38
rs1023243796 Chr.16: 86478526-86478526 on Build GRCh38
rs904166671 Chr.16: 86478486-86478486 on Build GRCh38
rs375385405 Chr.16: 86478289-86478289 on Build GRCh38
rs894809951 Chr.16: 86478559-86478559 on Build GRCh38
rs1051017712 Chr.16: 86478513-86478513 on Build GRCh38
rs995244779 Chr.16: 86478546-86478549 on Build GRCh38
rs149833868 Chr.16: 86478668-86478668 on Build GRCh38
rs545386702 Chr.16: 86478573-86478573 on Build GRCh38
rs1048407799 Chr.16: 86478399-86478399 on Build GRCh38
rs889926066 Chr.16: 86478605-86478605 on Build GRCh38
rs889595 Chr.16: 86478474-86478474 on Build GRCh38
rs898221788 Chr.16: 86478538-86478538 on Build GRCh38
rs1031786959 Chr.16: 86478660-86478660 on Build GRCh38
rs940017717 Chr.16: 86478408-86478408 on Build GRCh38
rs984010915 Chr.16: 86478552-86478552 on Build GRCh38
rs12934218 Chr.16: 86478524-86478524 on Build GRCh38
rs992417885 Chr.16: 86478441-86478441 on Build GRCh38
rs969935784 Chr.16: 86478550-86478550 on Build GRCh38
rs895553705 Chr.16: 86478667-86478667 on Build GRCh38
rs933295892 Chr.16: 86478655-86478655 on Build GRCh38
rs74387953 Chr.16: 86478612-86478612 on Build GRCh38
rs1021272894 Chr.16: 86478317-86478317 on Build GRCh38
rs118025818 Chr.16: 86478560-86478560 on Build GRCh38
rs189825218 Chr.16: 86478430-86478430 on Build GRCh38
rs563738678 Chr.16: 86478471-86478471 on Build GRCh38
rs1025596495 Chr.16: 86478451-86478451 on Build GRCh38
rs909809865 Chr.16: 86478556-86478556 on Build GRCh38
rs7187365 Chr.16: 86478309-86478309 on Build GRCh38
rs972771195 Chr.16: 86478581-86478581 on Build GRCh38
rs534222643 Chr.16: 86478487-86478487 on Build GRCh38
rs1037137346 Chr.16: 86478259-86478259 on Build GRCh38
rs561182930 Chr.16: 86478396-86478396 on Build GRCh38
rs890955207 Chr.16: 86478515-86478515 on Build GRCh38
rs1033382212 Chr.16: 86478557-86478557 on Build GRCh38
rs1036793482 Chr.16: 86478532-86478532 on Build GRCh38
rs921975139 Chr.16: 86478651-86478651 on Build GRCh38
rs533495966 Chr.16: 86478361-86478361 on Build GRCh38
rs1047105163 Chr.16: 86478719-86478719 on Build GRCh38
rs932572118 Chr.16: 86478387-86478387 on Build GRCh38
rs543276218 Chr.16: 86478675-86478675 on Build GRCh38
rs1049900732 Chr.16: 86478593-86478593 on Build GRCh38
rs776914830 Chr.16: 86478281-86478281 on Build GRCh38
rs191900668 Chr.16: 86478433-86478433 on Build GRCh38
rs563012955 Chr.16: 86478682-86478682 on Build GRCh38
rs147206814 Chr.16: 86478388-86478388 on Build GRCh38
rs565976040 Chr.16: 86478630-86478630 on Build GRCh38

View Full Product Details