Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15030877-15031386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT7
Entrez Gene ID
94150
Gene Name
CMT1A duplicated region transcript 7 (non-protein coding)
Gene Aliases
LINC00025, NCRNA00025
UniGene
Hs.147654
Gene Chromosome Location
Chr.17: 15030975-15031957 on Build GRCh38
Transcripts
NR_033371.1

SNP Detail

SNP ID Chromosome Location
rs8065954 Chr.17: 15031114-15031114 on Build GRCh38
rs116400898 Chr.17: 15031126-15031126 on Build GRCh38
rs1029946668 Chr.17: 15031294-15031294 on Build GRCh38
rs150258623 Chr.17: 15031317-15031317 on Build GRCh38
rs7210338 Chr.17: 15031170-15031170 on Build GRCh38
rs962578533 Chr.17: 15031362-15031362 on Build GRCh38
rs746594610 Chr.17: 15031296-15031296 on Build GRCh38
rs990066146 Chr.17: 15031067-15031067 on Build GRCh38
rs1043381699 Chr.17: 15031224-15031224 on Build GRCh38
rs907841517 Chr.17: 15030907-15030907 on Build GRCh38
rs552966624 Chr.17: 15031192-15031192 on Build GRCh38
rs1012380916 Chr.17: 15031261-15031261 on Build GRCh38
rs7208936 Chr.17: 15030928-15030928 on Build GRCh38
rs1031126036 Chr.17: 15030986-15030986 on Build GRCh38
rs556096428 Chr.17: 15031058-15031058 on Build GRCh38
rs557002114 Chr.17: 15031339-15031339 on Build GRCh38
rs978476341 Chr.17: 15031018-15031018 on Build GRCh38
rs73980953 Chr.17: 15031172-15031172 on Build GRCh38
rs996568480 Chr.17: 15031240-15031240 on Build GRCh38
rs1029463590 Chr.17: 15031066-15031066 on Build GRCh38
rs948203639 Chr.17: 15030953-15030953 on Build GRCh38
rs77198164 Chr.17: 15031205-15031205 on Build GRCh38
rs978190879 Chr.17: 15031302-15031302 on Build GRCh38
rs756552896 Chr.17: 15031052-15031052 on Build GRCh38
rs112228515 Chr.17: 15031347-15031347 on Build GRCh38
rs7210007 Chr.17: 15031015-15031015 on Build GRCh38
rs1045643853 Chr.17: 15031145-15031145 on Build GRCh38
rs1045208277 Chr.17: 15030968-15030968 on Build GRCh38
rs1015446093 Chr.17: 15031356-15031356 on Build GRCh38
rs892128632 Chr.17: 15030993-15030993 on Build GRCh38
rs943199102 Chr.17: 15031084-15031084 on Build GRCh38
rs899590219 Chr.17: 15031232-15031232 on Build GRCh38
rs1025168263 Chr.17: 15031265-15031265 on Build GRCh38
rs971193346 Chr.17: 15031292-15031292 on Build GRCh38
rs899367044 Chr.17: 15030938-15030938 on Build GRCh38
rs1022035907 Chr.17: 15031049-15031049 on Build GRCh38
rs8182247 Chr.17: 15031076-15031076 on Build GRCh38
rs948129867 Chr.17: 15031129-15031129 on Build GRCh38
rs886087521 Chr.17: 15031153-15031153 on Build GRCh38
rs906788002 Chr.17: 15030981-15030981 on Build GRCh38
rs188891014 Chr.17: 15031318-15031318 on Build GRCh38
rs551976315 Chr.17: 15030923-15030923 on Build GRCh38
rs1010596817 Chr.17: 15031214-15031214 on Build GRCh38
rs76110013 Chr.17: 15031325-15031325 on Build GRCh38
rs749659614 Chr.17: 15031120-15031120 on Build GRCh38
rs566534994 Chr.17: 15031002-15031002 on Build GRCh38
rs982714539 Chr.17: 15031083-15031083 on Build GRCh38
rs117144301 Chr.17: 15031098-15031098 on Build GRCh38
rs1052677530 Chr.17: 15031203-15031203 on Build GRCh38
rs973480722 Chr.17: 15031118-15031118 on Build GRCh38
rs959522941 Chr.17: 15030952-15030952 on Build GRCh38
rs1003761670 Chr.17: 15030985-15030985 on Build GRCh38
rs386795760 Chr.17: 15031170-15031172 on Build GRCh38

View Full Product Details