Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 15031606-15032105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDRT7
Entrez Gene ID
94150
Gene Name
CMT1A duplicated region transcript 7 (non-protein coding)
Gene Aliases
LINC00025, NCRNA00025
UniGene
Hs.147654
Gene Chromosome Location
Chr.17: 15030975-15031957 on Build GRCh38
Transcripts
NR_033371.1

SNP Detail

SNP ID Chromosome Location
rs530045627 Chr.17: 15031760-15031761 on Build GRCh38
rs905789849 Chr.17: 15031931-15031931 on Build GRCh38
rs968101044 Chr.17: 15031976-15031976 on Build GRCh38
rs986487506 Chr.17: 15031864-15031864 on Build GRCh38
rs368263270 Chr.17: 15031704-15031704 on Build GRCh38
rs969131205 Chr.17: 15032029-15032029 on Build GRCh38
rs898326208 Chr.17: 15031749-15031749 on Build GRCh38
rs939735522 Chr.17: 15031719-15031719 on Build GRCh38
rs939807722 Chr.17: 15031876-15031876 on Build GRCh38
rs979670049 Chr.17: 15031670-15031670 on Build GRCh38
rs1028874435 Chr.17: 15031995-15031995 on Build GRCh38
rs1002224800 Chr.17: 15031795-15031795 on Build GRCh38
rs890348191 Chr.17: 15031981-15031981 on Build GRCh38
rs979704220 Chr.17: 15031850-15031850 on Build GRCh38
rs954359908 Chr.17: 15032005-15032005 on Build GRCh38
rs893956259 Chr.17: 15031943-15031943 on Build GRCh38
rs946988903 Chr.17: 15031658-15031658 on Build GRCh38
rs1044595837 Chr.17: 15031916-15031916 on Build GRCh38
rs961010576 Chr.17: 15032028-15032028 on Build GRCh38
rs143774588 Chr.17: 15032010-15032010 on Build GRCh38
rs920958275 Chr.17: 15031678-15031678 on Build GRCh38
rs1022866100 Chr.17: 15031760-15031760 on Build GRCh38
rs916237920 Chr.17: 15031654-15031654 on Build GRCh38
rs187189920 Chr.17: 15031819-15031819 on Build GRCh38
rs960823377 Chr.17: 15031813-15031813 on Build GRCh38
rs114468442 Chr.17: 15032082-15032082 on Build GRCh38
rs964011272 Chr.17: 15031824-15031824 on Build GRCh38
rs575203378 Chr.17: 15031979-15031979 on Build GRCh38
rs372131298 Chr.17: 15031809-15031809 on Build GRCh38
rs932316983 Chr.17: 15031687-15031687 on Build GRCh38
rs59627551 Chr.17: 15031858-15031858 on Build GRCh38
rs546913703 Chr.17: 15031915-15031915 on Build GRCh38
rs750689474 Chr.17: 15031816-15031816 on Build GRCh38
rs190464387 Chr.17: 15031862-15031862 on Build GRCh38
rs906812533 Chr.17: 15031756-15031756 on Build GRCh38
rs777205234 Chr.17: 15031957-15031957 on Build GRCh38
rs947697035 Chr.17: 15031901-15031901 on Build GRCh38
rs4523961 Chr.17: 15031860-15031860 on Build GRCh38
rs180914705 Chr.17: 15031866-15031866 on Build GRCh38
rs767640112 Chr.17: 15031782-15031782 on Build GRCh38
rs1036865113 Chr.17: 15031727-15031727 on Build GRCh38
rs145869900 Chr.17: 15031849-15031849 on Build GRCh38
rs75101976 Chr.17: 15031827-15031827 on Build GRCh38
rs995412433 Chr.17: 15031751-15031751 on Build GRCh38
rs8182278 Chr.17: 15031942-15031942 on Build GRCh38
rs946923781 Chr.17: 15031946-15031946 on Build GRCh38
rs912259082 Chr.17: 15032024-15032024 on Build GRCh38
rs919692200 Chr.17: 15032062-15032062 on Build GRCh38
rs753203429 Chr.17: 15031902-15031902 on Build GRCh38
rs756722569 Chr.17: 15031920-15031920 on Build GRCh38
rs898360315 Chr.17: 15031898-15031898 on Build GRCh38
rs973133341 Chr.17: 15031889-15031890 on Build GRCh38
rs1005196044 Chr.17: 15031983-15031983 on Build GRCh38
rs914031928 Chr.17: 15031650-15031650 on Build GRCh38
rs1037343228 Chr.17: 15031641-15031641 on Build GRCh38
rs890891921 Chr.17: 15031712-15031712 on Build GRCh38
rs577811495 Chr.17: 15032065-15032065 on Build GRCh38
rs963206322 Chr.17: 15031998-15031998 on Build GRCh38
rs1030212433 Chr.17: 15031830-15031830 on Build GRCh38
rs1039752571 Chr.17: 15031951-15031951 on Build GRCh38
rs80340954 Chr.17: 15031938-15031938 on Build GRCh38
rs931031864 Chr.17: 15032063-15032063 on Build GRCh38
rs1046682557 Chr.17: 15031710-15031710 on Build GRCh38
rs779305905 Chr.17: 15031895-15031895 on Build GRCh38
rs1044240523 Chr.17: 15032068-15032068 on Build GRCh38
rs905754257 Chr.17: 15031781-15031781 on Build GRCh38
rs1051177715 Chr.17: 15031698-15031698 on Build GRCh38
rs1009680948 Chr.17: 15031968-15031968 on Build GRCh38
rs148971661 Chr.17: 15031974-15031974 on Build GRCh38
rs530255994 Chr.17: 15032043-15032043 on Build GRCh38

View Full Product Details