Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16439921-16440430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC75A-AS1
Entrez Gene ID
125144
Gene Name
LRRC75A antisense RNA 1
Gene Aliases
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
UniGene
Hs.368934
Gene Chromosome Location
Chr.17: 16438987-16470648 on Build GRCh38
Transcripts
NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027179.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045024.1 NR_045025.1 NR_045026.1 NR_045027.1 NR_045028.1 NR_045029.1 NR_002744.1

Gene Symbol
SNORD49A
Entrez Gene ID
26800
Gene Name
small nucleolar RNA, C/D box 49A
Gene Aliases
RNU49, U49, U49A
UniGene
Gene Chromosome Location
Chr.17: 16440036-16440106 on Build GRCh38
Transcripts
NR_027158.1 NR_027159.1 NR_027160.1 NR_027161.1 NR_027162.1 NR_027163.1 NR_027164.1 NR_027165.1 NR_027166.1 NR_027167.1 NR_027168.2 NR_027169.1 NR_027170.1 NR_027171.1 NR_027172.2 NR_027173.1 NR_027174.1 NR_027175.1 NR_027176.1 NR_027177.1 NR_027178.1 NR_027179.1 NR_027667.1 NR_045021.1 NR_045022.1 NR_045023.1 NR_045024.1 NR_045025.1 NR_045026.1 NR_045027.1 NR_045028.1 NR_045029.1 NR_002744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172382 c.362A>G p.Y121C Chr.17: 16440229-16440229 on Build GRCh38
7272952 c.325A>G p.I109V Chr.17: 16440192-16440192 on Build GRCh38
5758293 c.329G>A p.R110H Chr.17: 16440196-16440196 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750920579 Chr.17: 16440173-16440173 on Build GRCh38
rs769768006 Chr.17: 16440067-16440067 on Build GRCh38
rs750229460 Chr.17: 16439994-16439994 on Build GRCh38
rs903473606 Chr.17: 16440307-16440307 on Build GRCh38
rs969188291 Chr.17: 16440013-16440013 on Build GRCh38
rs759258701 Chr.17: 16440088-16440088 on Build GRCh38
rs957473627 Chr.17: 16440122-16440126 on Build GRCh38
rs773102772 Chr.17: 16440120-16440120 on Build GRCh38
rs748247756 Chr.17: 16440113-16440113 on Build GRCh38
rs112824075 Chr.17: 16440183-16440183 on Build GRCh38
rs774213377 Chr.17: 16440138-16440138 on Build GRCh38
rs760206686 Chr.17: 16440163-16440163 on Build GRCh38
rs776293455 Chr.17: 16440256-16440256 on Build GRCh38
rs548906956 Chr.17: 16440376-16440376 on Build GRCh38
rs986248730 Chr.17: 16440127-16440127 on Build GRCh38
rs1048790427 Chr.17: 16440209-16440209 on Build GRCh38
rs771100512 Chr.17: 16440097-16440100 on Build GRCh38
rs769911904 Chr.17: 16440114-16440114 on Build GRCh38
rs1034771342 Chr.17: 16440341-16440341 on Build GRCh38
rs987841412 Chr.17: 16440345-16440345 on Build GRCh38
rs765823391 Chr.17: 16440099-16440099 on Build GRCh38
rs749258947 Chr.17: 16440128-16440128 on Build GRCh38
rs999043274 Chr.17: 16440315-16440315 on Build GRCh38
rs147803224 Chr.17: 16440009-16440009 on Build GRCh38
rs773640815 Chr.17: 16440046-16440046 on Build GRCh38
rs780596147 Chr.17: 16440106-16440106 on Build GRCh38
rs545518399 Chr.17: 16440215-16440215 on Build GRCh38
rs775173379 Chr.17: 16440249-16440249 on Build GRCh38
rs562319300 Chr.17: 16440389-16440389 on Build GRCh38
rs563089420 Chr.17: 16440137-16440137 on Build GRCh38
rs781064592 Chr.17: 16440111-16440111 on Build GRCh38
rs1007936870 Chr.17: 16440349-16440349 on Build GRCh38
rs1027518110 Chr.17: 16440286-16440286 on Build GRCh38
rs747295607 Chr.17: 16440048-16440048 on Build GRCh38
rs755357352 Chr.17: 16440188-16440188 on Build GRCh38
rs145515927 Chr.17: 16439998-16439998 on Build GRCh38
rs967601429 Chr.17: 16440063-16440063 on Build GRCh38
rs763732684 Chr.17: 16440169-16440169 on Build GRCh38
rs770862528 Chr.17: 16440231-16440231 on Build GRCh38
rs747149217 Chr.17: 16440108-16440108 on Build GRCh38
rs748729580 Chr.17: 16439986-16439986 on Build GRCh38
rs771809254 Chr.17: 16440242-16440242 on Build GRCh38
rs763444294 Chr.17: 16440095-16440096 on Build GRCh38
rs778785949 Chr.17: 16440232-16440232 on Build GRCh38
rs528987524 Chr.17: 16440201-16440201 on Build GRCh38
rs766202202 Chr.17: 16440070-16440070 on Build GRCh38
rs749169206 Chr.17: 16440229-16440229 on Build GRCh38
rs776793174 Chr.17: 16440056-16440056 on Build GRCh38
rs764888595 Chr.17: 16439987-16439987 on Build GRCh38
rs745660052 Chr.17: 16440233-16440233 on Build GRCh38
rs759838585 Chr.17: 16440120-16440122 on Build GRCh38
rs966526317 Chr.17: 16440387-16440387 on Build GRCh38
rs11945 Chr.17: 16440129-16440129 on Build GRCh38
rs1034956419 Chr.17: 16440003-16440003 on Build GRCh38
rs758676536 Chr.17: 16439995-16439995 on Build GRCh38
rs182416931 Chr.17: 16440258-16440258 on Build GRCh38
rs763485722 Chr.17: 16440257-16440257 on Build GRCh38
rs1008743036 Chr.17: 16439960-16439960 on Build GRCh38
rs755643470 Chr.17: 16440094-16440094 on Build GRCh38
rs754419924 Chr.17: 16440008-16440008 on Build GRCh38
rs757798375 Chr.17: 16440031-16440031 on Build GRCh38
rs758800373 Chr.17: 16440105-16440105 on Build GRCh38
rs762715483 Chr.17: 16440068-16440068 on Build GRCh38
rs932053572 Chr.17: 16440157-16440157 on Build GRCh38
rs766790849 Chr.17: 16440186-16440186 on Build GRCh38
rs748360829 Chr.17: 16440213-16440213 on Build GRCh38
rs368864239 Chr.17: 16440253-16440253 on Build GRCh38
rs528049318 Chr.17: 16440267-16440267 on Build GRCh38
rs774271569 Chr.17: 16440115-16440115 on Build GRCh38
rs753913025 Chr.17: 16440297-16440297 on Build GRCh38
rs747976558 Chr.17: 16440058-16440058 on Build GRCh38
rs368573782 Chr.17: 16440079-16440079 on Build GRCh38
rs960329868 Chr.17: 16440344-16440344 on Build GRCh38
rs561588302 Chr.17: 16440155-16440155 on Build GRCh38
rs979435015 Chr.17: 16440404-16440404 on Build GRCh38
rs780397586 Chr.17: 16440042-16440042 on Build GRCh38
rs762511663 Chr.17: 16440077-16440077 on Build GRCh38
rs756345928 Chr.17: 16440282-16440282 on Build GRCh38
rs1008860540 Chr.17: 16440196-16440196 on Build GRCh38
rs758995570 Chr.17: 16440175-16440175 on Build GRCh38
rs772620121 Chr.17: 16440040-16440040 on Build GRCh38
rs752258898 Chr.17: 16440091-16440091 on Build GRCh38
rs913495591 Chr.17: 16440115-16440115 on Build GRCh38
rs767592119 Chr.17: 16440176-16440176 on Build GRCh38
rs775444612 Chr.17: 16440200-16440205 on Build GRCh38
rs937651985 Chr.17: 16440227-16440227 on Build GRCh38
rs897708551 Chr.17: 16439975-16439975 on Build GRCh38
rs374777955 Chr.17: 16440216-16440216 on Build GRCh38
rs781455040 Chr.17: 16440206-16440206 on Build GRCh38
rs1032959980 Chr.17: 16440094-16440096 on Build GRCh38
rs1001938282 Chr.17: 16440339-16440339 on Build GRCh38
rs746607207 Chr.17: 16440057-16440057 on Build GRCh38
rs755239784 Chr.17: 16440109-16440109 on Build GRCh38
rs980833024 Chr.17: 16440062-16440062 on Build GRCh38
rs1048757609 Chr.17: 16439983-16439983 on Build GRCh38
rs890467121 Chr.17: 16439945-16439945 on Build GRCh38
rs752919267 Chr.17: 16440268-16440268 on Build GRCh38
rs777766598 Chr.17: 16440288-16440288 on Build GRCh38
rs140063952 Chr.17: 16439980-16439980 on Build GRCh38
rs776167559 Chr.17: 16440149-16440149 on Build GRCh38
rs779351252 Chr.17: 16440034-16440034 on Build GRCh38
rs766882841 Chr.17: 16440090-16440090 on Build GRCh38
rs113150455 Chr.17: 16440002-16440002 on Build GRCh38
rs778249027 Chr.17: 16440023-16440023 on Build GRCh38
rs1027814423 Chr.17: 16440005-16440005 on Build GRCh38
rs760431683 Chr.17: 16440250-16440250 on Build GRCh38
rs753733464 Chr.17: 16440161-16440161 on Build GRCh38
rs1017689275 Chr.17: 16440350-16440350 on Build GRCh38
rs751262899 Chr.17: 16439999-16439999 on Build GRCh38
rs746227643 Chr.17: 16440037-16440037 on Build GRCh38
rs887561687 Chr.17: 16439985-16439985 on Build GRCh38
rs759046237 Chr.17: 16440141-16440141 on Build GRCh38
rs1054895055 Chr.17: 16440243-16440243 on Build GRCh38
rs376864181 Chr.17: 16440264-16440264 on Build GRCh38

View Full Product Details