Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64501103-64501614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX5
Entrez Gene ID
1655
Gene Name
DEAD-box helicase 5
Gene Aliases
G17P1, HLR1, HUMP68, p68
UniGene
Hs.279806
Gene Chromosome Location
Chr.17: 64498254-64507038 on Build GRCh38
Transcripts
NM_001320595.1 NM_001320596.1 NM_001320597.1 NM_004396.4 NR_039969.1

Gene Symbol
MIR5047
Entrez Gene ID
100616408
Gene Name
microRNA 5047
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 64501214-64501313 on Build GRCh38
Transcripts
NM_001320595.1 NM_001320596.1 NM_001320597.1 NM_004396.4 NR_039969.1

SNP Detail

SNP ID Chromosome Location
rs59298349 Chr.17: 64501412-64501412 on Build GRCh38
rs948160115 Chr.17: 64501218-64501218 on Build GRCh38
rs781953144 Chr.17: 64501565-64501565 on Build GRCh38
rs554378158 Chr.17: 64501284-64501284 on Build GRCh38
rs62073204 Chr.17: 64501194-64501194 on Build GRCh38
rs144818179 Chr.17: 64501309-64501309 on Build GRCh38
rs181146530 Chr.17: 64501308-64501308 on Build GRCh38
rs997823456 Chr.17: 64501495-64501495 on Build GRCh38
rs1045157665 Chr.17: 64501234-64501234 on Build GRCh38
rs782251887 Chr.17: 64501342-64501342 on Build GRCh38
rs553787494 Chr.17: 64501361-64501361 on Build GRCh38
rs1031043930 Chr.17: 64501551-64501551 on Build GRCh38
rs188327005 Chr.17: 64501182-64501182 on Build GRCh38
rs782352487 Chr.17: 64501539-64501539 on Build GRCh38
rs965067231 Chr.17: 64501485-64501485 on Build GRCh38
rs915438388 Chr.17: 64501585-64501585 on Build GRCh38
rs185787791 Chr.17: 64501426-64501426 on Build GRCh38
rs897073741 Chr.17: 64501318-64501318 on Build GRCh38
rs782655765 Chr.17: 64501214-64501214 on Build GRCh38
rs369110693 Chr.17: 64501423-64501423 on Build GRCh38
rs939346358 Chr.17: 64501297-64501297 on Build GRCh38
rs549577050 Chr.17: 64501141-64501141 on Build GRCh38
rs782424473 Chr.17: 64501383-64501383 on Build GRCh38
rs782123911 Chr.17: 64501570-64501570 on Build GRCh38
rs373566078 Chr.17: 64501241-64501241 on Build GRCh38

View Full Product Details