Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77881066-77881571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45079
Entrez Gene ID
400624
Gene Name
FLJ45079 protein
Gene Aliases
-
UniGene
Hs.514521
Gene Chromosome Location
Chr.17: 77879001-77884087 on Build GRCh38
Transcripts
NR_028337.1

SNP Detail

SNP ID Chromosome Location
rs906012519 Chr.17: 77881366-77881366 on Build GRCh38
rs35585089 Chr.17: 77881381-77881382 on Build GRCh38
rs189928946 Chr.17: 77881244-77881244 on Build GRCh38
rs775668083 Chr.17: 77881137-77881137 on Build GRCh38
rs996696234 Chr.17: 77881543-77881543 on Build GRCh38
rs963885764 Chr.17: 77881235-77881235 on Build GRCh38
rs1029346495 Chr.17: 77881229-77881229 on Build GRCh38
rs544249329 Chr.17: 77881477-77881477 on Build GRCh38
rs35095574 Chr.17: 77881382-77881383 on Build GRCh38
rs533080963 Chr.17: 77881090-77881090 on Build GRCh38
rs9912960 Chr.17: 77881200-77881200 on Build GRCh38
rs750430769 Chr.17: 77881456-77881456 on Build GRCh38
rs907847221 Chr.17: 77881131-77881131 on Build GRCh38
rs1044873216 Chr.17: 77881150-77881150 on Build GRCh38
rs542006392 Chr.17: 77881355-77881355 on Build GRCh38
rs948202646 Chr.17: 77881149-77881149 on Build GRCh38
rs973690948 Chr.17: 77881369-77881369 on Build GRCh38
rs962033214 Chr.17: 77881368-77881368 on Build GRCh38
rs563936149 Chr.17: 77881514-77881514 on Build GRCh38
rs530518583 Chr.17: 77881449-77881449 on Build GRCh38
rs975926133 Chr.17: 77881225-77881225 on Build GRCh38
rs372102030 Chr.17: 77881461-77881461 on Build GRCh38
rs2077228 Chr.17: 77881423-77881423 on Build GRCh38
rs932666645 Chr.17: 77881093-77881093 on Build GRCh38
rs937603420 Chr.17: 77881375-77881375 on Build GRCh38
rs907876053 Chr.17: 77881347-77881347 on Build GRCh38
rs928127589 Chr.17: 77881506-77881506 on Build GRCh38
rs950289395 Chr.17: 77881331-77881331 on Build GRCh38
rs936081857 Chr.17: 77881116-77881116 on Build GRCh38
rs73997813 Chr.17: 77881094-77881094 on Build GRCh38
rs80150020 Chr.17: 77881522-77881522 on Build GRCh38
rs1053158079 Chr.17: 77881126-77881126 on Build GRCh38
rs1004590924 Chr.17: 77881192-77881192 on Build GRCh38
rs555128517 Chr.17: 77881156-77881156 on Build GRCh38
rs935987095 Chr.17: 77881258-77881258 on Build GRCh38
rs73997814 Chr.17: 77881201-77881201 on Build GRCh38
rs899746519 Chr.17: 77881541-77881541 on Build GRCh38
rs1022127789 Chr.17: 77881217-77881217 on Build GRCh38
rs954977144 Chr.17: 77881291-77881291 on Build GRCh38
rs920842118 Chr.17: 77881403-77881403 on Build GRCh38
rs568548283 Chr.17: 77881173-77881173 on Build GRCh38
rs953155267 Chr.17: 77881536-77881536 on Build GRCh38
rs1015116217 Chr.17: 77881465-77881465 on Build GRCh38
rs987545966 Chr.17: 77881344-77881344 on Build GRCh38
rs887483470 Chr.17: 77881408-77881408 on Build GRCh38
rs1037926132 Chr.17: 77881140-77881140 on Build GRCh38
rs573131511 Chr.17: 77881281-77881281 on Build GRCh38
rs147071289 Chr.17: 77881234-77881234 on Build GRCh38
rs987859682 Chr.17: 77881109-77881109 on Build GRCh38
rs901574205 Chr.17: 77881483-77881483 on Build GRCh38
rs953291305 Chr.17: 77881230-77881230 on Build GRCh38
rs182137075 Chr.17: 77881525-77881525 on Build GRCh38
rs1045236787 Chr.17: 77881492-77881492 on Build GRCh38
rs892127398 Chr.17: 77881521-77881521 on Build GRCh38
rs5822224 Chr.17: 77881383-77881384 on Build GRCh38
rs1029923718 Chr.17: 77881545-77881545 on Build GRCh38
rs939109818 Chr.17: 77881507-77881507 on Build GRCh38
rs1001718126 Chr.17: 77881157-77881157 on Build GRCh38
rs186311058 Chr.17: 77881154-77881154 on Build GRCh38
rs147687952 Chr.17: 77881237-77881237 on Build GRCh38
rs1027290684 Chr.17: 77881159-77881159 on Build GRCh38
rs989337470 Chr.17: 77881285-77881285 on Build GRCh38
rs894545 Chr.17: 77881147-77881147 on Build GRCh38

View Full Product Details