Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77883030-77883520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45079
Entrez Gene ID
400624
Gene Name
FLJ45079 protein
Gene Aliases
-
UniGene
Hs.514521
Gene Chromosome Location
Chr.17: 77879001-77884087 on Build GRCh38
Transcripts
NR_028337.1

SNP Detail

SNP ID Chromosome Location
rs1014586798 Chr.17: 77883502-77883502 on Build GRCh38
rs1039098128 Chr.17: 77883353-77883353 on Build GRCh38
rs759105996 Chr.17: 77883257-77883257 on Build GRCh38
rs1034293369 Chr.17: 77883210-77883210 on Build GRCh38
rs1001267139 Chr.17: 77883452-77883452 on Build GRCh38
rs62078501 Chr.17: 77883155-77883155 on Build GRCh38
rs962573553 Chr.17: 77883215-77883215 on Build GRCh38
rs894776892 Chr.17: 77883422-77883422 on Build GRCh38
rs913039670 Chr.17: 77883059-77883059 on Build GRCh38
rs764855976 Chr.17: 77883406-77883406 on Build GRCh38
rs992727850 Chr.17: 77883246-77883246 on Build GRCh38
rs911866716 Chr.17: 77883301-77883301 on Build GRCh38
rs993925746 Chr.17: 77883174-77883174 on Build GRCh38
rs1041265823 Chr.17: 77883390-77883390 on Build GRCh38
rs890023574 Chr.17: 77883124-77883124 on Build GRCh38
rs551744558 Chr.17: 77883443-77883443 on Build GRCh38
rs776632089 Chr.17: 77883169-77883169 on Build GRCh38
rs560191296 Chr.17: 77883188-77883188 on Build GRCh38
rs902744773 Chr.17: 77883401-77883401 on Build GRCh38
rs888146381 Chr.17: 77883192-77883192 on Build GRCh38
rs761870023 Chr.17: 77883379-77883379 on Build GRCh38
rs534360469 Chr.17: 77883483-77883483 on Build GRCh38
rs148401792 Chr.17: 77883240-77883240 on Build GRCh38
rs565099784 Chr.17: 77883123-77883123 on Build GRCh38
rs936705678 Chr.17: 77883291-77883291 on Build GRCh38
rs1026732712 Chr.17: 77883182-77883182 on Build GRCh38
rs1001187362 Chr.17: 77883206-77883206 on Build GRCh38
rs1011577547 Chr.17: 77883054-77883054 on Build GRCh38
rs945474438 Chr.17: 77883103-77883103 on Build GRCh38
rs556474582 Chr.17: 77883423-77883423 on Build GRCh38
rs540351626 Chr.17: 77883156-77883156 on Build GRCh38
rs373180101 Chr.17: 77883333-77883333 on Build GRCh38
rs911202280 Chr.17: 77883363-77883363 on Build GRCh38
rs375470195 Chr.17: 77883409-77883409 on Build GRCh38
rs770923057 Chr.17: 77883149-77883149 on Build GRCh38
rs943442246 Chr.17: 77883325-77883327 on Build GRCh38
rs763675975 Chr.17: 77883487-77883487 on Build GRCh38
rs548956321 Chr.17: 77883298-77883298 on Build GRCh38
rs752264544 Chr.17: 77883410-77883410 on Build GRCh38
rs925291155 Chr.17: 77883109-77883109 on Build GRCh38
rs1020542465 Chr.17: 77883256-77883256 on Build GRCh38
rs773008290 Chr.17: 77883089-77883089 on Build GRCh38
rs760556799 Chr.17: 77883148-77883148 on Build GRCh38
rs189958005 Chr.17: 77883464-77883464 on Build GRCh38
rs993050458 Chr.17: 77883058-77883058 on Build GRCh38
rs944300606 Chr.17: 77883374-77883374 on Build GRCh38
rs959928200 Chr.17: 77883056-77883056 on Build GRCh38
rs978188270 Chr.17: 77883275-77883275 on Build GRCh38
rs918798042 Chr.17: 77883394-77883394 on Build GRCh38
rs544927958 Chr.17: 77883113-77883113 on Build GRCh38
rs978111192 Chr.17: 77883105-77883105 on Build GRCh38
rs925364650 Chr.17: 77883279-77883279 on Build GRCh38
rs985847905 Chr.17: 77883322-77883322 on Build GRCh38

View Full Product Details