Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 77883289-77883724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45079
Entrez Gene ID
400624
Gene Name
FLJ45079 protein
Gene Aliases
-
UniGene
Hs.514521
Gene Chromosome Location
Chr.17: 77879001-77884087 on Build GRCh38
Transcripts
NR_028337.1

SNP Detail

SNP ID Chromosome Location
rs985534601 Chr.17: 77883678-77883678 on Build GRCh38
rs1014586798 Chr.17: 77883502-77883502 on Build GRCh38
rs35815167 Chr.17: 77883612-77883613 on Build GRCh38
rs1039098128 Chr.17: 77883353-77883353 on Build GRCh38
rs999974335 Chr.17: 77883584-77883584 on Build GRCh38
rs556474582 Chr.17: 77883423-77883423 on Build GRCh38
rs924072687 Chr.17: 77883687-77883687 on Build GRCh38
rs1020200915 Chr.17: 77883507-77883507 on Build GRCh38
rs1001267139 Chr.17: 77883452-77883452 on Build GRCh38
rs373180101 Chr.17: 77883333-77883333 on Build GRCh38
rs536300899 Chr.17: 77883622-77883622 on Build GRCh38
rs911202280 Chr.17: 77883363-77883363 on Build GRCh38
rs375470195 Chr.17: 77883409-77883409 on Build GRCh38
rs188568564 Chr.17: 77883553-77883553 on Build GRCh38
rs943442246 Chr.17: 77883325-77883327 on Build GRCh38
rs894776892 Chr.17: 77883422-77883422 on Build GRCh38
rs763675975 Chr.17: 77883487-77883487 on Build GRCh38
rs79728577 Chr.17: 77883679-77883679 on Build GRCh38
rs764855976 Chr.17: 77883406-77883406 on Build GRCh38
rs752264544 Chr.17: 77883410-77883410 on Build GRCh38
rs189958005 Chr.17: 77883464-77883464 on Build GRCh38
rs944300606 Chr.17: 77883374-77883374 on Build GRCh38
rs1041265823 Chr.17: 77883390-77883390 on Build GRCh38
rs915314187 Chr.17: 77883664-77883664 on Build GRCh38
rs965368430 Chr.17: 77883694-77883694 on Build GRCh38
rs551744558 Chr.17: 77883443-77883443 on Build GRCh38
rs918798042 Chr.17: 77883394-77883394 on Build GRCh38
rs750743852 Chr.17: 77883524-77883524 on Build GRCh38
rs902744773 Chr.17: 77883401-77883401 on Build GRCh38
rs985847905 Chr.17: 77883322-77883322 on Build GRCh38
rs553944477 Chr.17: 77883548-77883548 on Build GRCh38
rs761870023 Chr.17: 77883379-77883379 on Build GRCh38
rs534360469 Chr.17: 77883483-77883483 on Build GRCh38
rs567361116 Chr.17: 77883601-77883601 on Build GRCh38
rs964703641 Chr.17: 77883645-77883645 on Build GRCh38
rs955329746 Chr.17: 77883522-77883522 on Build GRCh38
rs967220079 Chr.17: 77883508-77883508 on Build GRCh38
rs986680940 Chr.17: 77883564-77883565 on Build GRCh38
rs556200710 Chr.17: 77883646-77883646 on Build GRCh38

View Full Product Details