Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13419689-13420228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

Gene Symbol
LDLRAD4-AS1
Entrez Gene ID
100288122
Gene Name
LDLRAD4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.646264
Gene Chromosome Location
Chr.18: 13419499-13427534 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

SNP Detail

SNP ID Chromosome Location
rs990205824 Chr.18: 13420057-13420057 on Build GRCh38
rs570995761 Chr.18: 13420117-13420117 on Build GRCh38
rs1013956382 Chr.18: 13419955-13419955 on Build GRCh38
rs898311085 Chr.18: 13419923-13419923 on Build GRCh38
rs974892292 Chr.18: 13420001-13420001 on Build GRCh38
rs1038087393 Chr.18: 13419905-13419905 on Build GRCh38
rs561469818 Chr.18: 13419997-13419997 on Build GRCh38
rs118087637 Chr.18: 13420076-13420076 on Build GRCh38
rs1038056420 Chr.18: 13420112-13420112 on Build GRCh38
rs1031803449 Chr.18: 13420042-13420042 on Build GRCh38
rs556863912 Chr.18: 13419904-13419904 on Build GRCh38
rs914596947 Chr.18: 13420062-13420062 on Build GRCh38
rs937801361 Chr.18: 13420139-13420139 on Build GRCh38
rs943914930 Chr.18: 13419894-13419894 on Build GRCh38
rs149634832 Chr.18: 13419877-13419877 on Build GRCh38
rs984118621 Chr.18: 13420052-13420052 on Build GRCh38
rs577389456 Chr.18: 13420092-13420092 on Build GRCh38
rs539809185 Chr.18: 13419983-13419983 on Build GRCh38
rs767828527 Chr.18: 13419841-13419841 on Build GRCh38
rs982002137 Chr.18: 13419732-13419732 on Build GRCh38
rs974959665 Chr.18: 13419814-13419814 on Build GRCh38
rs182949661 Chr.18: 13419742-13419742 on Build GRCh38
rs143367025 Chr.18: 13420149-13420149 on Build GRCh38
rs924750851 Chr.18: 13419737-13419737 on Build GRCh38
rs1020922367 Chr.18: 13419974-13419974 on Build GRCh38
rs914544510 Chr.18: 13419824-13419824 on Build GRCh38
rs1037264323 Chr.18: 13419935-13419935 on Build GRCh38
rs912452635 Chr.18: 13419884-13419884 on Build GRCh38
rs566866041 Chr.18: 13419897-13419897 on Build GRCh38
rs923475415 Chr.18: 13420005-13420005 on Build GRCh38
rs557602904 Chr.18: 13419767-13419767 on Build GRCh38
rs919659262 Chr.18: 13420126-13420126 on Build GRCh38
rs943951526 Chr.18: 13420177-13420177 on Build GRCh38
rs939738763 Chr.18: 13419903-13419903 on Build GRCh38
rs77547273 Chr.18: 13420159-13420159 on Build GRCh38
rs545778340 Chr.18: 13419944-13419944 on Build GRCh38
rs572515165 Chr.18: 13419982-13419982 on Build GRCh38
rs537035203 Chr.18: 13419942-13419942 on Build GRCh38
rs896316850 Chr.18: 13420150-13420150 on Build GRCh38
rs187646981 Chr.18: 13419878-13419878 on Build GRCh38
rs11878138 Chr.18: 13420050-13420050 on Build GRCh38
rs531101956 Chr.18: 13419728-13419728 on Build GRCh38
rs150394322 Chr.18: 13419762-13419762 on Build GRCh38
rs573398440 Chr.18: 13420031-13420031 on Build GRCh38
rs1003081369 Chr.18: 13420028-13420028 on Build GRCh38
rs138346417 Chr.18: 13419808-13419808 on Build GRCh38
rs1042713421 Chr.18: 13420182-13420182 on Build GRCh38
rs191014131 Chr.18: 13420161-13420161 on Build GRCh38
rs1001979758 Chr.18: 13419928-13419928 on Build GRCh38
rs1032442995 Chr.18: 13419986-13419986 on Build GRCh38
rs534973024 Chr.18: 13419807-13419807 on Build GRCh38
rs7235364 Chr.18: 13419949-13419949 on Build GRCh38
rs780453143 Chr.18: 13419879-13419879 on Build GRCh38

View Full Product Details