Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13425821-13426321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD4
Entrez Gene ID
753
Gene Name
low density lipoprotein receptor class A domain containing 4
Gene Aliases
C18orf1
UniGene
Hs.731853
Gene Chromosome Location
Chr.18: 13217691-13652754 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

Gene Symbol
LDLRAD4-AS1
Entrez Gene ID
100288122
Gene Name
LDLRAD4 antisense RNA 1
Gene Aliases
-
UniGene
Hs.646264
Gene Chromosome Location
Chr.18: 13419499-13427534 on Build GRCh38
Transcripts
NM_181481.4 NM_181482.4 XM_005258140.1 XM_006722353.1 XM_006722354.1 XM_011525737.1 XM_011525738.1 XM_017025961.1 XM_017025962.1 XM_017025963.1 XM_017025964.1 XM_017025965.1 XM_017025966.1 XM_017025967.1 XM_017025968.1 NR_040031.1

SNP Detail

SNP ID Chromosome Location
rs1021594238 Chr.18: 13425864-13425864 on Build GRCh38
rs555196763 Chr.18: 13426255-13426255 on Build GRCh38
rs970593670 Chr.18: 13425852-13425852 on Build GRCh38
rs557585854 Chr.18: 13426172-13426172 on Build GRCh38
rs947466634 Chr.18: 13426076-13426076 on Build GRCh38
rs938871235 Chr.18: 13426023-13426023 on Build GRCh38
rs916224762 Chr.18: 13425998-13425998 on Build GRCh38
rs779295117 Chr.18: 13425843-13425843 on Build GRCh38
rs940916665 Chr.18: 13426008-13426008 on Build GRCh38
rs2114427 Chr.18: 13426246-13426246 on Build GRCh38
rs539348001 Chr.18: 13426024-13426024 on Build GRCh38
rs891883865 Chr.18: 13426065-13426065 on Build GRCh38
rs978022309 Chr.18: 13425895-13425895 on Build GRCh38
rs1006740561 Chr.18: 13425891-13425891 on Build GRCh38
rs972587370 Chr.18: 13425979-13425979 on Build GRCh38
rs573661853 Chr.18: 13426286-13426286 on Build GRCh38
rs201713669 Chr.18: 13426107-13426107 on Build GRCh38
rs777987603 Chr.18: 13426032-13426032 on Build GRCh38
rs938913607 Chr.18: 13426264-13426264 on Build GRCh38
rs868182079 Chr.18: 13426082-13426082 on Build GRCh38
rs142579039 Chr.18: 13425904-13425905 on Build GRCh38
rs1036601511 Chr.18: 13426021-13426021 on Build GRCh38
rs1019526738 Chr.18: 13426168-13426168 on Build GRCh38
rs970562792 Chr.18: 13426147-13426147 on Build GRCh38
rs921936860 Chr.18: 13426209-13426209 on Build GRCh38
rs969077193 Chr.18: 13426206-13426206 on Build GRCh38
rs1024790901 Chr.18: 13426140-13426140 on Build GRCh38
rs899433629 Chr.18: 13426031-13426031 on Build GRCh38
rs932029745 Chr.18: 13426252-13426252 on Build GRCh38
rs768208125 Chr.18: 13426200-13426200 on Build GRCh38
rs368830984 Chr.18: 13425986-13425986 on Build GRCh38
rs747046407 Chr.18: 13426089-13426089 on Build GRCh38
rs72874889 Chr.18: 13426105-13426105 on Build GRCh38
rs994413148 Chr.18: 13426183-13426183 on Build GRCh38
rs922039555 Chr.18: 13425958-13425958 on Build GRCh38
rs748335929 Chr.18: 13425943-13425943 on Build GRCh38
rs978358819 Chr.18: 13426186-13426186 on Build GRCh38
rs540608997 Chr.18: 13426017-13426017 on Build GRCh38
rs947682506 Chr.18: 13426005-13426005 on Build GRCh38
rs1043471462 Chr.18: 13426104-13426104 on Build GRCh38
rs989399568 Chr.18: 13426282-13426282 on Build GRCh38
rs987527767 Chr.18: 13426261-13426261 on Build GRCh38
rs1027100521 Chr.18: 13426041-13426041 on Build GRCh38
rs894862539 Chr.18: 13426084-13426084 on Build GRCh38
rs76637460 Chr.18: 13426203-13426203 on Build GRCh38
rs139955315 Chr.18: 13425968-13425968 on Build GRCh38
rs901056367 Chr.18: 13426179-13426179 on Build GRCh38
rs369384383 Chr.18: 13425921-13425922 on Build GRCh38
rs936673680 Chr.18: 13425953-13425953 on Build GRCh38
rs59792741 Chr.18: 13426287-13426287 on Build GRCh38
rs923910415 Chr.18: 13425928-13425928 on Build GRCh38
rs953978289 Chr.18: 13425889-13425889 on Build GRCh38
rs780872742 Chr.18: 13426273-13426274 on Build GRCh38
rs1006835374 Chr.18: 13426165-13426165 on Build GRCh38
rs78167209 Chr.18: 13426193-13426193 on Build GRCh38
rs772239964 Chr.18: 13425984-13425984 on Build GRCh38
rs984203872 Chr.18: 13425989-13425989 on Build GRCh38
rs34178898 Chr.18: 13426209-13426210 on Build GRCh38
rs371495680 Chr.18: 13425957-13425957 on Build GRCh38

View Full Product Details