Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26930562-26931073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_017026033.1 XM_017026034.1

SNP Detail

SNP ID Chromosome Location
rs1019512827 Chr.18: 26930670-26930670 on Build GRCh38
rs1029088609 Chr.18: 26931027-26931027 on Build GRCh38
rs893216068 Chr.18: 26931028-26931028 on Build GRCh38
rs772497788 Chr.18: 26930652-26930652 on Build GRCh38
rs966681157 Chr.18: 26931045-26931045 on Build GRCh38
rs879708299 Chr.18: 26930934-26930934 on Build GRCh38
rs927774033 Chr.18: 26930768-26930768 on Build GRCh38
rs543823382 Chr.18: 26930949-26930953 on Build GRCh38
rs577109325 Chr.18: 26930798-26930798 on Build GRCh38
rs1029060646 Chr.18: 26930707-26930707 on Build GRCh38
rs748588694 Chr.18: 26930596-26930596 on Build GRCh38
rs173075 Chr.18: 26930880-26930880 on Build GRCh38
rs781382758 Chr.18: 26930876-26930876 on Build GRCh38
rs180147 Chr.18: 26930869-26930869 on Build GRCh38
rs1020683419 Chr.18: 26931034-26931034 on Build GRCh38
rs548288821 Chr.18: 26930665-26930665 on Build GRCh38
rs752568186 Chr.18: 26930929-26930929 on Build GRCh38
rs760900747 Chr.18: 26930684-26930684 on Build GRCh38
rs574898199 Chr.18: 26930859-26930859 on Build GRCh38
rs955928848 Chr.18: 26930944-26930944 on Build GRCh38
rs232346 Chr.18: 26930958-26930958 on Build GRCh38
rs71353413 Chr.18: 26930681-26930681 on Build GRCh38
rs886717378 Chr.18: 26930868-26930868 on Build GRCh38
rs556479384 Chr.18: 26930841-26930841 on Build GRCh38
rs939169072 Chr.18: 26930783-26930783 on Build GRCh38
rs996278068 Chr.18: 26930692-26930692 on Build GRCh38
rs930346990 Chr.18: 26930840-26930840 on Build GRCh38
rs1048711039 Chr.18: 26930857-26930857 on Build GRCh38
rs369399029 Chr.18: 26930756-26930756 on Build GRCh38
rs163224 Chr.18: 26930789-26930789 on Build GRCh38
rs1011620771 Chr.18: 26931033-26931033 on Build GRCh38
rs373346665 Chr.18: 26930874-26930874 on Build GRCh38
rs996307837 Chr.18: 26930973-26930973 on Build GRCh38
rs149258 Chr.18: 26930729-26930729 on Build GRCh38
rs1049309675 Chr.18: 26930604-26930604 on Build GRCh38
rs774982167 Chr.18: 26930589-26930589 on Build GRCh38
rs886683867 Chr.18: 26930650-26930650 on Build GRCh38
rs143598762 Chr.18: 26930929-26930933 on Build GRCh38
rs572489314 Chr.18: 26931050-26931050 on Build GRCh38
rs990114567 Chr.18: 26930753-26930753 on Build GRCh38
rs163225 Chr.18: 26930799-26930799 on Build GRCh38
rs191418 Chr.18: 26930993-26930993 on Build GRCh38
rs112041808 Chr.18: 26930755-26930755 on Build GRCh38
rs957362091 Chr.18: 26930733-26930733 on Build GRCh38

View Full Product Details