Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 49049280-49049782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYM
Entrez Gene ID
54808
Gene Name
dymeclin
Gene Aliases
DMC, SMC
UniGene
Hs.162996
Gene Chromosome Location
Chr.18: 49041474-49460709 on Build GRCh38
Transcripts
NM_017653.3 XM_006722488.2 XM_011526036.1 XM_011526037.1 XM_011526038.1 XM_011526040.1 XM_011526041.1 XM_017025795.1 XM_017025796.1 XM_017025797.1 XM_017025798.1 XM_017025799.1 XM_017025800.1 XM_017025801.1 NR_039898.1

Gene Symbol
MIR4744
Entrez Gene ID
100616420
Gene Name
microRNA 4744
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.18: 49049687-49049768 on Build GRCh38
Transcripts
NM_017653.3 XM_006722488.2 XM_011526036.1 XM_011526037.1 XM_011526038.1 XM_011526040.1 XM_011526041.1 XM_017025795.1 XM_017025796.1 XM_017025797.1 XM_017025798.1 XM_017025799.1 XM_017025800.1 XM_017025801.1 NR_039898.1

SNP Detail

SNP ID Chromosome Location
rs954795047 Chr.18: 49049451-49049451 on Build GRCh38
rs990220772 Chr.18: 49049516-49049516 on Build GRCh38
rs894192321 Chr.18: 49049330-49049330 on Build GRCh38
rs185388564 Chr.18: 49049672-49049672 on Build GRCh38
rs12456845 Chr.18: 49049688-49049688 on Build GRCh38
rs143628225 Chr.18: 49049383-49049384 on Build GRCh38
rs942756647 Chr.18: 49049484-49049484 on Build GRCh38
rs545949365 Chr.18: 49049563-49049563 on Build GRCh38
rs1005993748 Chr.18: 49049334-49049334 on Build GRCh38
rs973574550 Chr.18: 49049326-49049326 on Build GRCh38
rs767551266 Chr.18: 49049309-49049309 on Build GRCh38
rs768247446 Chr.18: 49049617-49049617 on Build GRCh38
rs375615754 Chr.18: 49049318-49049318 on Build GRCh38
rs566127727 Chr.18: 49049576-49049576 on Build GRCh38
rs1005961103 Chr.18: 49049723-49049723 on Build GRCh38
rs931835347 Chr.18: 49049614-49049614 on Build GRCh38
rs946273223 Chr.18: 49049553-49049553 on Build GRCh38
rs913234361 Chr.18: 49049581-49049581 on Build GRCh38
rs969102762 Chr.18: 49049692-49049692 on Build GRCh38
rs1016077179 Chr.18: 49049349-49049349 on Build GRCh38
rs947383043 Chr.18: 49049582-49049582 on Build GRCh38
rs558808963 Chr.18: 49049546-49049546 on Build GRCh38
rs139567920 Chr.18: 49049724-49049724 on Build GRCh38
rs994494492 Chr.18: 49049687-49049687 on Build GRCh38
rs755330746 Chr.18: 49049742-49049742 on Build GRCh38
rs577086598 Chr.18: 49049433-49049433 on Build GRCh38
rs1051593757 Chr.18: 49049623-49049623 on Build GRCh38
rs868513843 Chr.18: 49049559-49049559 on Build GRCh38
rs746705743 Chr.18: 49049500-49049500 on Build GRCh38
rs970223277 Chr.18: 49049525-49049525 on Build GRCh38
rs750496708 Chr.18: 49049400-49049400 on Build GRCh38
rs898844855 Chr.18: 49049678-49049678 on Build GRCh38
rs900571428 Chr.18: 49049610-49049610 on Build GRCh38
rs960166662 Chr.18: 49049746-49049746 on Build GRCh38
rs754296345 Chr.18: 49049703-49049703 on Build GRCh38
rs534009369 Chr.18: 49049671-49049671 on Build GRCh38
rs556382233 Chr.18: 49049745-49049745 on Build GRCh38
rs938934400 Chr.18: 49049575-49049575 on Build GRCh38
rs890252233 Chr.18: 49049662-49049662 on Build GRCh38
rs538979278 Chr.18: 49049396-49049396 on Build GRCh38
rs980227517 Chr.18: 49049532-49049532 on Build GRCh38
rs1043554119 Chr.18: 49049593-49049593 on Build GRCh38
rs1030281321 Chr.18: 49049403-49049403 on Build GRCh38
rs911240403 Chr.18: 49049465-49049465 on Build GRCh38
rs756652619 Chr.18: 49049560-49049560 on Build GRCh38

View Full Product Details