Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1952686-1953190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G2
Entrez Gene ID
1455
Gene Name
casein kinase 1 gamma 2
Gene Aliases
CK1g2
UniGene
Hs.651905
Gene Chromosome Location
Chr.19: 1941149-1981338 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259500.1 XM_017026297.1 NR_033400.1

Gene Symbol
CSNK1G2-AS1
Entrez Gene ID
255193
Gene Name
CSNK1G2 antisense RNA 1
Gene Aliases
C19orf34
UniGene
Hs.374717
Gene Chromosome Location
Chr.19: 1952527-1954549 on Build GRCh38
Transcripts
NM_001319.6 XM_005259498.1 XM_005259500.1 XM_017026297.1 NR_033400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892367 c.317G>A p.R106Q Chr.19: 1953009-1953009 on Build GRCh38
6636806 c.378G>A p.R126R Chr.19: 1952948-1952948 on Build GRCh38
3101284 c.409G>A p.E137K Chr.19: 1952917-1952917 on Build GRCh38
3101287 c.389G>A p.R130H Chr.19: 1952937-1952937 on Build GRCh38
993326 c.362C>T p.S121L Chr.19: 1952964-1952964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16991001 Chr.19: 1952894-1952894 on Build GRCh38
rs1042762666 Chr.19: 1952750-1952750 on Build GRCh38
rs1053110424 Chr.19: 1952906-1952906 on Build GRCh38
rs116294380 Chr.19: 1953165-1953165 on Build GRCh38
rs963433132 Chr.19: 1952828-1952828 on Build GRCh38
rs752927782 Chr.19: 1952953-1952953 on Build GRCh38
rs376946152 Chr.19: 1952917-1952917 on Build GRCh38
rs533227816 Chr.19: 1952741-1952741 on Build GRCh38
rs551695486 Chr.19: 1952935-1952935 on Build GRCh38
rs764347931 Chr.19: 1953029-1953029 on Build GRCh38
rs779082287 Chr.19: 1953043-1953043 on Build GRCh38
rs943206953 Chr.19: 1953056-1953056 on Build GRCh38
rs763529796 Chr.19: 1952746-1952746 on Build GRCh38
rs748557040 Chr.19: 1952837-1952837 on Build GRCh38
rs117450701 Chr.19: 1952813-1952813 on Build GRCh38
rs368351443 Chr.19: 1952869-1952869 on Build GRCh38
rs775014021 Chr.19: 1952849-1952849 on Build GRCh38
rs768288432 Chr.19: 1952967-1952967 on Build GRCh38
rs756154000 Chr.19: 1953012-1953012 on Build GRCh38
rs1001512135 Chr.19: 1952772-1952772 on Build GRCh38
rs929108443 Chr.19: 1952790-1952790 on Build GRCh38
rs561767183 Chr.19: 1952715-1952715 on Build GRCh38
rs750374883 Chr.19: 1952833-1952833 on Build GRCh38
rs138242650 Chr.19: 1952717-1952717 on Build GRCh38
rs749701489 Chr.19: 1952961-1952961 on Build GRCh38
rs1033744632 Chr.19: 1952754-1952754 on Build GRCh38
rs1026609332 Chr.19: 1953008-1953008 on Build GRCh38
rs899249886 Chr.19: 1953155-1953155 on Build GRCh38
rs146078839 Chr.19: 1953001-1953001 on Build GRCh38
rs2396366 Chr.19: 1952842-1952842 on Build GRCh38
rs757223642 Chr.19: 1952868-1952868 on Build GRCh38
rs541104343 Chr.19: 1953138-1953138 on Build GRCh38
rs539784942 Chr.19: 1953010-1953010 on Build GRCh38
rs771427768 Chr.19: 1952899-1952899 on Build GRCh38
rs774689349 Chr.19: 1952902-1952902 on Build GRCh38
rs571797038 Chr.19: 1953042-1953042 on Build GRCh38
rs761493059 Chr.19: 1952932-1952932 on Build GRCh38
rs986821346 Chr.19: 1953104-1953104 on Build GRCh38
rs904076870 Chr.19: 1952766-1952766 on Build GRCh38
rs750878956 Chr.19: 1952757-1952757 on Build GRCh38
rs375885034 Chr.19: 1952745-1952745 on Build GRCh38
rs912855791 Chr.19: 1952978-1952978 on Build GRCh38
rs528956865 Chr.19: 1952874-1952874 on Build GRCh38
rs367874771 Chr.19: 1952933-1952933 on Build GRCh38
rs779057287 Chr.19: 1952867-1952867 on Build GRCh38
rs773737133 Chr.19: 1952925-1952937 on Build GRCh38
rs559372893 Chr.19: 1952918-1952918 on Build GRCh38
rs957092258 Chr.19: 1952800-1952800 on Build GRCh38
rs553716688 Chr.19: 1952808-1952808 on Build GRCh38
rs770126668 Chr.19: 1952838-1952838 on Build GRCh38
rs538890642 Chr.19: 1953124-1953124 on Build GRCh38
rs773773464 Chr.19: 1952840-1952840 on Build GRCh38
rs374851514 Chr.19: 1952937-1952937 on Build GRCh38
rs138686657 Chr.19: 1953116-1953116 on Build GRCh38
rs992456398 Chr.19: 1952778-1952778 on Build GRCh38
rs954150496 Chr.19: 1953097-1953097 on Build GRCh38
rs746358052 Chr.19: 1952964-1952964 on Build GRCh38
rs758476608 Chr.19: 1952834-1952834 on Build GRCh38
rs759408893 Chr.19: 1953002-1953002 on Build GRCh38
rs528892841 Chr.19: 1952716-1952716 on Build GRCh38
rs891277891 Chr.19: 1952726-1952726 on Build GRCh38
rs1052792189 Chr.19: 1953076-1953076 on Build GRCh38
rs55814949 Chr.19: 1953167-1953167 on Build GRCh38
rs778139799 Chr.19: 1952960-1952960 on Build GRCh38
rs760275133 Chr.19: 1952850-1952850 on Build GRCh38
rs762766875 Chr.19: 1952723-1952723 on Build GRCh38
rs558179673 Chr.19: 1953064-1953064 on Build GRCh38
rs183212922 Chr.19: 1952945-1952945 on Build GRCh38
rs767604856 Chr.19: 1953009-1953009 on Build GRCh38
rs747512732 Chr.19: 1953069-1953069 on Build GRCh38
rs949623277 Chr.19: 1953081-1953081 on Build GRCh38
rs547416087 Chr.19: 1952976-1952976 on Build GRCh38
rs112167855 Chr.19: 1952936-1952936 on Build GRCh38
rs1046652182 Chr.19: 1953096-1953096 on Build GRCh38
rs768877974 Chr.19: 1952836-1952836 on Build GRCh38
rs756278429 Chr.19: 1952954-1952954 on Build GRCh38
rs1004729738 Chr.19: 1953053-1953053 on Build GRCh38
rs535541050 Chr.19: 1952963-1952963 on Build GRCh38
rs373554362 Chr.19: 1952835-1952835 on Build GRCh38
rs748362563 Chr.19: 1952876-1952876 on Build GRCh38
rs954722321 Chr.19: 1953168-1953168 on Build GRCh38
rs767032802 Chr.19: 1953014-1953015 on Build GRCh38
rs894888230 Chr.19: 1953024-1953025 on Build GRCh38
rs561634721 Chr.19: 1952873-1952873 on Build GRCh38
rs945622553 Chr.19: 1953123-1953123 on Build GRCh38
rs767515046 Chr.19: 1952952-1952952 on Build GRCh38
rs770145263 Chr.19: 1952888-1952888 on Build GRCh38
rs559435936 Chr.19: 1953156-1953156 on Build GRCh38
rs766079950 Chr.19: 1952985-1952985 on Build GRCh38
rs772713641 Chr.19: 1952983-1952983 on Build GRCh38
rs557976661 Chr.19: 1952822-1952822 on Build GRCh38
rs556372664 Chr.19: 1952770-1952770 on Build GRCh38
rs753992850 Chr.19: 1953033-1953033 on Build GRCh38
rs372199329 Chr.19: 1952843-1952843 on Build GRCh38
rs566004349 Chr.19: 1952732-1952732 on Build GRCh38
rs537271254 Chr.19: 1953080-1953080 on Build GRCh38
rs753520464 Chr.19: 1952858-1952858 on Build GRCh38
rs747827817 Chr.19: 1952980-1952980 on Build GRCh38
rs113771051 Chr.19: 1952728-1952728 on Build GRCh38
rs568500608 Chr.19: 1952789-1952789 on Build GRCh38
rs780024054 Chr.19: 1952871-1952871 on Build GRCh38
rs774347785 Chr.19: 1952999-1952999 on Build GRCh38
rs1051600331 Chr.19: 1952864-1952864 on Build GRCh38
rs1013709031 Chr.19: 1953025-1953025 on Build GRCh38
rs757708264 Chr.19: 1953035-1953035 on Build GRCh38
rs932607947 Chr.19: 1952962-1952962 on Build GRCh38
rs772526406 Chr.19: 1952883-1952884 on Build GRCh38
rs746579486 Chr.19: 1952903-1952903 on Build GRCh38
rs45437201 Chr.19: 1952851-1952851 on Build GRCh38
rs549113127 Chr.19: 1952938-1952938 on Build GRCh38
rs74415283 Chr.19: 1952809-1952809 on Build GRCh38
rs77965237 Chr.19: 1953015-1953015 on Build GRCh38
rs769534920 Chr.19: 1952981-1952981 on Build GRCh38
rs1055295818 Chr.19: 1953164-1953164 on Build GRCh38
rs116465395 Chr.19: 1952806-1952806 on Build GRCh38
rs934288094 Chr.19: 1953075-1953075 on Build GRCh38
rs762859796 Chr.19: 1952984-1952984 on Build GRCh38
rs761239851 Chr.19: 1952973-1952973 on Build GRCh38
rs867223252 Chr.19: 1952854-1952854 on Build GRCh38
rs551634462 Chr.19: 1952742-1952742 on Build GRCh38
rs1055243701 Chr.19: 1953021-1953021 on Build GRCh38
rs909190209 Chr.19: 1952823-1952823 on Build GRCh38
rs186967736 Chr.19: 1953068-1953068 on Build GRCh38
rs774857131 Chr.19: 1953015-1953015 on Build GRCh38
rs909093800 Chr.19: 1952709-1952709 on Build GRCh38
rs142852376 Chr.19: 1952760-1952760 on Build GRCh38
rs1001008917 Chr.19: 1952869-1952871 on Build GRCh38
rs761759186 Chr.19: 1952866-1952866 on Build GRCh38

View Full Product Details