Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46660227-46660723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3
Entrez Gene ID
147906
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
DAPPER3, RRR1
UniGene
Hs.515490
Gene Chromosome Location
Chr.19: 46647564-46661138 on Build GRCh38
Transcripts
NM_145056.2 NR_040042.1

Gene Symbol
DACT3-AS1
Entrez Gene ID
100506068
Gene Name
DACT3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434327
Gene Chromosome Location
Chr.19: 46660364-46677456 on Build GRCh38
Transcripts
NM_145056.2 NR_040042.1

SNP Detail

SNP ID Chromosome Location
rs907587810 Chr.19: 46660483-46660483 on Build GRCh38
rs1032613450 Chr.19: 46660653-46660653 on Build GRCh38
rs1042456215 Chr.19: 46660627-46660627 on Build GRCh38
rs559624214 Chr.19: 46660437-46660437 on Build GRCh38
rs964919774 Chr.19: 46660364-46660364 on Build GRCh38
rs977643902 Chr.19: 46660466-46660466 on Build GRCh38
rs182295109 Chr.19: 46660311-46660311 on Build GRCh38
rs961815088 Chr.19: 46660577-46660577 on Build GRCh38
rs902519424 Chr.19: 46660641-46660641 on Build GRCh38
rs375337093 Chr.19: 46660534-46660534 on Build GRCh38
rs999699725 Chr.19: 46660394-46660394 on Build GRCh38
rs372261415 Chr.19: 46660575-46660575 on Build GRCh38
rs575731014 Chr.19: 46660645-46660645 on Build GRCh38
rs920461209 Chr.19: 46660620-46660620 on Build GRCh38
rs901312029 Chr.19: 46660370-46660370 on Build GRCh38
rs1032971701 Chr.19: 46660362-46660362 on Build GRCh38
rs1031216554 Chr.19: 46660500-46660500 on Build GRCh38
rs534524854 Chr.19: 46660471-46660471 on Build GRCh38
rs896593237 Chr.19: 46660266-46660266 on Build GRCh38
rs1020998791 Chr.19: 46660303-46660303 on Build GRCh38
rs895642776 Chr.19: 46660569-46660569 on Build GRCh38
rs185234044 Chr.19: 46660519-46660519 on Build GRCh38
rs763042698 Chr.19: 46660603-46660603 on Build GRCh38
rs1013714444 Chr.19: 46660609-46660609 on Build GRCh38
rs1057015378 Chr.19: 46660565-46660565 on Build GRCh38
rs911187207 Chr.19: 46660664-46660664 on Build GRCh38
rs571882861 Chr.19: 46660423-46660423 on Build GRCh38
rs1037634136 Chr.19: 46660511-46660511 on Build GRCh38
rs923422837 Chr.19: 46660404-46660404 on Build GRCh38
rs371956704 Chr.19: 46660334-46660334 on Build GRCh38
rs873286 Chr.19: 46660517-46660517 on Build GRCh38
rs555883360 Chr.19: 46660540-46660540 on Build GRCh38
rs975292762 Chr.19: 46660384-46660384 on Build GRCh38
rs941653946 Chr.19: 46660510-46660510 on Build GRCh38
rs867101240 Chr.19: 46660358-46660358 on Build GRCh38
rs201524895 Chr.19: 46660575-46660575 on Build GRCh38
rs1013661856 Chr.19: 46660294-46660294 on Build GRCh38
rs1023706430 Chr.19: 46660538-46660538 on Build GRCh38
rs942727166 Chr.19: 46660679-46660679 on Build GRCh38
rs1048496236 Chr.19: 46660659-46660659 on Build GRCh38
rs1033779849 Chr.19: 46660260-46660260 on Build GRCh38
rs1001126456 Chr.19: 46660359-46660359 on Build GRCh38
rs971868019 Chr.19: 46660613-46660613 on Build GRCh38

View Full Product Details