Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24971989-24972491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 NR_034113.1

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 NR_034113.1

SNP Detail

SNP ID Chromosome Location
rs1055011504 Chr.2: 24972415-24972415 on Build GRCh38
rs535460137 Chr.2: 24972114-24972114 on Build GRCh38
rs553932691 Chr.2: 24972110-24972110 on Build GRCh38
rs931213438 Chr.2: 24972413-24972413 on Build GRCh38
rs895001696 Chr.2: 24972439-24972439 on Build GRCh38
rs1022853428 Chr.2: 24972192-24972192 on Build GRCh38
rs974973726 Chr.2: 24972301-24972301 on Build GRCh38
rs949784430 Chr.2: 24972022-24972022 on Build GRCh38
rs1013629143 Chr.2: 24972055-24972055 on Build GRCh38
rs539639738 Chr.2: 24972184-24972184 on Build GRCh38
rs981131394 Chr.2: 24972033-24972033 on Build GRCh38
rs562068279 Chr.2: 24972239-24972239 on Build GRCh38
rs1046547350 Chr.2: 24972462-24972462 on Build GRCh38
rs999381146 Chr.2: 24972287-24972287 on Build GRCh38
rs986531237 Chr.2: 24972356-24972356 on Build GRCh38
rs919891564 Chr.2: 24972411-24972411 on Build GRCh38
rs986479047 Chr.2: 24972099-24972099 on Build GRCh38
rs899858347 Chr.2: 24972125-24972125 on Build GRCh38
rs988037010 Chr.2: 24972376-24972376 on Build GRCh38
rs966663137 Chr.2: 24972061-24972061 on Build GRCh38
rs898345302 Chr.2: 24972035-24972035 on Build GRCh38
rs751882952 Chr.2: 24972242-24972242 on Build GRCh38
rs982961380 Chr.2: 24972226-24972226 on Build GRCh38
rs973615994 Chr.2: 24972352-24972352 on Build GRCh38
rs564998660 Chr.2: 24972298-24972298 on Build GRCh38
rs918967405 Chr.2: 24972211-24972211 on Build GRCh38
rs957304122 Chr.2: 24972186-24972186 on Build GRCh38
rs182287849 Chr.2: 24972116-24972116 on Build GRCh38
rs903277217 Chr.2: 24972179-24972179 on Build GRCh38
rs1039955909 Chr.2: 24972247-24972247 on Build GRCh38
rs568032861 Chr.2: 24972082-24972082 on Build GRCh38
rs1047677673 Chr.2: 24972066-24972066 on Build GRCh38
rs957520768 Chr.2: 24972014-24972014 on Build GRCh38
rs892934937 Chr.2: 24972313-24972313 on Build GRCh38
rs187423055 Chr.2: 24972200-24972200 on Build GRCh38
rs915613253 Chr.2: 24972021-24972021 on Build GRCh38
rs929086879 Chr.2: 24972218-24972218 on Build GRCh38
rs1033299736 Chr.2: 24972185-24972185 on Build GRCh38
rs888701354 Chr.2: 24972074-24972074 on Build GRCh38
rs962381602 Chr.2: 24972031-24972031 on Build GRCh38
rs933945608 Chr.2: 24972259-24972259 on Build GRCh38
rs910069516 Chr.2: 24972238-24972238 on Build GRCh38
rs991836619 Chr.2: 24972190-24972190 on Build GRCh38
rs190853585 Chr.2: 24972398-24972398 on Build GRCh38
rs1054284254 Chr.2: 24972295-24972295 on Build GRCh38
rs1036963121 Chr.2: 24972150-24972150 on Build GRCh38
rs998204829 Chr.2: 24972134-24972134 on Build GRCh38
rs776418752 Chr.2: 24972336-24972336 on Build GRCh38
rs1026569231 Chr.2: 24972354-24972354 on Build GRCh38
rs1022800918 Chr.2: 24972323-24972323 on Build GRCh38
rs573931997 Chr.2: 24972281-24972281 on Build GRCh38
rs546931828 Chr.2: 24972081-24972081 on Build GRCh38
rs978387727 Chr.2: 24972064-24972064 on Build GRCh38
rs991170493 Chr.2: 24972017-24972017 on Build GRCh38
rs1032979713 Chr.2: 24972072-24972072 on Build GRCh38
rs185186554 Chr.2: 24972422-24972422 on Build GRCh38
rs1051100338 Chr.2: 24972262-24972262 on Build GRCh38
rs975299240 Chr.2: 24972080-24972080 on Build GRCh38
rs573113068 Chr.2: 24972207-24972207 on Build GRCh38
rs1012714483 Chr.2: 24972315-24972315 on Build GRCh38
rs941604309 Chr.2: 24972240-24972240 on Build GRCh38
rs949172897 Chr.2: 24972461-24972461 on Build GRCh38
rs912231969 Chr.2: 24972120-24972120 on Build GRCh38
rs757641555 Chr.2: 24972252-24972252 on Build GRCh38
rs1029723463 Chr.2: 24972151-24972151 on Build GRCh38
rs575335585 Chr.2: 24972128-24972128 on Build GRCh38
rs931184641 Chr.2: 24972160-24972160 on Build GRCh38
rs180955443 Chr.2: 24972401-24972401 on Build GRCh38
rs959423354 Chr.2: 24972052-24972052 on Build GRCh38
rs562832926 Chr.2: 24972290-24972290 on Build GRCh38
rs202073500 Chr.2: 24972367-24972367 on Build GRCh38
rs1029124898 Chr.2: 24972308-24972308 on Build GRCh38
rs972790585 Chr.2: 24972405-24972405 on Build GRCh38
rs1026432654 Chr.2: 24972469-24972469 on Build GRCh38
rs544629751 Chr.2: 24972289-24972289 on Build GRCh38
rs898392121 Chr.2: 24972153-24972153 on Build GRCh38
rs950982406 Chr.2: 24972369-24972369 on Build GRCh38
rs534868695 Chr.2: 24972038-24972038 on Build GRCh38
rs759500264 Chr.2: 24972047-24972047 on Build GRCh38
rs540493454 Chr.2: 24972228-24972228 on Build GRCh38
rs932395979 Chr.2: 24972087-24972087 on Build GRCh38

View Full Product Details