Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25036260-25036766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NR_034113.1

SNP Detail

SNP ID Chromosome Location
rs921760034 Chr.2: 25036403-25036403 on Build GRCh38
rs948454402 Chr.2: 25036492-25036492 on Build GRCh38
rs60626550 Chr.2: 25036409-25036409 on Build GRCh38
rs996671855 Chr.2: 25036413-25036413 on Build GRCh38
rs987316409 Chr.2: 25036697-25036697 on Build GRCh38
rs989909312 Chr.2: 25036465-25036465 on Build GRCh38
rs1000621465 Chr.2: 25036327-25036327 on Build GRCh38
rs890975663 Chr.2: 25036618-25036618 on Build GRCh38
rs763373655 Chr.2: 25036643-25036643 on Build GRCh38
rs541872793 Chr.2: 25036354-25036354 on Build GRCh38
rs747939268 Chr.2: 25036453-25036453 on Build GRCh38
rs926793362 Chr.2: 25036513-25036513 on Build GRCh38
rs1017461329 Chr.2: 25036370-25036370 on Build GRCh38
rs925441557 Chr.2: 25036660-25036660 on Build GRCh38
rs887127680 Chr.2: 25036337-25036337 on Build GRCh38
rs773183895 Chr.2: 25036564-25036564 on Build GRCh38
rs895977843 Chr.2: 25036572-25036572 on Build GRCh38
rs955257270 Chr.2: 25036422-25036422 on Build GRCh38
rs905408171 Chr.2: 25036287-25036287 on Build GRCh38
rs774086078 Chr.2: 25036410-25036410 on Build GRCh38
rs536180315 Chr.2: 25036351-25036351 on Build GRCh38
rs996892421 Chr.2: 25036576-25036576 on Build GRCh38
rs569357610 Chr.2: 25036698-25036698 on Build GRCh38
rs752085132 Chr.2: 25036729-25036729 on Build GRCh38
rs148259697 Chr.2: 25036548-25036548 on Build GRCh38
rs78822393 Chr.2: 25036532-25036532 on Build GRCh38
rs546368401 Chr.2: 25036415-25036415 on Build GRCh38
rs914364890 Chr.2: 25036476-25036476 on Build GRCh38
rs1045223987 Chr.2: 25036505-25036505 on Build GRCh38
rs141180023 Chr.2: 25036633-25036633 on Build GRCh38
rs954504777 Chr.2: 25036686-25036686 on Build GRCh38
rs753045055 Chr.2: 25036634-25036634 on Build GRCh38
rs1028084439 Chr.2: 25036611-25036611 on Build GRCh38
rs903445779 Chr.2: 25036607-25036607 on Build GRCh38
rs887103564 Chr.2: 25036544-25036544 on Build GRCh38
rs1003805546 Chr.2: 25036313-25036313 on Build GRCh38
rs893588 Chr.2: 25036651-25036651 on Build GRCh38
rs563631872 Chr.2: 25036397-25036397 on Build GRCh38
rs939562195 Chr.2: 25036514-25036514 on Build GRCh38
rs893589 Chr.2: 25036573-25036573 on Build GRCh38
rs770671870 Chr.2: 25036368-25036368 on Build GRCh38
rs765881175 Chr.2: 25036584-25036584 on Build GRCh38
rs1041925943 Chr.2: 25036581-25036581 on Build GRCh38
rs1038449417 Chr.2: 25036549-25036549 on Build GRCh38
rs776311766 Chr.2: 25036620-25036620 on Build GRCh38
rs536640219 Chr.2: 25036717-25036717 on Build GRCh38
rs114784708 Chr.2: 25036459-25036459 on Build GRCh38
rs1021309705 Chr.2: 25036673-25036673 on Build GRCh38
rs565858294 Chr.2: 25036624-25036624 on Build GRCh38
rs568738246 Chr.2: 25036339-25036339 on Build GRCh38
rs1056441205 Chr.2: 25036318-25036318 on Build GRCh38
rs878912532 Chr.2: 25036712-25036712 on Build GRCh38
rs987135327 Chr.2: 25036452-25036452 on Build GRCh38

View Full Product Details