Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25036850-25037357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NR_034113.1

SNP Detail

SNP ID Chromosome Location
rs569598801 Chr.2: 25037119-25037119 on Build GRCh38
rs546206775 Chr.2: 25036964-25036964 on Build GRCh38
rs575038916 Chr.2: 25036922-25036922 on Build GRCh38
rs940285814 Chr.2: 25036879-25036879 on Build GRCh38
rs548117984 Chr.2: 25037106-25037106 on Build GRCh38
rs969737680 Chr.2: 25037101-25037101 on Build GRCh38
rs575750552 Chr.2: 25036959-25036959 on Build GRCh38
rs879235963 Chr.2: 25037123-25037123 on Build GRCh38
rs180710747 Chr.2: 25037147-25037147 on Build GRCh38
rs777466493 Chr.2: 25037204-25037204 on Build GRCh38
rs940236095 Chr.2: 25037161-25037161 on Build GRCh38
rs898514337 Chr.2: 25036909-25036909 on Build GRCh38
rs919909148 Chr.2: 25037172-25037172 on Build GRCh38
rs114044417 Chr.2: 25037214-25037214 on Build GRCh38
rs936211366 Chr.2: 25037224-25037224 on Build GRCh38
rs373372098 Chr.2: 25037129-25037129 on Build GRCh38
rs891225209 Chr.2: 25037065-25037065 on Build GRCh38
rs2196792 Chr.2: 25037084-25037084 on Build GRCh38
rs751550089 Chr.2: 25037035-25037036 on Build GRCh38
rs1008697900 Chr.2: 25037066-25037066 on Build GRCh38
rs1008519155 Chr.2: 25036942-25036942 on Build GRCh38
rs6737356 Chr.2: 25036980-25036980 on Build GRCh38
rs534671060 Chr.2: 25037226-25037226 on Build GRCh38
rs992695270 Chr.2: 25037128-25037128 on Build GRCh38
rs973717461 Chr.2: 25037122-25037122 on Build GRCh38
rs553474020 Chr.2: 25037169-25037169 on Build GRCh38
rs746924963 Chr.2: 25037305-25037305 on Build GRCh38
rs756822491 Chr.2: 25037243-25037243 on Build GRCh38
rs188256699 Chr.2: 25037102-25037102 on Build GRCh38
rs528557568 Chr.2: 25037048-25037048 on Build GRCh38
rs540884750 Chr.2: 25037049-25037049 on Build GRCh38
rs185726074 Chr.2: 25037255-25037255 on Build GRCh38
rs375308426 Chr.2: 25036894-25036894 on Build GRCh38
rs139001924 Chr.2: 25036928-25036928 on Build GRCh38
rs776830835 Chr.2: 25037151-25037151 on Build GRCh38
rs750527183 Chr.2: 25037055-25037055 on Build GRCh38
rs143442351 Chr.2: 25037175-25037175 on Build GRCh38
rs1031507725 Chr.2: 25036975-25036975 on Build GRCh38
rs996869102 Chr.2: 25037032-25037032 on Build GRCh38
rs780247881 Chr.2: 25037093-25037093 on Build GRCh38
rs150748129 Chr.2: 25036923-25036923 on Build GRCh38
rs1042285277 Chr.2: 25036913-25036913 on Build GRCh38
rs1054749095 Chr.2: 25036950-25036950 on Build GRCh38
rs1049791496 Chr.2: 25037181-25037181 on Build GRCh38
rs568685794 Chr.2: 25037299-25037299 on Build GRCh38
rs1055098478 Chr.2: 25037278-25037280 on Build GRCh38

View Full Product Details