Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25038161-25038664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27-AS1
Entrez Gene ID
729723
Gene Name
DNAJC27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.436366
Gene Chromosome Location
Chr.2: 24972112-25039694 on Build GRCh38
Transcripts
NR_034113.1

SNP Detail

SNP ID Chromosome Location
rs940605895 Chr.2: 25038455-25038455 on Build GRCh38
rs958524556 Chr.2: 25038595-25038595 on Build GRCh38
rs978827195 Chr.2: 25038582-25038582 on Build GRCh38
rs917211573 Chr.2: 25038453-25038453 on Build GRCh38
rs1010916685 Chr.2: 25038552-25038552 on Build GRCh38
rs1050161283 Chr.2: 25038495-25038495 on Build GRCh38
rs1055170250 Chr.2: 25038305-25038305 on Build GRCh38
rs6704969 Chr.2: 25038242-25038242 on Build GRCh38
rs546275233 Chr.2: 25038524-25038524 on Build GRCh38
rs898855500 Chr.2: 25038461-25038461 on Build GRCh38
rs541637592 Chr.2: 25038606-25038606 on Build GRCh38
rs927091720 Chr.2: 25038422-25038422 on Build GRCh38
rs151066860 Chr.2: 25038383-25038383 on Build GRCh38
rs1034467463 Chr.2: 25038593-25038593 on Build GRCh38
rs912839366 Chr.2: 25038217-25038217 on Build GRCh38
rs968759269 Chr.2: 25038354-25038354 on Build GRCh38
rs778645801 Chr.2: 25038546-25038546 on Build GRCh38
rs757025185 Chr.2: 25038460-25038460 on Build GRCh38
rs930376619 Chr.2: 25038484-25038484 on Build GRCh38
rs892828362 Chr.2: 25038487-25038487 on Build GRCh38
rs961965930 Chr.2: 25038624-25038624 on Build GRCh38
rs1007542828 Chr.2: 25038246-25038246 on Build GRCh38
rs35282713 Chr.2: 25038640-25038640 on Build GRCh38
rs931410450 Chr.2: 25038292-25038295 on Build GRCh38
rs968727911 Chr.2: 25038573-25038573 on Build GRCh38
rs551170640 Chr.2: 25038184-25038184 on Build GRCh38
rs1019901718 Chr.2: 25038277-25038277 on Build GRCh38
rs140919941 Chr.2: 25038408-25038408 on Build GRCh38
rs994560195 Chr.2: 25038226-25038226 on Build GRCh38
rs965758289 Chr.2: 25038288-25038288 on Build GRCh38
rs555857919 Chr.2: 25038434-25038434 on Build GRCh38
rs775180594 Chr.2: 25038401-25038401 on Build GRCh38
rs954173153 Chr.2: 25038604-25038604 on Build GRCh38
rs937186599 Chr.2: 25038450-25038450 on Build GRCh38
rs1020205633 Chr.2: 25038532-25038532 on Build GRCh38
rs889946338 Chr.2: 25038515-25038515 on Build GRCh38
rs901389527 Chr.2: 25038537-25038537 on Build GRCh38
rs1028102070 Chr.2: 25038566-25038566 on Build GRCh38
rs767043242 Chr.2: 25038307-25038308 on Build GRCh38
rs972414730 Chr.2: 25038633-25038633 on Build GRCh38
rs907125397 Chr.2: 25038343-25038343 on Build GRCh38
rs142848890 Chr.2: 25038338-25038338 on Build GRCh38
rs1011314167 Chr.2: 25038496-25038496 on Build GRCh38
rs769406231 Chr.2: 25038339-25038339 on Build GRCh38
rs1031334276 Chr.2: 25038459-25038459 on Build GRCh38
rs192113790 Chr.2: 25038639-25038639 on Build GRCh38
rs200584122 Chr.2: 25038479-25038481 on Build GRCh38
rs540098426 Chr.2: 25038263-25038263 on Build GRCh38
rs917083725 Chr.2: 25038612-25038612 on Build GRCh38
rs574021284 Chr.2: 25038550-25038550 on Build GRCh38
rs993145404 Chr.2: 25038452-25038452 on Build GRCh38
rs1027086648 Chr.2: 25038237-25038237 on Build GRCh38
rs865872987 Chr.2: 25038241-25038241 on Build GRCh38
rs1020270671 Chr.2: 25038564-25038564 on Build GRCh38

View Full Product Details