Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 145072376-145072908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs989066436 Chr.2: 145072495-145072495 on Build GRCh38
rs925265139 Chr.2: 145072542-145072542 on Build GRCh38
rs1039099155 Chr.2: 145072577-145072577 on Build GRCh38
rs943326393 Chr.2: 145072876-145072876 on Build GRCh38
rs1040113377 Chr.2: 145072575-145072575 on Build GRCh38
rs1051306159 Chr.2: 145072656-145072656 on Build GRCh38
rs574213875 Chr.2: 145072803-145072803 on Build GRCh38
rs969730509 Chr.2: 145072403-145072403 on Build GRCh38
rs890901231 Chr.2: 145072661-145072661 on Build GRCh38
rs1006955736 Chr.2: 145072685-145072685 on Build GRCh38
rs968572789 Chr.2: 145072856-145072856 on Build GRCh38
rs1043538766 Chr.2: 145072527-145072527 on Build GRCh38
rs935037704 Chr.2: 145072604-145072604 on Build GRCh38
rs1006421797 Chr.2: 145072727-145072727 on Build GRCh38
rs190942000 Chr.2: 145072422-145072422 on Build GRCh38
rs943378566 Chr.2: 145072567-145072567 on Build GRCh38
rs1002889902 Chr.2: 145072755-145072755 on Build GRCh38
rs576558387 Chr.2: 145072748-145072748 on Build GRCh38
rs901942696 Chr.2: 145072632-145072632 on Build GRCh38
rs541135091 Chr.2: 145072427-145072427 on Build GRCh38
rs572257020 Chr.2: 145072833-145072833 on Build GRCh38
rs539756285 Chr.2: 145072538-145072538 on Build GRCh38
rs535059193 Chr.2: 145072414-145072414 on Build GRCh38
rs988620360 Chr.2: 145072837-145072837 on Build GRCh38
rs1039406241 Chr.2: 145072689-145072689 on Build GRCh38
rs991895446 Chr.2: 145072778-145072778 on Build GRCh38
rs564444372 Chr.2: 145072862-145072862 on Build GRCh38
rs947286775 Chr.2: 145072506-145072506 on Build GRCh38
rs541402056 Chr.2: 145072806-145072806 on Build GRCh38
rs958851123 Chr.2: 145072774-145072774 on Build GRCh38
rs928399248 Chr.2: 145072467-145072467 on Build GRCh38
rs979810064 Chr.2: 145072463-145072463 on Build GRCh38
rs1048365420 Chr.2: 145072691-145072691 on Build GRCh38
rs746258270 Chr.2: 145072747-145072747 on Build GRCh38
rs1043032347 Chr.2: 145072531-145072531 on Build GRCh38
rs115985042 Chr.2: 145072791-145072791 on Build GRCh38
rs187221012 Chr.2: 145072410-145072410 on Build GRCh38
rs995719333 Chr.2: 145072696-145072696 on Build GRCh38
rs377450735 Chr.2: 145072652-145072652 on Build GRCh38
rs559792282 Chr.2: 145072813-145072813 on Build GRCh38
rs763585860 Chr.2: 145072861-145072861 on Build GRCh38
rs558286457 Chr.2: 145072675-145072675 on Build GRCh38
rs12476923 Chr.2: 145072486-145072486 on Build GRCh38
rs913112210 Chr.2: 145072498-145072498 on Build GRCh38
rs147617647 Chr.2: 145072855-145072855 on Build GRCh38
rs886648380 Chr.2: 145072718-145072718 on Build GRCh38
rs34029033 Chr.2: 145072761-145072761 on Build GRCh38
rs1016500530 Chr.2: 145072735-145072735 on Build GRCh38
rs751365823 Chr.2: 145072425-145072425 on Build GRCh38

View Full Product Details