Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 145072796-145073343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs923258064 Chr.2: 145073309-145073309 on Build GRCh38
rs1020073126 Chr.2: 145073148-145073148 on Build GRCh38
rs1035764360 Chr.2: 145073077-145073077 on Build GRCh38
rs895519588 Chr.2: 145073080-145073080 on Build GRCh38
rs796332933 Chr.2: 145073207-145073207 on Build GRCh38
rs998712378 Chr.2: 145072985-145072985 on Build GRCh38
rs1010391642 Chr.2: 145073117-145073117 on Build GRCh38
rs943326393 Chr.2: 145072876-145072876 on Build GRCh38
rs768036142 Chr.2: 145073210-145073210 on Build GRCh38
rs1037895937 Chr.2: 145073024-145073024 on Build GRCh38
rs751237121 Chr.2: 145072980-145072980 on Build GRCh38
rs530732724 Chr.2: 145073307-145073307 on Build GRCh38
rs968520561 Chr.2: 145073184-145073184 on Build GRCh38
rs182284190 Chr.2: 145073243-145073243 on Build GRCh38
rs923346364 Chr.2: 145072933-145072933 on Build GRCh38
rs780712067 Chr.2: 145073161-145073161 on Build GRCh38
rs968572789 Chr.2: 145072856-145072856 on Build GRCh38
rs569919805 Chr.2: 145073282-145073282 on Build GRCh38
rs886597373 Chr.2: 145072979-145072979 on Build GRCh38
rs397777104 Chr.2: 145073005-145073005 on Build GRCh38
rs995688659 Chr.2: 145072999-145072999 on Build GRCh38
rs561420853 Chr.2: 145073236-145073236 on Build GRCh38
rs1003441768 Chr.2: 145073125-145073125 on Build GRCh38
rs1047842265 Chr.2: 145072973-145072973 on Build GRCh38
rs543238590 Chr.2: 145072971-145072971 on Build GRCh38
rs975160905 Chr.2: 145072895-145072895 on Build GRCh38
rs942504544 Chr.2: 145072891-145072891 on Build GRCh38
rs35500812 Chr.2: 145073004-145073004 on Build GRCh38
rs906140331 Chr.2: 145073060-145073060 on Build GRCh38
rs763585860 Chr.2: 145072861-145072861 on Build GRCh38
rs572257020 Chr.2: 145072833-145072833 on Build GRCh38
rs35834661 Chr.2: 145073219-145073219 on Build GRCh38
rs1001177277 Chr.2: 145073062-145073062 on Build GRCh38
rs1039738482 Chr.2: 145072953-145072953 on Build GRCh38
rs963963989 Chr.2: 145073250-145073250 on Build GRCh38
rs1022012710 Chr.2: 145073197-145073197 on Build GRCh38
rs923840861 Chr.2: 145073213-145073213 on Build GRCh38
rs988620360 Chr.2: 145072837-145072837 on Build GRCh38
rs978609429 Chr.2: 145073190-145073190 on Build GRCh38
rs531387212 Chr.2: 145072911-145072911 on Build GRCh38
rs147617647 Chr.2: 145072855-145072855 on Build GRCh38
rs968724685 Chr.2: 145073201-145073201 on Build GRCh38
rs564444372 Chr.2: 145072862-145072862 on Build GRCh38
rs887385340 Chr.2: 145073043-145073043 on Build GRCh38

View Full Product Details