Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 145073267-145073800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs963435584 Chr.2: 145073658-145073658 on Build GRCh38
rs927456517 Chr.2: 145073673-145073673 on Build GRCh38
rs556012161 Chr.2: 145073682-145073682 on Build GRCh38
rs887356336 Chr.2: 145073530-145073530 on Build GRCh38
rs1018123217 Chr.2: 145073500-145073500 on Build GRCh38
rs942176487 Chr.2: 145073375-145073375 on Build GRCh38
rs907933386 Chr.2: 145073620-145073620 on Build GRCh38
rs530732724 Chr.2: 145073307-145073307 on Build GRCh38
rs78005876 Chr.2: 145073688-145073688 on Build GRCh38
rs975678437 Chr.2: 145073732-145073732 on Build GRCh38
rs558032754 Chr.2: 145073697-145073697 on Build GRCh38
rs550121855 Chr.2: 145073595-145073595 on Build GRCh38
rs919666002 Chr.2: 145073743-145073743 on Build GRCh38
rs998847454 Chr.2: 145073631-145073631 on Build GRCh38
rs1009927009 Chr.2: 145073469-145073469 on Build GRCh38
rs187267562 Chr.2: 145073350-145073350 on Build GRCh38
rs912400903 Chr.2: 145073348-145073348 on Build GRCh38
rs796368223 Chr.2: 145073506-145073506 on Build GRCh38
rs532992256 Chr.2: 145073585-145073585 on Build GRCh38
rs764897517 Chr.2: 145073493-145073493 on Build GRCh38
rs80251109 Chr.2: 145073687-145073687 on Build GRCh38
rs919572199 Chr.2: 145073415-145073415 on Build GRCh38
rs879944491 Chr.2: 145073462-145073462 on Build GRCh38
rs76868621 Chr.2: 145073685-145073685 on Build GRCh38
rs908223801 Chr.2: 145073767-145073767 on Build GRCh38
rs537620870 Chr.2: 145073654-145073654 on Build GRCh38
rs567720977 Chr.2: 145073748-145073748 on Build GRCh38
rs963946079 Chr.2: 145073522-145073522 on Build GRCh38
rs565544696 Chr.2: 145073515-145073515 on Build GRCh38
rs927513371 Chr.2: 145073398-145073398 on Build GRCh38
rs1057474656 Chr.2: 145073584-145073584 on Build GRCh38
rs982372930 Chr.2: 145073764-145073764 on Build GRCh38
rs868564528 Chr.2: 145073664-145073664 on Build GRCh38
rs923258064 Chr.2: 145073309-145073309 on Build GRCh38
rs1057499764 Chr.2: 145073676-145073676 on Build GRCh38
rs1057402407 Chr.2: 145073428-145073428 on Build GRCh38
rs1019837597 Chr.2: 145073703-145073703 on Build GRCh38
rs80251847 Chr.2: 145073606-145073606 on Build GRCh38
rs937551905 Chr.2: 145073674-145073674 on Build GRCh38
rs772938982 Chr.2: 145073494-145073494 on Build GRCh38
rs762746054 Chr.2: 145073499-145073499 on Build GRCh38
rs751441821 Chr.2: 145073610-145073610 on Build GRCh38
rs895456656 Chr.2: 145073442-145073442 on Build GRCh38
rs963932380 Chr.2: 145073728-145073728 on Build GRCh38
rs79052724 Chr.2: 145073686-145073686 on Build GRCh38
rs931100314 Chr.2: 145073755-145073755 on Build GRCh38
rs937585086 Chr.2: 145073421-145073421 on Build GRCh38
rs761337534 Chr.2: 145073321-145073321 on Build GRCh38
rs1031717021 Chr.2: 145073668-145073668 on Build GRCh38
rs986743766 Chr.2: 145073698-145073698 on Build GRCh38
rs983924180 Chr.2: 145073339-145073339 on Build GRCh38
rs938947837 Chr.2: 145073575-145073575 on Build GRCh38
rs1041463950 Chr.2: 145073482-145073482 on Build GRCh38
rs756094449 Chr.2: 145073507-145073507 on Build GRCh38
rs998498061 Chr.2: 145073578-145073578 on Build GRCh38
rs1037843827 Chr.2: 145073377-145073377 on Build GRCh38
rs930963559 Chr.2: 145073440-145073440 on Build GRCh38
rs1039471805 Chr.2: 145073361-145073361 on Build GRCh38

View Full Product Details