Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 145074959-145075506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs974965630 Chr.2: 145075244-145075244 on Build GRCh38
rs113898170 Chr.2: 145075163-145075163 on Build GRCh38
rs997093254 Chr.2: 145075041-145075041 on Build GRCh38
rs1033173791 Chr.2: 145075014-145075014 on Build GRCh38
rs142459288 Chr.2: 145075387-145075387 on Build GRCh38
rs549688719 Chr.2: 145075217-145075217 on Build GRCh38
rs984715567 Chr.2: 145075349-145075349 on Build GRCh38
rs955046461 Chr.2: 145075054-145075054 on Build GRCh38
rs917839192 Chr.2: 145075218-145075218 on Build GRCh38
rs1054572113 Chr.2: 145075396-145075396 on Build GRCh38
rs1045764243 Chr.2: 145075251-145075251 on Build GRCh38
rs893288032 Chr.2: 145075408-145075408 on Build GRCh38
rs930630150 Chr.2: 145075093-145075093 on Build GRCh38
rs1036831795 Chr.2: 145075108-145075108 on Build GRCh38
rs368025624 Chr.2: 145075086-145075086 on Build GRCh38
rs71428050 Chr.2: 145075234-145075234 on Build GRCh38
rs554879980 Chr.2: 145074990-145074990 on Build GRCh38
rs922012633 Chr.2: 145075300-145075300 on Build GRCh38
rs957558031 Chr.2: 145075027-145075027 on Build GRCh38
rs999950574 Chr.2: 145075470-145075470 on Build GRCh38
rs547191225 Chr.2: 145075286-145075286 on Build GRCh38
rs919752818 Chr.2: 145075132-145075132 on Build GRCh38
rs903275285 Chr.2: 145074995-145074995 on Build GRCh38
rs1013981788 Chr.2: 145075139-145075139 on Build GRCh38
rs528671573 Chr.2: 145075283-145075283 on Build GRCh38
rs116444502 Chr.2: 145075435-145075435 on Build GRCh38
rs1049083532 Chr.2: 145075096-145075096 on Build GRCh38
rs956239174 Chr.2: 145075210-145075210 on Build GRCh38
rs1028547913 Chr.2: 145075423-145075423 on Build GRCh38
rs1023119191 Chr.2: 145075151-145075151 on Build GRCh38
rs966590403 Chr.2: 145075239-145075239 on Build GRCh38
rs777198249 Chr.2: 145075053-145075054 on Build GRCh38
rs998983261 Chr.2: 145075009-145075009 on Build GRCh38
rs939941805 Chr.2: 145075099-145075099 on Build GRCh38
rs1032719340 Chr.2: 145075183-145075183 on Build GRCh38
rs765479256 Chr.2: 145075417-145075417 on Build GRCh38
rs1029017979 Chr.2: 145075049-145075049 on Build GRCh38
rs767513943 Chr.2: 145075111-145075111 on Build GRCh38
rs1044273017 Chr.2: 145075455-145075455 on Build GRCh38
rs986887103 Chr.2: 145075078-145075078 on Build GRCh38
rs1007855727 Chr.2: 145075456-145075456 on Build GRCh38
rs937871282 Chr.2: 145075138-145075138 on Build GRCh38
rs961115286 Chr.2: 145075098-145075098 on Build GRCh38
rs956312311 Chr.2: 145075031-145075031 on Build GRCh38
rs975249151 Chr.2: 145075072-145075072 on Build GRCh38
rs997444648 Chr.2: 145075413-145075413 on Build GRCh38
rs957292 Chr.2: 145075168-145075168 on Build GRCh38
rs905642168 Chr.2: 145075164-145075165 on Build GRCh38
rs546624341 Chr.2: 145075396-145075396 on Build GRCh38
rs571613438 Chr.2: 145075341-145075341 on Build GRCh38
rs751314583 Chr.2: 145075220-145075223 on Build GRCh38
rs539135048 Chr.2: 145075380-145075380 on Build GRCh38

View Full Product Details