Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 145075319-145075862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX41
Entrez Gene ID
401014
Gene Name
testis expressed 41 (non-protein coding)
Gene Aliases
DKFZp686O1327, LINC00953
UniGene
Hs.742290
Gene Chromosome Location
Chr.2: 144667967-145076729 on Build GRCh38
Transcripts
NR_033870.2

SNP Detail

SNP ID Chromosome Location
rs796551882 Chr.2: 145075833-145075833 on Build GRCh38
rs553149229 Chr.2: 145075623-145075623 on Build GRCh38
rs891334525 Chr.2: 145075820-145075820 on Build GRCh38
rs116444502 Chr.2: 145075435-145075435 on Build GRCh38
rs961062234 Chr.2: 145075487-145075487 on Build GRCh38
rs867519735 Chr.2: 145075617-145075617 on Build GRCh38
rs966351830 Chr.2: 145075674-145075674 on Build GRCh38
rs1028547913 Chr.2: 145075423-145075423 on Build GRCh38
rs142459288 Chr.2: 145075387-145075387 on Build GRCh38
rs978035337 Chr.2: 145075664-145075664 on Build GRCh38
rs917802154 Chr.2: 145075579-145075579 on Build GRCh38
rs563414037 Chr.2: 145075694-145075694 on Build GRCh38
rs765479256 Chr.2: 145075417-145075417 on Build GRCh38
rs748724521 Chr.2: 145075775-145075775 on Build GRCh38
rs751981646 Chr.2: 145075592-145075592 on Build GRCh38
rs555227120 Chr.2: 145075585-145075585 on Build GRCh38
rs781726896 Chr.2: 145075717-145075717 on Build GRCh38
rs984715567 Chr.2: 145075349-145075349 on Build GRCh38
rs193012270 Chr.2: 145075542-145075542 on Build GRCh38
rs1044273017 Chr.2: 145075455-145075455 on Build GRCh38
rs1054572113 Chr.2: 145075396-145075396 on Build GRCh38
rs991068209 Chr.2: 145075576-145075576 on Build GRCh38
rs545082345 Chr.2: 145075640-145075640 on Build GRCh38
rs1009927030 Chr.2: 145075615-145075615 on Build GRCh38
rs1007855727 Chr.2: 145075456-145075456 on Build GRCh38
rs893288032 Chr.2: 145075408-145075408 on Build GRCh38
rs1015211997 Chr.2: 145075484-145075484 on Build GRCh38
rs185580848 Chr.2: 145075700-145075700 on Build GRCh38
rs758741194 Chr.2: 145075530-145075530 on Build GRCh38
rs577618381 Chr.2: 145075637-145075637 on Build GRCh38
rs924591569 Chr.2: 145075702-145075702 on Build GRCh38
rs917148823 Chr.2: 145075826-145075826 on Build GRCh38
rs992201690 Chr.2: 145075798-145075798 on Build GRCh38
rs997444648 Chr.2: 145075413-145075413 on Build GRCh38
rs534491647 Chr.2: 145075622-145075622 on Build GRCh38
rs957293 Chr.2: 145075620-145075620 on Build GRCh38
rs546624341 Chr.2: 145075396-145075396 on Build GRCh38
rs932803169 Chr.2: 145075788-145075788 on Build GRCh38
rs541073603 Chr.2: 145075619-145075619 on Build GRCh38
rs1050352778 Chr.2: 145075792-145075792 on Build GRCh38
rs1032647162 Chr.2: 145075515-145075515 on Build GRCh38
rs1054522427 Chr.2: 145075768-145075768 on Build GRCh38
rs780247202 Chr.2: 145075570-145075570 on Build GRCh38
rs999950574 Chr.2: 145075470-145075470 on Build GRCh38
rs907857953 Chr.2: 145075782-145075782 on Build GRCh38
rs973780169 Chr.2: 145075535-145075535 on Build GRCh38
rs539135048 Chr.2: 145075380-145075380 on Build GRCh38

View Full Product Details