Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210618071-210618575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPS1
Entrez Gene ID
1373
Gene Name
carbamoyl-phosphate synthase 1
Gene Aliases
CPSASE1, PHN
UniGene
Hs.149252
Gene Chromosome Location
Chr.2: 210477682-210679107 on Build GRCh38
Transcripts
NM_001122633.2 NM_001122634.3 NM_001875.4 XM_011510640.1 XM_011510641.2 XM_011510642.2 XM_011510643.2 XM_011510644.2 NR_002763.2

Gene Symbol
CPS1-IT1
Entrez Gene ID
29034
Gene Name
CPS1 intronic transcript 1
Gene Aliases
CPS1-IT, CPS1IT, CPS1IT1, PRO0132
UniGene
Hs.658398
Gene Chromosome Location
Chr.2: 210617571-210619876 on Build GRCh38
Transcripts
NM_001122633.2 NM_001122634.3 NM_001875.4 XM_011510640.1 XM_011510641.2 XM_011510642.2 XM_011510643.2 XM_011510644.2 NR_002763.2

SNP Detail

SNP ID Chromosome Location
rs747304938 Chr.2: 210618503-210618503 on Build GRCh38
rs553526578 Chr.2: 210618266-210618266 on Build GRCh38
rs189981705 Chr.2: 210618221-210618221 on Build GRCh38
rs181215111 Chr.2: 210618473-210618473 on Build GRCh38
rs934115618 Chr.2: 210618457-210618457 on Build GRCh38
rs540396136 Chr.2: 210618248-210618248 on Build GRCh38
rs892062969 Chr.2: 210618492-210618492 on Build GRCh38
rs150875886 Chr.2: 210618223-210618223 on Build GRCh38
rs558156389 Chr.2: 210618100-210618100 on Build GRCh38
rs35804978 Chr.2: 210618247-210618248 on Build GRCh38
rs1016542330 Chr.2: 210618230-210618230 on Build GRCh38
rs1006158282 Chr.2: 210618229-210618229 on Build GRCh38
rs954824478 Chr.2: 210618461-210618461 on Build GRCh38
rs568744838 Chr.2: 210618192-210618192 on Build GRCh38
rs986581181 Chr.2: 210618463-210618463 on Build GRCh38
rs1009900040 Chr.2: 210618499-210618499 on Build GRCh38
rs573403165 Chr.2: 210618401-210618401 on Build GRCh38
rs980322864 Chr.2: 210618173-210618173 on Build GRCh38
rs1055380862 Chr.2: 210618138-210618138 on Build GRCh38
rs939990696 Chr.2: 210618339-210618339 on Build GRCh38
rs764024971 Chr.2: 210618381-210618381 on Build GRCh38
rs971207174 Chr.2: 210618145-210618145 on Build GRCh38
rs913652486 Chr.2: 210618477-210618477 on Build GRCh38
rs757541880 Chr.2: 210618532-210618532 on Build GRCh38
rs186655963 Chr.2: 210618096-210618096 on Build GRCh38
rs113259035 Chr.2: 210618486-210618486 on Build GRCh38
rs570087335 Chr.2: 210618100-210618103 on Build GRCh38
rs979079459 Chr.2: 210618511-210618511 on Build GRCh38
rs972742209 Chr.2: 210618382-210618382 on Build GRCh38
rs1023998811 Chr.2: 210618500-210618500 on Build GRCh38
rs879936054 Chr.2: 210618475-210618475 on Build GRCh38
rs1009911362 Chr.2: 210618109-210618109 on Build GRCh38
rs922685241 Chr.2: 210618409-210618409 on Build GRCh38
rs974999848 Chr.2: 210618278-210618278 on Build GRCh38
rs1034213582 Chr.2: 210618213-210618213 on Build GRCh38
rs924943813 Chr.2: 210618512-210618512 on Build GRCh38
rs896299335 Chr.2: 210618163-210618163 on Build GRCh38
rs1050336740 Chr.2: 210618464-210618464 on Build GRCh38
rs574577523 Chr.2: 210618224-210618224 on Build GRCh38
rs185488558 Chr.2: 210618533-210618533 on Build GRCh38
rs545503284 Chr.2: 210618458-210618458 on Build GRCh38
rs758784070 Chr.2: 210618242-210618242 on Build GRCh38

View Full Product Details