Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 217283913-217284412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRC3
Entrez Gene ID
729582
Gene Name
disrupted in renal carcinoma 3
Gene Aliases
-
UniGene
Hs.572788
Gene Chromosome Location
Chr.2: 217284023-217756593 on Build GRCh38
Transcripts
NR_026597.1 NR_133642.1

Gene Symbol
DIRC3-AS1
Entrez Gene ID
105373877
Gene Name
DIRC3 antisense RNA 1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 217282733-217344204 on Build GRCh38
Transcripts
NR_026597.1 NR_133642.1

SNP Detail

SNP ID Chromosome Location
rs978608275 Chr.2: 217284168-217284168 on Build GRCh38
rs957982511 Chr.2: 217284294-217284294 on Build GRCh38
rs997233170 Chr.2: 217284137-217284137 on Build GRCh38
rs397787708 Chr.2: 217284177-217284178 on Build GRCh38
rs202233026 Chr.2: 217284174-217284174 on Build GRCh38
rs1042906778 Chr.2: 217283950-217283950 on Build GRCh38
rs57021693 Chr.2: 217284175-217284176 on Build GRCh38
rs937215928 Chr.2: 217284228-217284228 on Build GRCh38
rs148721366 Chr.2: 217284130-217284130 on Build GRCh38
rs1046412995 Chr.2: 217284239-217284239 on Build GRCh38
rs182746720 Chr.2: 217284070-217284070 on Build GRCh38
rs1046817451 Chr.2: 217284060-217284060 on Build GRCh38
rs199875535 Chr.2: 217284175-217284175 on Build GRCh38
rs762180047 Chr.2: 217283991-217283991 on Build GRCh38
rs113470911 Chr.2: 217284043-217284043 on Build GRCh38
rs546833340 Chr.2: 217284347-217284347 on Build GRCh38
rs115170918 Chr.2: 217284243-217284243 on Build GRCh38
rs187714880 Chr.2: 217284103-217284103 on Build GRCh38
rs539381946 Chr.2: 217283942-217283942 on Build GRCh38
rs35338405 Chr.2: 217284134-217284134 on Build GRCh38
rs546153275 Chr.2: 217284234-217284234 on Build GRCh38
rs773492694 Chr.2: 217284087-217284087 on Build GRCh38
rs532400498 Chr.2: 217284374-217284377 on Build GRCh38
rs912858628 Chr.2: 217284161-217284161 on Build GRCh38
rs1027160039 Chr.2: 217284114-217284114 on Build GRCh38
rs3051596 Chr.2: 217284138-217284138 on Build GRCh38
rs1053834689 Chr.2: 217284004-217284004 on Build GRCh38
rs994058862 Chr.2: 217284223-217284223 on Build GRCh38
rs546981510 Chr.2: 217284147-217284147 on Build GRCh38
rs989581598 Chr.2: 217284340-217284340 on Build GRCh38
rs865959861 Chr.2: 217284222-217284222 on Build GRCh38
rs532876738 Chr.2: 217284275-217284275 on Build GRCh38
rs994805033 Chr.2: 217284103-217284103 on Build GRCh38
rs981161472 Chr.2: 217283975-217283975 on Build GRCh38
rs962199477 Chr.2: 217284186-217284186 on Build GRCh38
rs936729147 Chr.2: 217284000-217284000 on Build GRCh38
rs368511338 Chr.2: 217284091-217284091 on Build GRCh38
rs1050508089 Chr.2: 217284141-217284141 on Build GRCh38
rs114060581 Chr.2: 217284249-217284249 on Build GRCh38
rs965481962 Chr.2: 217284150-217284150 on Build GRCh38
rs895234357 Chr.2: 217284040-217284040 on Build GRCh38
rs1041665908 Chr.2: 217284129-217284129 on Build GRCh38
rs561478827 Chr.2: 217283983-217283983 on Build GRCh38
rs12927 Chr.2: 217284160-217284160 on Build GRCh38
rs1055172973 Chr.2: 217284003-217284003 on Build GRCh38
rs925763407 Chr.2: 217283951-217283951 on Build GRCh38
rs949441965 Chr.2: 217283947-217283947 on Build GRCh38
rs1007055622 Chr.2: 217284137-217284138 on Build GRCh38
rs897109978 Chr.2: 217284284-217284284 on Build GRCh38
rs141459646 Chr.2: 217284233-217284233 on Build GRCh38
rs765194743 Chr.2: 217284140-217284140 on Build GRCh38
rs926732855 Chr.2: 217283986-217283986 on Build GRCh38
rs945926336 Chr.2: 217284383-217284383 on Build GRCh38
rs888748566 Chr.2: 217284144-217284144 on Build GRCh38
rs2113824 Chr.2: 217284166-217284166 on Build GRCh38
rs150858495 Chr.2: 217284368-217284368 on Build GRCh38

View Full Product Details