Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 217284132-217284576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRC3
Entrez Gene ID
729582
Gene Name
disrupted in renal carcinoma 3
Gene Aliases
-
UniGene
Hs.572788
Gene Chromosome Location
Chr.2: 217284023-217756593 on Build GRCh38
Transcripts
NR_026597.1 NR_133642.1

Gene Symbol
DIRC3-AS1
Entrez Gene ID
105373877
Gene Name
DIRC3 antisense RNA 1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.2: 217282733-217344204 on Build GRCh38
Transcripts
NR_026597.1 NR_133642.1

SNP Detail

SNP ID Chromosome Location
rs978608275 Chr.2: 217284168-217284168 on Build GRCh38
rs989581598 Chr.2: 217284340-217284340 on Build GRCh38
rs957982511 Chr.2: 217284294-217284294 on Build GRCh38
rs397787708 Chr.2: 217284177-217284178 on Build GRCh38
rs865959861 Chr.2: 217284222-217284222 on Build GRCh38
rs532876738 Chr.2: 217284275-217284275 on Build GRCh38
rs550416543 Chr.2: 217284417-217284417 on Build GRCh38
rs993719770 Chr.2: 217284545-217284545 on Build GRCh38
rs555630502 Chr.2: 217284516-217284516 on Build GRCh38
rs13412814 Chr.2: 217284485-217284485 on Build GRCh38
rs933243304 Chr.2: 217284449-217284449 on Build GRCh38
rs1019949236 Chr.2: 217284483-217284483 on Build GRCh38
rs1002868973 Chr.2: 217284549-217284549 on Build GRCh38
rs202233026 Chr.2: 217284174-217284174 on Build GRCh38
rs139144456 Chr.2: 217284515-217284515 on Build GRCh38
rs1007086992 Chr.2: 217284406-217284406 on Build GRCh38
rs962199477 Chr.2: 217284186-217284186 on Build GRCh38
rs57021693 Chr.2: 217284175-217284176 on Build GRCh38
rs970883831 Chr.2: 217284547-217284547 on Build GRCh38
rs937215928 Chr.2: 217284228-217284228 on Build GRCh38
rs1046412995 Chr.2: 217284239-217284239 on Build GRCh38
rs112626183 Chr.2: 217284550-217284550 on Build GRCh38
rs965168883 Chr.2: 217284452-217284457 on Build GRCh38
rs114060581 Chr.2: 217284249-217284249 on Build GRCh38
rs13412710 Chr.2: 217284436-217284436 on Build GRCh38
rs199875535 Chr.2: 217284175-217284175 on Build GRCh38
rs115170918 Chr.2: 217284243-217284243 on Build GRCh38
rs546833340 Chr.2: 217284347-217284347 on Build GRCh38
rs12927 Chr.2: 217284160-217284160 on Build GRCh38
rs546153275 Chr.2: 217284234-217284234 on Build GRCh38
rs532400498 Chr.2: 217284374-217284377 on Build GRCh38
rs998318021 Chr.2: 217284456-217284456 on Build GRCh38
rs1050266006 Chr.2: 217284480-217284480 on Build GRCh38
rs923249710 Chr.2: 217284445-217284445 on Build GRCh38
rs557695213 Chr.2: 217284486-217284486 on Build GRCh38
rs912858628 Chr.2: 217284161-217284161 on Build GRCh38
rs897109978 Chr.2: 217284284-217284284 on Build GRCh38
rs528988033 Chr.2: 217284411-217284411 on Build GRCh38
rs566811898 Chr.2: 217284502-217284502 on Build GRCh38
rs141459646 Chr.2: 217284233-217284233 on Build GRCh38
rs573865513 Chr.2: 217284546-217284546 on Build GRCh38
rs945926336 Chr.2: 217284383-217284383 on Build GRCh38
rs1037442949 Chr.2: 217284514-217284514 on Build GRCh38
rs2113824 Chr.2: 217284166-217284166 on Build GRCh38
rs994058862 Chr.2: 217284223-217284223 on Build GRCh38
rs150858495 Chr.2: 217284368-217284368 on Build GRCh38

View Full Product Details