Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 217600265-217600776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIRC3
Entrez Gene ID
729582
Gene Name
disrupted in renal carcinoma 3
Gene Aliases
-
UniGene
Hs.572788
Gene Chromosome Location
Chr.2: 217284023-217756593 on Build GRCh38
Transcripts
NR_026597.1

SNP Detail

SNP ID Chromosome Location
rs564333894 Chr.2: 217600384-217600384 on Build GRCh38
rs898830442 Chr.2: 217600343-217600343 on Build GRCh38
rs1053618828 Chr.2: 217600647-217600647 on Build GRCh38
rs769807095 Chr.2: 217600667-217600667 on Build GRCh38
rs1045415130 Chr.2: 217600605-217600605 on Build GRCh38
rs905089795 Chr.2: 217600614-217600614 on Build GRCh38
rs372029467 Chr.2: 217600631-217600631 on Build GRCh38
rs781200963 Chr.2: 217600491-217600491 on Build GRCh38
rs1023471511 Chr.2: 217600305-217600305 on Build GRCh38
rs540027708 Chr.2: 217600573-217600573 on Build GRCh38
rs1009220051 Chr.2: 217600364-217600364 on Build GRCh38
rs1008375165 Chr.2: 217600465-217600465 on Build GRCh38
rs34241787 Chr.2: 217600350-217600351 on Build GRCh38
rs778036184 Chr.2: 217600681-217600681 on Build GRCh38
rs989024493 Chr.2: 217600535-217600535 on Build GRCh38
rs58899448 Chr.2: 217600347-217600347 on Build GRCh38
rs927391559 Chr.2: 217600622-217600622 on Build GRCh38
rs868147540 Chr.2: 217600453-217600453 on Build GRCh38
rs773699684 Chr.2: 217600481-217600488 on Build GRCh38
rs748258472 Chr.2: 217600586-217600586 on Build GRCh38
rs908662272 Chr.2: 217600498-217600498 on Build GRCh38
rs1021592593 Chr.2: 217600565-217600565 on Build GRCh38
rs1019203731 Chr.2: 217600366-217600366 on Build GRCh38
rs1000690593 Chr.2: 217600342-217600342 on Build GRCh38
rs1009230086 Chr.2: 217600703-217600703 on Build GRCh38
rs144429831 Chr.2: 217600594-217600594 on Build GRCh38
rs917216285 Chr.2: 217600590-217600590 on Build GRCh38
rs921457647 Chr.2: 217600460-217600460 on Build GRCh38
rs60796764 Chr.2: 217600299-217600299 on Build GRCh38
rs558128542 Chr.2: 217600575-217600575 on Build GRCh38
rs907729343 Chr.2: 217600712-217600712 on Build GRCh38
rs151215489 Chr.2: 217600639-217600639 on Build GRCh38
rs186880264 Chr.2: 217600407-217600407 on Build GRCh38
rs940079902 Chr.2: 217600560-217600560 on Build GRCh38
rs556334694 Chr.2: 217600351-217600351 on Build GRCh38
rs555321582 Chr.2: 217600735-217600735 on Build GRCh38
rs758501224 Chr.2: 217600526-217600526 on Build GRCh38
rs376696240 Chr.2: 217600716-217600716 on Build GRCh38
rs191775307 Chr.2: 217600456-217600456 on Build GRCh38
rs992630782 Chr.2: 217600654-217600654 on Build GRCh38
rs1057162141 Chr.2: 217600579-217600579 on Build GRCh38
rs904994382 Chr.2: 217600334-217600334 on Build GRCh38
rs953022555 Chr.2: 217600489-217600489 on Build GRCh38

View Full Product Details